Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136942677-136943169 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW5
Entrez Gene ID
54461
Gene Name
F-box and WD repeat domain containing 5
Gene Aliases
Fbw5
UniGene
Hs.522507
Gene Chromosome Location
Chr.9: 136940433-136944774 on Build GRCh38
Transcripts
NM_018998.3 XM_005266089.2 XM_011518789.1 XM_017014812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5420993 c.531C>T p.S177S Chr.9: 136942794-136942794 on Build GRCh38
3216756 c.452C>T p.S151L Chr.9: 136942843-136942843 on Build GRCh38
6662468 c.473T>C p.V158A Chr.9: 136942822-136942822 on Build GRCh38
4784446 c.421G>A p.A141T Chr.9: 136942904-136942904 on Build GRCh38
4784986 c.453C>A p.T151T Chr.9: 136942872-136942872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750768869 Chr.9: 136942699-136942700 on Build GRCh38
rs560242649 Chr.9: 136942955-136942955 on Build GRCh38
rs553877251 Chr.9: 136943056-136943056 on Build GRCh38
rs751799285 Chr.9: 136943006-136943006 on Build GRCh38
rs774807950 Chr.9: 136942925-136942925 on Build GRCh38
rs371500857 Chr.9: 136942903-136942903 on Build GRCh38
rs144989211 Chr.9: 136942725-136942726 on Build GRCh38
rs746089632 Chr.9: 136942742-136942742 on Build GRCh38
rs373825023 Chr.9: 136942753-136942753 on Build GRCh38
rs766954091 Chr.9: 136942757-136942757 on Build GRCh38
rs780236095 Chr.9: 136942810-136942810 on Build GRCh38
rs532239633 Chr.9: 136943146-136943146 on Build GRCh38
rs763943676 Chr.9: 136942841-136942841 on Build GRCh38
rs989409693 Chr.9: 136943131-136943131 on Build GRCh38
rs374021447 Chr.9: 136942924-136942924 on Build GRCh38
rs768929141 Chr.9: 136942787-136942787 on Build GRCh38
rs552411617 Chr.9: 136942964-136942964 on Build GRCh38
rs748605164 Chr.9: 136942819-136942819 on Build GRCh38
rs574859062 Chr.9: 136942793-136942793 on Build GRCh38
rs749499047 Chr.9: 136943021-136943022 on Build GRCh38
rs754369450 Chr.9: 136943019-136943019 on Build GRCh38
rs758547574 Chr.9: 136942914-136942914 on Build GRCh38
rs757957345 Chr.9: 136942944-136942944 on Build GRCh38
rs372187895 Chr.9: 136942709-136942709 on Build GRCh38
rs886831224 Chr.9: 136943064-136943064 on Build GRCh38
rs778808301 Chr.9: 136942910-136942910 on Build GRCh38
rs1002827378 Chr.9: 136943113-136943113 on Build GRCh38
rs752600247 Chr.9: 136942803-136942803 on Build GRCh38
rs766079091 Chr.9: 136942761-136942761 on Build GRCh38
rs546645083 Chr.9: 136942737-136942737 on Build GRCh38
rs749869265 Chr.9: 136942759-136942759 on Build GRCh38
rs1041925451 Chr.9: 136943026-136943026 on Build GRCh38
rs78290834 Chr.9: 136943128-136943128 on Build GRCh38
rs766087360 Chr.9: 136942709-136942710 on Build GRCh38
rs756434861 Chr.9: 136942703-136942704 on Build GRCh38
rs760513187 Chr.9: 136942996-136942996 on Build GRCh38
rs756295513 Chr.9: 136942809-136942809 on Build GRCh38
rs376926032 Chr.9: 136942780-136942780 on Build GRCh38
rs758288123 Chr.9: 136942794-136942795 on Build GRCh38
rs975725477 Chr.9: 136942718-136942718 on Build GRCh38
rs11554213 Chr.9: 136942846-136942846 on Build GRCh38
rs763057345 Chr.9: 136943003-136943003 on Build GRCh38
rs758112282 Chr.9: 136942806-136942806 on Build GRCh38
rs201523922 Chr.9: 136942719-136942719 on Build GRCh38
rs568248938 Chr.9: 136943018-136943018 on Build GRCh38
rs758284886 Chr.9: 136942769-136942769 on Build GRCh38
rs146126767 Chr.9: 136942818-136942818 on Build GRCh38
rs556340512 Chr.9: 136943083-136943083 on Build GRCh38
rs956112971 Chr.9: 136943130-136943130 on Build GRCh38
rs138984211 Chr.9: 136942827-136942827 on Build GRCh38
rs774188507 Chr.9: 136942991-136942991 on Build GRCh38
rs754600763 Chr.9: 136942798-136942798 on Build GRCh38
rs546064512 Chr.9: 136942717-136942717 on Build GRCh38
rs147211191 Chr.9: 136942905-136942905 on Build GRCh38
rs559856592 Chr.9: 136943136-136943136 on Build GRCh38
rs771926213 Chr.9: 136942993-136942993 on Build GRCh38
rs370380736 Chr.9: 136942763-136942763 on Build GRCh38
rs576199707 Chr.9: 136943099-136943099 on Build GRCh38
rs1024709003 Chr.9: 136943074-136943074 on Build GRCh38
rs752266640 Chr.9: 136942832-136942832 on Build GRCh38
rs749403708 Chr.9: 136943115-136943115 on Build GRCh38
rs149461505 Chr.9: 136942904-136942904 on Build GRCh38
rs767390757 Chr.9: 136942950-136942950 on Build GRCh38
rs145866995 Chr.9: 136942774-136942774 on Build GRCh38
rs764563932 Chr.9: 136942725-136942725 on Build GRCh38
rs1020627107 Chr.9: 136942795-136942795 on Build GRCh38
rs750823975 Chr.9: 136943014-136943014 on Build GRCh38
rs74333867 Chr.9: 136943010-136943010 on Build GRCh38
rs918476088 Chr.9: 136942984-136942984 on Build GRCh38
rs377326923 Chr.9: 136942756-136942756 on Build GRCh38
rs770441219 Chr.9: 136942828-136942828 on Build GRCh38
rs746740174 Chr.9: 136943066-136943066 on Build GRCh38
rs768515548 Chr.9: 136942702-136942702 on Build GRCh38
rs368747499 Chr.9: 136942708-136942708 on Build GRCh38
rs369820296 Chr.9: 136942972-136942972 on Build GRCh38
rs374535247 Chr.9: 136942949-136942949 on Build GRCh38
rs546424327 Chr.9: 136942872-136942872 on Build GRCh38
rs538857883 Chr.9: 136942754-136942754 on Build GRCh38
rs766401515 Chr.9: 136942768-136942768 on Build GRCh38
rs770841577 Chr.9: 136942811-136942812 on Build GRCh38
rs201118247 Chr.9: 136942902-136942902 on Build GRCh38
rs778366386 Chr.9: 136942737-136942738 on Build GRCh38
rs1041185700 Chr.9: 136942926-136942926 on Build GRCh38
rs370546816 Chr.9: 136942725-136942731 on Build GRCh38
rs375215463 Chr.9: 136943090-136943090 on Build GRCh38
rs913656355 Chr.9: 136943134-136943141 on Build GRCh38
rs771494126 Chr.9: 136942878-136942878 on Build GRCh38
rs142910179 Chr.9: 136942791-136942791 on Build GRCh38
rs746403703 Chr.9: 136942979-136942979 on Build GRCh38
rs142701655 Chr.9: 136942714-136942715 on Build GRCh38
rs542131101 Chr.9: 136943110-136943110 on Build GRCh38
rs753509107 Chr.9: 136942797-136942797 on Build GRCh38
rs773697937 Chr.9: 136942792-136942792 on Build GRCh38
rs754536040 Chr.9: 136942736-136942737 on Build GRCh38
rs769612103 Chr.9: 136942697-136942697 on Build GRCh38
rs562076005 Chr.9: 136943111-136943111 on Build GRCh38
rs540266891 Chr.9: 136943135-136943135 on Build GRCh38
rs141202282 Chr.9: 136942830-136942830 on Build GRCh38
rs746780893 Chr.9: 136942802-136942802 on Build GRCh38
rs763403695 Chr.9: 136942724-136942724 on Build GRCh38
rs762845142 Chr.9: 136942837-136942837 on Build GRCh38
rs757415833 Chr.9: 136943008-136943008 on Build GRCh38
rs774342063 Chr.9: 136942706-136942706 on Build GRCh38
rs567366453 Chr.9: 136942760-136942760 on Build GRCh38
rs752976346 Chr.9: 136942913-136942913 on Build GRCh38
rs1010472596 Chr.9: 136942932-136942932 on Build GRCh38
rs562733381 Chr.9: 136942895-136942895 on Build GRCh38
rs1025436675 Chr.9: 136943096-136943096 on Build GRCh38
rs375934624 Chr.9: 136942965-136942965 on Build GRCh38
rs778755224 Chr.9: 136942733-136942733 on Build GRCh38
rs73668377 Chr.9: 136943036-136943036 on Build GRCh38
rs758459447 Chr.9: 136942959-136942959 on Build GRCh38
rs199987293 Chr.9: 136942817-136942817 on Build GRCh38
rs577245382 Chr.9: 136942847-136942847 on Build GRCh38
rs757206684 Chr.9: 136942843-136942843 on Build GRCh38
rs1031998434 Chr.9: 136943060-136943060 on Build GRCh38
rs984481748 Chr.9: 136943104-136943104 on Build GRCh38
rs531880221 Chr.9: 136942726-136942726 on Build GRCh38
rs202042512 Chr.9: 136942916-136942916 on Build GRCh38
rs1041873348 Chr.9: 136943145-136943145 on Build GRCh38
rs776523690 Chr.9: 136943000-136943000 on Build GRCh38
rs539507141 Chr.9: 136943068-136943068 on Build GRCh38
rs1020576436 Chr.9: 136942940-136942940 on Build GRCh38
rs748289429 Chr.9: 136942850-136942850 on Build GRCh38
rs1039672317 Chr.9: 136943048-136943048 on Build GRCh38
rs150752994 Chr.9: 136942842-136942842 on Build GRCh38
rs780481327 Chr.9: 136942856-136942857 on Build GRCh38
rs928822281 Chr.9: 136942836-136942836 on Build GRCh38
rs555007815 Chr.9: 136942786-136942786 on Build GRCh38
rs769969613 Chr.9: 136942781-136942781 on Build GRCh38
rs773738626 Chr.9: 136942825-136942825 on Build GRCh38
rs747657165 Chr.9: 136942754-136942755 on Build GRCh38
rs569028461 Chr.9: 136942986-136942986 on Build GRCh38
rs202168534 Chr.9: 136942894-136942894 on Build GRCh38
rs765833265 Chr.9: 136942794-136942794 on Build GRCh38
rs781741068 Chr.9: 136942920-136942920 on Build GRCh38
rs538165049 Chr.9: 136943004-136943004 on Build GRCh38
rs764995479 Chr.9: 136942831-136942831 on Build GRCh38
rs747764299 Chr.9: 136942992-136942992 on Build GRCh38
rs754016834 Chr.9: 136942845-136942845 on Build GRCh38
rs760469417 Chr.9: 136942957-136942957 on Build GRCh38
rs749488509 Chr.9: 136942811-136942811 on Build GRCh38
rs367779676 Chr.9: 136942736-136942736 on Build GRCh38
rs566880015 Chr.9: 136942729-136942729 on Build GRCh38
rs62580408 Chr.9: 136942958-136942958 on Build GRCh38
rs139022111 Chr.9: 136942848-136942848 on Build GRCh38
rs764345523 Chr.9: 136942963-136942963 on Build GRCh38
rs375975356 Chr.9: 136942773-136942773 on Build GRCh38
rs374271255 Chr.9: 136942700-136942700 on Build GRCh38
rs140614835 Chr.9: 136942909-136942909 on Build GRCh38
rs772814123 Chr.9: 136942995-136942995 on Build GRCh38
rs890868807 Chr.9: 136943087-136943087 on Build GRCh38
rs138491092 Chr.9: 136942784-136942784 on Build GRCh38
rs576337189 Chr.9: 136942897-136942897 on Build GRCh38
rs772446117 Chr.9: 136942866-136942866 on Build GRCh38
rs775839145 Chr.9: 136942748-136942748 on Build GRCh38
rs772304052 Chr.9: 136942821-136942821 on Build GRCh38
rs368227017 Chr.9: 136942918-136942918 on Build GRCh38
rs749150527 Chr.9: 136942730-136942730 on Build GRCh38
rs371142063 Chr.9: 136942732-136942732 on Build GRCh38
rs80337213 Chr.9: 136942917-136942917 on Build GRCh38
rs764945619 Chr.9: 136942800-136942800 on Build GRCh38
rs371585295 Chr.9: 136942876-136942876 on Build GRCh38
rs759782408 Chr.9: 136942710-136942710 on Build GRCh38
rs756515070 Chr.9: 136942985-136942985 on Build GRCh38
rs995448306 Chr.9: 136943103-136943103 on Build GRCh38
rs768493165 Chr.9: 136942936-136942936 on Build GRCh38
rs770203825 Chr.9: 136942743-136942743 on Build GRCh38
rs777706097 Chr.9: 136942798-136942800 on Build GRCh38
rs577307420 Chr.9: 136943065-136943065 on Build GRCh38
rs970558782 Chr.9: 136942967-136942967 on Build GRCh38
rs746218548 Chr.9: 136942987-136942987 on Build GRCh38
rs768259879 Chr.9: 136942990-136942990 on Build GRCh38
rs774723521 Chr.9: 136942790-136942790 on Build GRCh38
rs972517763 Chr.9: 136943091-136943091 on Build GRCh38
rs1000862310 Chr.9: 136943052-136943052 on Build GRCh38
rs1005316560 Chr.9: 136943070-136943070 on Build GRCh38
rs746318883 Chr.9: 136942922-136942922 on Build GRCh38
rs756492092 Chr.9: 136943015-136943015 on Build GRCh38
rs779779412 Chr.9: 136942698-136942698 on Build GRCh38
rs566561880 Chr.9: 136942739-136942739 on Build GRCh38
rs368508014 Chr.9: 136942727-136942727 on Build GRCh38
rs374269642 Chr.9: 136942891-136942891 on Build GRCh38
rs376304753 Chr.9: 136942931-136942931 on Build GRCh38
rs898298887 Chr.9: 136943100-136943100 on Build GRCh38
rs754569744 Chr.9: 136943022-136943022 on Build GRCh38

View Full Product Details