Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136943260-136943764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBXW5
Entrez Gene ID
54461
Gene Name
F-box and WD repeat domain containing 5
Gene Aliases
Fbw5
UniGene
Hs.522507
Gene Chromosome Location
Chr.9: 136940433-136944774 on Build GRCh38
Transcripts
NM_018998.3 XM_005266089.2 XM_011518789.1 XM_017014812.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5553726 c.214G>A p.E72K Chr.9: 136943486-136943486 on Build GRCh38
5567200 c.307T>A p.S103T Chr.9: 136943393-136943393 on Build GRCh38
3216768 c.258G>A p.Q86Q Chr.9: 136943442-136943442 on Build GRCh38
3216766 c.270A>G p.E90E Chr.9: 136943430-136943430 on Build GRCh38
4917952 c.224G>A p.R75Q Chr.9: 136943476-136943476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs998791666 Chr.9: 136943667-136943667 on Build GRCh38
rs766711538 Chr.9: 136943552-136943553 on Build GRCh38
rs371391427 Chr.9: 136943575-136943575 on Build GRCh38
rs145714396 Chr.9: 136943665-136943665 on Build GRCh38
rs570838613 Chr.9: 136943526-136943526 on Build GRCh38
rs535344554 Chr.9: 136943591-136943591 on Build GRCh38
rs112789924 Chr.9: 136943344-136943344 on Build GRCh38
rs978671394 Chr.9: 136943707-136943707 on Build GRCh38
rs982819706 Chr.9: 136943611-136943611 on Build GRCh38
rs576213493 Chr.9: 136943658-136943658 on Build GRCh38
rs754704316 Chr.9: 136943542-136943542 on Build GRCh38
rs761086926 Chr.9: 136943537-136943538 on Build GRCh38
rs578244898 Chr.9: 136943633-136943633 on Build GRCh38
rs747203931 Chr.9: 136943454-136943454 on Build GRCh38
rs541203052 Chr.9: 136943680-136943680 on Build GRCh38
rs770652572 Chr.9: 136943378-136943378 on Build GRCh38
rs200112008 Chr.9: 136943364-136943364 on Build GRCh38
rs770295691 Chr.9: 136943514-136943514 on Build GRCh38
rs369599390 Chr.9: 136943340-136943340 on Build GRCh38
rs929560237 Chr.9: 136943610-136943610 on Build GRCh38
rs114180840 Chr.9: 136943496-136943496 on Build GRCh38
rs768734164 Chr.9: 136943328-136943328 on Build GRCh38
rs535518742 Chr.9: 136943549-136943549 on Build GRCh38
rs761896721 Chr.9: 136943326-136943328 on Build GRCh38
rs1026446170 Chr.9: 136943694-136943694 on Build GRCh38
rs773287390 Chr.9: 136943453-136943453 on Build GRCh38
rs377580494 Chr.9: 136943408-136943408 on Build GRCh38
rs760267225 Chr.9: 136943537-136943537 on Build GRCh38
rs780820222 Chr.9: 136943439-136943439 on Build GRCh38
rs749213136 Chr.9: 136943448-136943448 on Build GRCh38
rs546257786 Chr.9: 136943460-136943460 on Build GRCh38
rs200304281 Chr.9: 136943539-136943539 on Build GRCh38
rs201091313 Chr.9: 136943510-136943510 on Build GRCh38
rs776968692 Chr.9: 136943458-136943458 on Build GRCh38
rs555933723 Chr.9: 136943556-136943556 on Build GRCh38
rs927008005 Chr.9: 136943678-136943679 on Build GRCh38
rs909582109 Chr.9: 136943617-136943617 on Build GRCh38
rs906243556 Chr.9: 136943292-136943292 on Build GRCh38
rs572710741 Chr.9: 136943341-136943341 on Build GRCh38
rs573107936 Chr.9: 136943732-136943733 on Build GRCh38
rs750046824 Chr.9: 136943490-136943490 on Build GRCh38
rs531431416 Chr.9: 136943444-136943444 on Build GRCh38
rs569670967 Chr.9: 136943702-136943702 on Build GRCh38
rs774324435 Chr.9: 136943316-136943317 on Build GRCh38
rs574828937 Chr.9: 136943666-136943667 on Build GRCh38
rs375441221 Chr.9: 136943326-136943326 on Build GRCh38
rs775856329 Chr.9: 136943522-136943522 on Build GRCh38
rs112696621 Chr.9: 136943347-136943347 on Build GRCh38
rs775883396 Chr.9: 136943464-136943464 on Build GRCh38
rs781362393 Chr.9: 136943550-136943550 on Build GRCh38
rs775394119 Chr.9: 136943380-136943380 on Build GRCh38
rs558202047 Chr.9: 136943599-136943599 on Build GRCh38
rs369341027 Chr.9: 136943415-136943415 on Build GRCh38
rs542085216 Chr.9: 136943447-136943447 on Build GRCh38
rs771100836 Chr.9: 136943505-136943505 on Build GRCh38
rs773765013 Chr.9: 136943522-136943523 on Build GRCh38
rs944885735 Chr.9: 136943733-136943734 on Build GRCh38
rs1046743187 Chr.9: 136943441-136943441 on Build GRCh38
rs756865141 Chr.9: 136943434-136943434 on Build GRCh38
rs767624995 Chr.9: 136943312-136943312 on Build GRCh38
rs564249055 Chr.9: 136943686-136943686 on Build GRCh38
rs527459357 Chr.9: 136943677-136943677 on Build GRCh38
rs774699686 Chr.9: 136943529-136943529 on Build GRCh38
rs904499481 Chr.9: 136943604-136943604 on Build GRCh38
rs561939155 Chr.9: 136943672-136943672 on Build GRCh38
rs373598007 Chr.9: 136943389-136943389 on Build GRCh38
rs763220161 Chr.9: 136943523-136943523 on Build GRCh38
rs772070924 Chr.9: 136943450-136943450 on Build GRCh38
rs971563701 Chr.9: 136943569-136943569 on Build GRCh38
rs954829477 Chr.9: 136943435-136943435 on Build GRCh38
rs199680044 Chr.9: 136943546-136943546 on Build GRCh38
rs747158780 Chr.9: 136943504-136943504 on Build GRCh38
rs897972308 Chr.9: 136943742-136943742 on Build GRCh38
rs142573296 Chr.9: 136943730-136943730 on Build GRCh38
rs1050934216 Chr.9: 136943649-136943649 on Build GRCh38
rs145188167 Chr.9: 136943501-136943501 on Build GRCh38
rs763323783 Chr.9: 136943391-136943391 on Build GRCh38
rs370810053 Chr.9: 136943521-136943521 on Build GRCh38
rs575667307 Chr.9: 136943291-136943291 on Build GRCh38
rs151282744 Chr.9: 136943427-136943427 on Build GRCh38
rs141420875 Chr.9: 136943476-136943476 on Build GRCh38
rs768848289 Chr.9: 136943411-136943412 on Build GRCh38
rs996382189 Chr.9: 136943410-136943410 on Build GRCh38
rs566114480 Chr.9: 136943578-136943578 on Build GRCh38
rs774349633 Chr.9: 136943330-136943330 on Build GRCh38
rs147821001 Chr.9: 136943355-136943355 on Build GRCh38
rs549775984 Chr.9: 136943691-136943691 on Build GRCh38
rs369034489 Chr.9: 136943506-136943506 on Build GRCh38
rs766975145 Chr.9: 136943531-136943531 on Build GRCh38
rs1009068146 Chr.9: 136943646-136943646 on Build GRCh38
rs747805029 Chr.9: 136943298-136943298 on Build GRCh38
rs556186222 Chr.9: 136943639-136943639 on Build GRCh38
rs372544615 Chr.9: 136943457-136943457 on Build GRCh38
rs765890413 Chr.9: 136943404-136943404 on Build GRCh38
rs970480492 Chr.9: 136943652-136943677 on Build GRCh38
rs182304407 Chr.9: 136943306-136943306 on Build GRCh38
rs866704263 Chr.9: 136943280-136943280 on Build GRCh38
rs951023375 Chr.9: 136943697-136943697 on Build GRCh38
rs201433254 Chr.9: 136943359-136943359 on Build GRCh38
rs771686512 Chr.9: 136943305-136943305 on Build GRCh38
rs142320232 Chr.9: 136943379-136943379 on Build GRCh38
rs1039478794 Chr.9: 136943637-136943637 on Build GRCh38
rs773341364 Chr.9: 136943392-136943392 on Build GRCh38
rs375551647 Chr.9: 136943473-136943473 on Build GRCh38
rs779853817 Chr.9: 136943493-136943493 on Build GRCh38
rs377091309 Chr.9: 136943463-136943463 on Build GRCh38
rs145640873 Chr.9: 136943466-136943466 on Build GRCh38
rs767690357 Chr.9: 136943336-136943336 on Build GRCh38
rs899706301 Chr.9: 136943520-136943520 on Build GRCh38
rs1023552994 Chr.9: 136943565-136943565 on Build GRCh38
rs777200032 Chr.9: 136943398-136943398 on Build GRCh38
rs761949947 Chr.9: 136943335-136943335 on Build GRCh38
rs985097581 Chr.9: 136943717-136943717 on Build GRCh38
rs766865644 Chr.9: 136943397-136943397 on Build GRCh38
rs754880763 Chr.9: 136943498-136943498 on Build GRCh38
rs202161680 Chr.9: 136943406-136943406 on Build GRCh38
rs888213217 Chr.9: 136943324-136943324 on Build GRCh38
rs768895125 Chr.9: 136943315-136943316 on Build GRCh38
rs916126190 Chr.9: 136943282-136943282 on Build GRCh38
rs187429985 Chr.9: 136943690-136943690 on Build GRCh38
rs545862264 Chr.9: 136943638-136943638 on Build GRCh38
rs967395432 Chr.9: 136943374-136943374 on Build GRCh38
rs746091585 Chr.9: 136943552-136943552 on Build GRCh38
rs975497292 Chr.9: 136943740-136943740 on Build GRCh38
rs752638293 Chr.9: 136943548-136943548 on Build GRCh38
rs556201721 Chr.9: 136943285-136943285 on Build GRCh38
rs777330320 Chr.9: 136943377-136943377 on Build GRCh38
rs754555466 Chr.9: 136943553-136943553 on Build GRCh38
rs761354603 Chr.9: 136943530-136943530 on Build GRCh38
rs146624839 Chr.9: 136943475-136943475 on Build GRCh38
rs759419002 Chr.9: 136943327-136943327 on Build GRCh38
rs770728500 Chr.9: 136943317-136943317 on Build GRCh38
rs2271872 Chr.9: 136943538-136943538 on Build GRCh38
rs759963739 Chr.9: 136943401-136943401 on Build GRCh38
rs2271871 Chr.9: 136943596-136943596 on Build GRCh38
rs376636930 Chr.9: 136943519-136943519 on Build GRCh38
rs750102632 Chr.9: 136943534-136943534 on Build GRCh38
rs909519208 Chr.9: 136943726-136943726 on Build GRCh38
rs148467044 Chr.9: 136943376-136943376 on Build GRCh38
rs371064957 Chr.9: 136943472-136943472 on Build GRCh38
rs752309281 Chr.9: 136943375-136943375 on Build GRCh38
rs140432662 Chr.9: 136943440-136943440 on Build GRCh38
rs763610609 Chr.9: 136943414-136943414 on Build GRCh38
rs1036919478 Chr.9: 136943733-136943733 on Build GRCh38

View Full Product Details