Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136992340-136992837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf142
Entrez Gene ID
286257
Gene Name
chromosome 9 open reading frame 142
Gene Aliases
PAXX, XLS
UniGene
Hs.409582
Gene Chromosome Location
Chr.9: 136992418-136993976 on Build GRCh38
Transcripts
NM_183241.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6201291 c.180+10T>G p.? Chr.9: 136992710-136992710 on Build GRCh38
1624765 c.2T>A p.M1K Chr.9: 136992445-136992445 on Build GRCh38
6407219 c.121G>T p.V41L Chr.9: 136992641-136992641 on Build GRCh38
7250486 c.159G>T p.T53T Chr.9: 136992679-136992679 on Build GRCh38
7292080 c.122T>C p.V41A Chr.9: 136992642-136992642 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs866558858 Chr.9: 136992575-136992575 on Build GRCh38
rs749598349 Chr.9: 136992466-136992466 on Build GRCh38
rs1047203292 Chr.9: 136992549-136992549 on Build GRCh38
rs977356580 Chr.9: 136992653-136992653 on Build GRCh38
rs1022110500 Chr.9: 136992488-136992488 on Build GRCh38
rs1053087150 Chr.9: 136992478-136992478 on Build GRCh38
rs919651415 Chr.9: 136992666-136992666 on Build GRCh38
rs368371162 Chr.9: 136992733-136992733 on Build GRCh38
rs771681023 Chr.9: 136992753-136992753 on Build GRCh38
rs943547548 Chr.9: 136992462-136992462 on Build GRCh38
rs990188351 Chr.9: 136992445-136992445 on Build GRCh38
rs1035953648 Chr.9: 136992568-136992568 on Build GRCh38
rs928167095 Chr.9: 136992710-136992710 on Build GRCh38
rs542096667 Chr.9: 136992560-136992560 on Build GRCh38
rs10870159 Chr.9: 136992590-136992590 on Build GRCh38
rs976679572 Chr.9: 136992808-136992808 on Build GRCh38
rs576291300 Chr.9: 136992541-136992541 on Build GRCh38
rs762218332 Chr.9: 136992497-136992497 on Build GRCh38
rs199779601 Chr.9: 136992730-136992730 on Build GRCh38
rs1053432690 Chr.9: 136992760-136992760 on Build GRCh38
rs753583449 Chr.9: 136992677-136992677 on Build GRCh38
rs1035921070 Chr.9: 136992388-136992388 on Build GRCh38
rs535514105 Chr.9: 136992483-136992483 on Build GRCh38
rs766091924 Chr.9: 136992654-136992654 on Build GRCh38
rs1049648845 Chr.9: 136992444-136992444 on Build GRCh38
rs764931530 Chr.9: 136992694-136992694 on Build GRCh38
rs576642531 Chr.9: 136992423-136992423 on Build GRCh38
rs958532892 Chr.9: 136992415-136992415 on Build GRCh38
rs939828016 Chr.9: 136992430-136992430 on Build GRCh38
rs781299945 Chr.9: 136992739-136992739 on Build GRCh38
rs971747741 Chr.9: 136992639-136992639 on Build GRCh38
rs957368654 Chr.9: 136992719-136992719 on Build GRCh38
rs960376105 Chr.9: 136992390-136992390 on Build GRCh38
rs891013038 Chr.9: 136992450-136992450 on Build GRCh38
rs932466668 Chr.9: 136992436-136992436 on Build GRCh38
rs917168762 Chr.9: 136992419-136992419 on Build GRCh38
rs780312291 Chr.9: 136992452-136992452 on Build GRCh38
rs980375442 Chr.9: 136992382-136992382 on Build GRCh38
rs929562259 Chr.9: 136992511-136992511 on Build GRCh38
rs991805198 Chr.9: 136992394-136992394 on Build GRCh38
rs973872086 Chr.9: 136992648-136992648 on Build GRCh38
rs1001676081 Chr.9: 136992491-136992491 on Build GRCh38
rs906049664 Chr.9: 136992489-136992489 on Build GRCh38
rs1021829245 Chr.9: 136992373-136992373 on Build GRCh38
rs771744936 Chr.9: 136992569-136992569 on Build GRCh38
rs961318268 Chr.9: 136992623-136992623 on Build GRCh38
rs187680896 Chr.9: 136992484-136992484 on Build GRCh38
rs970201208 Chr.9: 136992375-136992375 on Build GRCh38
rs974219293 Chr.9: 136992433-136992433 on Build GRCh38
rs772822840 Chr.9: 136992582-136992582 on Build GRCh38
rs894203700 Chr.9: 136992583-136992583 on Build GRCh38
rs545649201 Chr.9: 136992451-136992451 on Build GRCh38
rs759321877 Chr.9: 136992693-136992693 on Build GRCh38
rs750456007 Chr.9: 136992748-136992748 on Build GRCh38
rs991288895 Chr.9: 136992416-136992416 on Build GRCh38
rs913256084 Chr.9: 136992788-136992788 on Build GRCh38
rs919786935 Chr.9: 136992434-136992434 on Build GRCh38
rs774980912 Chr.9: 136992493-136992493 on Build GRCh38
rs891787821 Chr.9: 136992481-136992481 on Build GRCh38
rs541557085 Chr.9: 136992766-136992766 on Build GRCh38
rs1032126725 Chr.9: 136992669-136992669 on Build GRCh38

View Full Product Details