Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136995306-136995803 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLIC3
Entrez Gene ID
9022
Gene Name
chloride intracellular channel 3
Gene Aliases
-
UniGene
Hs.64746
Gene Chromosome Location
Chr.9: 136994608-136997929 on Build GRCh38
Transcripts
NM_004669.2 XM_017015281.1 XM_017015282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1461269 c.254C>T p.T85M Chr.9: 136995457-136995457 on Build GRCh38
7085572 c.192C>T p.I64I Chr.9: 136995519-136995519 on Build GRCh38
7224423 c.234C>A p.I78I Chr.9: 136995477-136995477 on Build GRCh38
1461270 c.176G>A p.G59D Chr.9: 136995535-136995535 on Build GRCh38
7292482 c.220G>A p.D74N Chr.9: 136995491-136995491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1048852238 Chr.9: 136995604-136995604 on Build GRCh38
rs770762522 Chr.9: 136995783-136995783 on Build GRCh38
rs776176446 Chr.9: 136995510-136995511 on Build GRCh38
rs200873514 Chr.9: 136995627-136995627 on Build GRCh38
rs774809536 Chr.9: 136995461-136995461 on Build GRCh38
rs766170206 Chr.9: 136995654-136995654 on Build GRCh38
rs544773888 Chr.9: 136995745-136995745 on Build GRCh38
rs530577870 Chr.9: 136995777-136995777 on Build GRCh38
rs777261805 Chr.9: 136995498-136995498 on Build GRCh38
rs781245274 Chr.9: 136995445-136995445 on Build GRCh38
rs778833124 Chr.9: 136995455-136995455 on Build GRCh38
rs1025971821 Chr.9: 136995762-136995762 on Build GRCh38
rs768602084 Chr.9: 136995342-136995342 on Build GRCh38
rs918924157 Chr.9: 136995532-136995532 on Build GRCh38
rs750689358 Chr.9: 136995671-136995671 on Build GRCh38
rs751362930 Chr.9: 136995437-136995437 on Build GRCh38
rs369332047 Chr.9: 136995626-136995626 on Build GRCh38
rs746331480 Chr.9: 136995504-136995504 on Build GRCh38
rs559155036 Chr.9: 136995634-136995634 on Build GRCh38
rs765382396 Chr.9: 136995481-136995482 on Build GRCh38
rs776993376 Chr.9: 136995444-136995444 on Build GRCh38
rs755954306 Chr.9: 136995567-136995567 on Build GRCh38
rs780316097 Chr.9: 136995337-136995337 on Build GRCh38
rs376321183 Chr.9: 136995679-136995679 on Build GRCh38
rs141765089 Chr.9: 136995653-136995653 on Build GRCh38
rs781390689 Chr.9: 136995623-136995623 on Build GRCh38
rs770755146 Chr.9: 136995728-136995728 on Build GRCh38
rs747551826 Chr.9: 136995774-136995774 on Build GRCh38
rs1012525534 Chr.9: 136995418-136995418 on Build GRCh38
rs951180183 Chr.9: 136995780-136995780 on Build GRCh38
rs771065326 Chr.9: 136995435-136995436 on Build GRCh38
rs774184280 Chr.9: 136995645-136995645 on Build GRCh38
rs76917530 Chr.9: 136995743-136995743 on Build GRCh38
rs774817854 Chr.9: 136995510-136995510 on Build GRCh38
rs377369832 Chr.9: 136995448-136995448 on Build GRCh38
rs909081689 Chr.9: 136995561-136995561 on Build GRCh38
rs747863493 Chr.9: 136995707-136995707 on Build GRCh38
rs767181377 Chr.9: 136995555-136995555 on Build GRCh38
rs571344485 Chr.9: 136995386-136995386 on Build GRCh38
rs930258281 Chr.9: 136995562-136995562 on Build GRCh38
rs764651549 Chr.9: 136995423-136995423 on Build GRCh38
rs142940457 Chr.9: 136995408-136995408 on Build GRCh38
rs950513029 Chr.9: 136995479-136995479 on Build GRCh38
rs758632102 Chr.9: 136995613-136995613 on Build GRCh38
rs199992644 Chr.9: 136995443-136995443 on Build GRCh38
rs374558339 Chr.9: 136995467-136995467 on Build GRCh38
rs13795 Chr.9: 136995677-136995677 on Build GRCh38
rs761532138 Chr.9: 136995650-136995650 on Build GRCh38
rs754416229 Chr.9: 136995768-136995768 on Build GRCh38
rs772540716 Chr.9: 136995662-136995662 on Build GRCh38
rs566659158 Chr.9: 136995447-136995447 on Build GRCh38
rs752705232 Chr.9: 136995581-136995581 on Build GRCh38
rs979483084 Chr.9: 136995402-136995403 on Build GRCh38
rs757374146 Chr.9: 136995670-136995670 on Build GRCh38
rs4880183 Chr.9: 136995392-136995392 on Build GRCh38
rs752435024 Chr.9: 136995425-136995425 on Build GRCh38
rs769396851 Chr.9: 136995506-136995506 on Build GRCh38
rs780540657 Chr.9: 136995761-136995761 on Build GRCh38
rs764183952 Chr.9: 136995736-136995736 on Build GRCh38
rs778027393 Chr.9: 136995619-136995619 on Build GRCh38
rs904914269 Chr.9: 136995615-136995615 on Build GRCh38
rs759751710 Chr.9: 136995459-136995461 on Build GRCh38
rs372161131 Chr.9: 136995711-136995711 on Build GRCh38
rs189896800 Chr.9: 136995387-136995387 on Build GRCh38
rs376619199 Chr.9: 136995431-136995431 on Build GRCh38
rs371956272 Chr.9: 136995352-136995352 on Build GRCh38
rs774305456 Chr.9: 136995537-136995537 on Build GRCh38
rs755500070 Chr.9: 136995769-136995769 on Build GRCh38
rs777682118 Chr.9: 136995328-136995328 on Build GRCh38
rs748754059 Chr.9: 136995635-136995635 on Build GRCh38
rs773892929 Chr.9: 136995676-136995676 on Build GRCh38
rs964498064 Chr.9: 136995588-136995588 on Build GRCh38
rs778515010 Chr.9: 136995686-136995686 on Build GRCh38
rs760444643 Chr.9: 136995725-136995725 on Build GRCh38
rs772471867 Chr.9: 136995403-136995403 on Build GRCh38
rs764610236 Chr.9: 136995650-136995651 on Build GRCh38
rs761762969 Chr.9: 136995539-136995539 on Build GRCh38
rs968022831 Chr.9: 136995694-136995694 on Build GRCh38
rs201523771 Chr.9: 136995663-136995663 on Build GRCh38
rs751600174 Chr.9: 136995329-136995329 on Build GRCh38
rs747229512 Chr.9: 136995621-136995621 on Build GRCh38
rs200303239 Chr.9: 136995638-136995638 on Build GRCh38
rs750318826 Chr.9: 136995446-136995446 on Build GRCh38
rs933344805 Chr.9: 136995651-136995651 on Build GRCh38
rs763777358 Chr.9: 136995536-136995536 on Build GRCh38
rs368301000 Chr.9: 136995773-136995773 on Build GRCh38
rs930310786 Chr.9: 136995781-136995781 on Build GRCh38
rs779257717 Chr.9: 136995391-136995391 on Build GRCh38
rs763398803 Chr.9: 136995542-136995542 on Build GRCh38
rs1036065231 Chr.9: 136995519-136995519 on Build GRCh38
rs759447972 Chr.9: 136995730-136995730 on Build GRCh38
rs551584036 Chr.9: 136995380-136995380 on Build GRCh38
rs559372697 Chr.9: 136995712-136995712 on Build GRCh38
rs886095254 Chr.9: 136995776-136995776 on Build GRCh38
rs968136511 Chr.9: 136995396-136995396 on Build GRCh38
rs750420478 Chr.9: 136995560-136995560 on Build GRCh38
rs200527589 Chr.9: 136995779-136995779 on Build GRCh38
rs749327182 Chr.9: 136995338-136995338 on Build GRCh38
rs776591303 Chr.9: 136995729-136995729 on Build GRCh38
rs756470338 Chr.9: 136995756-136995756 on Build GRCh38
rs765387966 Chr.9: 136995571-136995571 on Build GRCh38
rs750648054 Chr.9: 136995753-136995753 on Build GRCh38
rs762729158 Chr.9: 136995526-136995526 on Build GRCh38
rs202245191 Chr.9: 136995444-136995444 on Build GRCh38
rs375188203 Chr.9: 136995737-136995737 on Build GRCh38
rs984499581 Chr.9: 136995540-136995540 on Build GRCh38
rs373867099 Chr.9: 136995417-136995417 on Build GRCh38
rs757214720 Chr.9: 136995334-136995334 on Build GRCh38
rs560426731 Chr.9: 136995652-136995652 on Build GRCh38
rs370612864 Chr.9: 136995456-136995456 on Build GRCh38
rs758577027 Chr.9: 136995457-136995457 on Build GRCh38
rs1034959369 Chr.9: 136995365-136995365 on Build GRCh38
rs761199590 Chr.9: 136995406-136995406 on Build GRCh38
rs112943493 Chr.9: 136995477-136995477 on Build GRCh38
rs2292923 Chr.9: 136995678-136995678 on Build GRCh38
rs994429299 Chr.9: 136995744-136995744 on Build GRCh38
rs775073323 Chr.9: 136995628-136995628 on Build GRCh38
rs776018612 Chr.9: 136995409-136995409 on Build GRCh38
rs768327294 Chr.9: 136995685-136995685 on Build GRCh38
rs757681804 Chr.9: 136995622-136995622 on Build GRCh38

View Full Product Details