Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137027631-137028122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA2
Entrez Gene ID
20
Gene Name
ATP binding cassette subfamily A member 2
Gene Aliases
ABC2
UniGene
Hs.421202
Gene Chromosome Location
Chr.9: 137007234-137028922 on Build GRCh38
Transcripts
NM_001606.4 NM_212533.2 XM_006716996.3 NM_207511.2

Gene Symbol
C9orf139
Entrez Gene ID
401563
Gene Name
chromosome 9 open reading frame 139
Gene Aliases
-
UniGene
Hs.522509
Gene Chromosome Location
Chr.9: 137027464-137037770 on Build GRCh38
Transcripts
NM_001606.4 NM_212533.2 XM_006716996.3 NM_207511.2

SNP Detail

SNP ID Chromosome Location
rs892711908 Chr.9: 137027723-137027723 on Build GRCh38
rs961982402 Chr.9: 137027787-137027787 on Build GRCh38
rs890194331 Chr.9: 137027970-137027970 on Build GRCh38
rs200808193 Chr.9: 137027913-137027918 on Build GRCh38
rs980814005 Chr.9: 137027883-137027883 on Build GRCh38
rs562894000 Chr.9: 137027727-137027727 on Build GRCh38
rs374198396 Chr.9: 137027715-137027715 on Build GRCh38
rs778516144 Chr.9: 137027650-137027650 on Build GRCh38
rs796609135 Chr.9: 137027771-137027772 on Build GRCh38
rs534452154 Chr.9: 137027829-137027829 on Build GRCh38
rs929198821 Chr.9: 137028011-137028012 on Build GRCh38
rs568416386 Chr.9: 137027806-137027806 on Build GRCh38
rs995853141 Chr.9: 137027758-137027758 on Build GRCh38
rs1052655067 Chr.9: 137027721-137027721 on Build GRCh38
rs531575298 Chr.9: 137027776-137027776 on Build GRCh38
rs533220946 Chr.9: 137027916-137027921 on Build GRCh38
rs371973735 Chr.9: 137027777-137027778 on Build GRCh38
rs954499734 Chr.9: 137027960-137027960 on Build GRCh38
rs548673742 Chr.9: 137027791-137027791 on Build GRCh38
rs969178664 Chr.9: 137028020-137028020 on Build GRCh38
rs900027525 Chr.9: 137027753-137027753 on Build GRCh38
rs928043100 Chr.9: 137027884-137027884 on Build GRCh38
rs1052445627 Chr.9: 137027917-137027917 on Build GRCh38
rs1011615139 Chr.9: 137027732-137027732 on Build GRCh38
rs980593094 Chr.9: 137028022-137028022 on Build GRCh38
rs1014570947 Chr.9: 137027996-137027996 on Build GRCh38
rs958677802 Chr.9: 137027777-137027777 on Build GRCh38
rs879896625 Chr.9: 137027763-137027763 on Build GRCh38
rs1034891417 Chr.9: 137028038-137028038 on Build GRCh38
rs960763577 Chr.9: 137028042-137028042 on Build GRCh38
rs899973646 Chr.9: 137027944-137027945 on Build GRCh38
rs913915151 Chr.9: 137027936-137027936 on Build GRCh38
rs988659486 Chr.9: 137028053-137028053 on Build GRCh38
rs1028944134 Chr.9: 137027934-137027934 on Build GRCh38
rs992723978 Chr.9: 137027825-137027825 on Build GRCh38
rs920414057 Chr.9: 137027815-137027815 on Build GRCh38
rs1000008365 Chr.9: 137027747-137027747 on Build GRCh38
rs748141321 Chr.9: 137027816-137027816 on Build GRCh38
rs1050100532 Chr.9: 137027969-137027969 on Build GRCh38
rs1027988928 Chr.9: 137028018-137028018 on Build GRCh38
rs962029547 Chr.9: 137027998-137027998 on Build GRCh38
rs946976466 Chr.9: 137027937-137027937 on Build GRCh38
rs376662865 Chr.9: 137027764-137027764 on Build GRCh38
rs1015036161 Chr.9: 137027778-137027778 on Build GRCh38
rs996972577 Chr.9: 137027955-137027955 on Build GRCh38
rs913874607 Chr.9: 137028078-137028078 on Build GRCh38
rs1044383048 Chr.9: 137027939-137027939 on Build GRCh38
rs1008638209 Chr.9: 137027985-137027985 on Build GRCh38

View Full Product Details