Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137112593-137113084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPP7
Entrez Gene ID
29952
Gene Name
dipeptidyl peptidase 7
Gene Aliases
DPP2, DPPII, QPP
UniGene
Hs.37916
Gene Chromosome Location
Chr.9: 137110540-137117980 on Build GRCh38
Transcripts
NM_013379.2 XM_005266075.3 XM_006717083.3 XM_011518599.2 XM_011518600.1 XM_017014651.1 XM_017014652.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1193211 c.914G>T p.G305V Chr.9: 137112762-137112762 on Build GRCh38
3088662 c.813G>A p.M271I Chr.9: 137113010-137113010 on Build GRCh38
6421600 c.896A>G p.E299G Chr.9: 137112780-137112780 on Build GRCh38
1250660 c.802G>A p.V268M Chr.9: 137113021-137113021 on Build GRCh38
5979829 c.754G>A p.E252K Chr.9: 137113069-137113069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775876701 Chr.9: 137112827-137112827 on Build GRCh38
rs773920660 Chr.9: 137112835-137112835 on Build GRCh38
rs765630193 Chr.9: 137112727-137112727 on Build GRCh38
rs747158449 Chr.9: 137112726-137112732 on Build GRCh38
rs772726876 Chr.9: 137112931-137112931 on Build GRCh38
rs929044198 Chr.9: 137112741-137112741 on Build GRCh38
rs766298928 Chr.9: 137113018-137113018 on Build GRCh38
rs200976004 Chr.9: 137112695-137112695 on Build GRCh38
rs373456341 Chr.9: 137113039-137113039 on Build GRCh38
rs760226794 Chr.9: 137112984-137112984 on Build GRCh38
rs1018899667 Chr.9: 137112700-137112700 on Build GRCh38
rs755747267 Chr.9: 137112849-137112849 on Build GRCh38
rs781780025 Chr.9: 137112763-137112763 on Build GRCh38
rs776435505 Chr.9: 137112992-137112992 on Build GRCh38
rs748479319 Chr.9: 137113030-137113030 on Build GRCh38
rs760136963 Chr.9: 137112725-137112725 on Build GRCh38
rs372734807 Chr.9: 137112741-137112742 on Build GRCh38
rs769009949 Chr.9: 137112970-137112970 on Build GRCh38
rs766881531 Chr.9: 137112719-137112719 on Build GRCh38
rs761791313 Chr.9: 137112701-137112701 on Build GRCh38
rs879024952 Chr.9: 137113052-137113052 on Build GRCh38
rs774354079 Chr.9: 137113056-137113056 on Build GRCh38
rs760293298 Chr.9: 137112932-137112932 on Build GRCh38
rs764090406 Chr.9: 137113012-137113012 on Build GRCh38
rs770086351 Chr.9: 137112826-137112826 on Build GRCh38
rs774533402 Chr.9: 137112698-137112698 on Build GRCh38
rs536088659 Chr.9: 137112676-137112676 on Build GRCh38
rs373957363 Chr.9: 137112831-137112831 on Build GRCh38
rs754270721 Chr.9: 137112662-137112662 on Build GRCh38
rs368738117 Chr.9: 137112772-137112772 on Build GRCh38
rs753176014 Chr.9: 137113007-137113009 on Build GRCh38
rs892419831 Chr.9: 137112888-137112888 on Build GRCh38
rs751244732 Chr.9: 137112734-137112734 on Build GRCh38
rs199693681 Chr.9: 137112756-137112756 on Build GRCh38
rs765147199 Chr.9: 137113006-137113006 on Build GRCh38
rs534667797 Chr.9: 137112890-137112890 on Build GRCh38
rs749652953 Chr.9: 137112920-137112920 on Build GRCh38
rs755027514 Chr.9: 137112994-137112995 on Build GRCh38
rs140480385 Chr.9: 137112792-137112792 on Build GRCh38
rs779296786 Chr.9: 137113028-137113028 on Build GRCh38
rs780313143 Chr.9: 137112966-137112966 on Build GRCh38
rs554888500 Chr.9: 137112884-137112884 on Build GRCh38
rs377535179 Chr.9: 137112630-137112630 on Build GRCh38
rs750045518 Chr.9: 137112847-137112847 on Build GRCh38
rs147788989 Chr.9: 137112959-137112959 on Build GRCh38
rs775865359 Chr.9: 137112770-137112770 on Build GRCh38
rs751745608 Chr.9: 137112903-137112903 on Build GRCh38
rs75319311 Chr.9: 137113040-137113040 on Build GRCh38
rs151014148 Chr.9: 137112764-137112764 on Build GRCh38
rs764558195 Chr.9: 137112834-137112834 on Build GRCh38
rs747053214 Chr.9: 137112817-137112817 on Build GRCh38
rs781293711 Chr.9: 137112905-137112905 on Build GRCh38
rs75417246 Chr.9: 137112824-137112824 on Build GRCh38
rs139554508 Chr.9: 137112765-137112765 on Build GRCh38
rs1028111169 Chr.9: 137112676-137112681 on Build GRCh38
rs538153439 Chr.9: 137112862-137112862 on Build GRCh38
rs745566509 Chr.9: 137113043-137113043 on Build GRCh38
rs756146666 Chr.9: 137112743-137112743 on Build GRCh38
rs144421503 Chr.9: 137112807-137112807 on Build GRCh38
rs780324880 Chr.9: 137112918-137112918 on Build GRCh38
rs142674168 Chr.9: 137112762-137112762 on Build GRCh38
rs772394261 Chr.9: 137112706-137112706 on Build GRCh38
rs1021763266 Chr.9: 137112666-137112666 on Build GRCh38
rs761611563 Chr.9: 137113062-137113062 on Build GRCh38
rs745622423 Chr.9: 137112742-137112742 on Build GRCh38
rs758886708 Chr.9: 137113034-137113034 on Build GRCh38
rs766021457 Chr.9: 137112933-137112933 on Build GRCh38
rs777198687 Chr.9: 137112732-137112732 on Build GRCh38
rs755141973 Chr.9: 137113022-137113022 on Build GRCh38
rs375048462 Chr.9: 137112823-137112823 on Build GRCh38
rs775495838 Chr.9: 137113045-137113045 on Build GRCh38
rs763743146 Chr.9: 137113051-137113051 on Build GRCh38
rs147834845 Chr.9: 137113036-137113036 on Build GRCh38
rs1040228378 Chr.9: 137112879-137112879 on Build GRCh38
rs753974881 Chr.9: 137112992-137112992 on Build GRCh38
rs371140971 Chr.9: 137112958-137112958 on Build GRCh38
rs771256319 Chr.9: 137112758-137112758 on Build GRCh38
rs749458514 Chr.9: 137112967-137112967 on Build GRCh38
rs746037548 Chr.9: 137112825-137112825 on Build GRCh38
rs752713672 Chr.9: 137112946-137112946 on Build GRCh38
rs762236478 Chr.9: 137112928-137112928 on Build GRCh38
rs200279791 Chr.9: 137112715-137112715 on Build GRCh38
rs765772074 Chr.9: 137113002-137113003 on Build GRCh38
rs766523571 Chr.9: 137112924-137112924 on Build GRCh38
rs903870431 Chr.9: 137112898-137112898 on Build GRCh38
rs887426941 Chr.9: 137112895-137112895 on Build GRCh38
rs371957757 Chr.9: 137112730-137112730 on Build GRCh38
rs35650945 Chr.9: 137112658-137112658 on Build GRCh38
rs905852369 Chr.9: 137112645-137112646 on Build GRCh38
rs760821896 Chr.9: 137112779-137112782 on Build GRCh38
rs756858445 Chr.9: 137112736-137112736 on Build GRCh38
rs760381951 Chr.9: 137112629-137112629 on Build GRCh38
rs779827484 Chr.9: 137112793-137112793 on Build GRCh38
rs759391084 Chr.9: 137112943-137112943 on Build GRCh38
rs375067444 Chr.9: 137112731-137112731 on Build GRCh38
rs753645270 Chr.9: 137112855-137112855 on Build GRCh38
rs753069120 Chr.9: 137112661-137112661 on Build GRCh38
rs111226653 Chr.9: 137113021-137113021 on Build GRCh38
rs759225356 Chr.9: 137113001-137113001 on Build GRCh38
rs779840075 Chr.9: 137112749-137112749 on Build GRCh38
rs908786388 Chr.9: 137112655-137112655 on Build GRCh38
rs546250926 Chr.9: 137113010-137113010 on Build GRCh38
rs748673481 Chr.9: 137112976-137112976 on Build GRCh38
rs762797962 Chr.9: 137113049-137113049 on Build GRCh38
rs144328006 Chr.9: 137112841-137112841 on Build GRCh38
rs769541149 Chr.9: 137113044-137113044 on Build GRCh38
rs760910172 Chr.9: 137112718-137112718 on Build GRCh38
rs200537558 Chr.9: 137112930-137112930 on Build GRCh38
rs770719144 Chr.9: 137112675-137112675 on Build GRCh38
rs562697255 Chr.9: 137112669-137112669 on Build GRCh38
rs772606508 Chr.9: 137112979-137112979 on Build GRCh38
rs369979070 Chr.9: 137112989-137112989 on Build GRCh38
rs757418683 Chr.9: 137112904-137112904 on Build GRCh38
rs766106562 Chr.9: 137112854-137112854 on Build GRCh38
rs968041628 Chr.9: 137112759-137112759 on Build GRCh38
rs750530772 Chr.9: 137113013-137113013 on Build GRCh38
rs763027764 Chr.9: 137113011-137113011 on Build GRCh38
rs758892447 Chr.9: 137113035-137113035 on Build GRCh38
rs1034204289 Chr.9: 137112912-137112912 on Build GRCh38
rs767155285 Chr.9: 137112791-137112791 on Build GRCh38
rs746102370 Chr.9: 137112917-137112917 on Build GRCh38
rs778010133 Chr.9: 137113054-137113054 on Build GRCh38
rs751050637 Chr.9: 137112773-137112773 on Build GRCh38
rs372674817 Chr.9: 137112795-137112795 on Build GRCh38
rs756946294 Chr.9: 137112774-137112774 on Build GRCh38
rs979989431 Chr.9: 137112883-137112883 on Build GRCh38
rs199608486 Chr.9: 137112945-137112945 on Build GRCh38
rs771308357 Chr.9: 137112738-137112738 on Build GRCh38
rs766952839 Chr.9: 137112845-137112845 on Build GRCh38
rs772636240 Chr.9: 137112983-137112983 on Build GRCh38
rs764537384 Chr.9: 137112942-137112942 on Build GRCh38
rs746395277 Chr.9: 137113055-137113055 on Build GRCh38
rs919728148 Chr.9: 137112716-137112716 on Build GRCh38
rs773381853 Chr.9: 137112757-137112757 on Build GRCh38
rs181036640 Chr.9: 137113014-137113014 on Build GRCh38
rs769212647 Chr.9: 137112927-137112927 on Build GRCh38
rs1015724003 Chr.9: 137112939-137112939 on Build GRCh38
rs548442146 Chr.9: 137112679-137112679 on Build GRCh38
rs867564714 Chr.9: 137112973-137112973 on Build GRCh38
rs184507405 Chr.9: 137112654-137112654 on Build GRCh38
rs779204928 Chr.9: 137112972-137112972 on Build GRCh38
rs755006035 Chr.9: 137112856-137112856 on Build GRCh38
rs369048728 Chr.9: 137112842-137112842 on Build GRCh38
rs1049927664 Chr.9: 137112816-137112816 on Build GRCh38

View Full Product Details