Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137255811-137256304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELFB
Entrez Gene ID
25920
Gene Name
negative elongation factor complex member B
Gene Aliases
COBRA1, NELF-B
UniGene
Hs.521809
Gene Chromosome Location
Chr.9: 137255300-137273548 on Build GRCh38
Transcripts
NM_015456.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4163637 c.220C>T p.L74L Chr.9: 137256024-137256024 on Build GRCh38
753042 c.251G>A p.G84E Chr.9: 137256055-137256055 on Build GRCh38
1756068 c.128C>T p.S43F Chr.9: 137255932-137255932 on Build GRCh38
5885808 c.103-9delC p.? Chr.9: 137255898-137255898 on Build GRCh38
6581216 c.103-3C>G p.? Chr.9: 137255904-137255904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1017145380 Chr.9: 137256227-137256227 on Build GRCh38
rs914751461 Chr.9: 137256113-137256113 on Build GRCh38
rs934840947 Chr.9: 137255833-137255833 on Build GRCh38
rs200129862 Chr.9: 137255896-137255896 on Build GRCh38
rs1056069186 Chr.9: 137256205-137256205 on Build GRCh38
rs754967234 Chr.9: 137255927-137255927 on Build GRCh38
rs182428492 Chr.9: 137255901-137255901 on Build GRCh38
rs763514372 Chr.9: 137255926-137255926 on Build GRCh38
rs1053143170 Chr.9: 137256137-137256137 on Build GRCh38
rs11533155 Chr.9: 137255858-137255858 on Build GRCh38
rs761405414 Chr.9: 137255872-137255872 on Build GRCh38
rs973949441 Chr.9: 137256073-137256073 on Build GRCh38
rs759501533 Chr.9: 137256049-137256049 on Build GRCh38
rs900703618 Chr.9: 137256189-137256189 on Build GRCh38
rs370208665 Chr.9: 137255951-137255951 on Build GRCh38
rs544817077 Chr.9: 137255829-137255829 on Build GRCh38
rs757816908 Chr.9: 137256110-137256110 on Build GRCh38
rs749738091 Chr.9: 137255984-137255984 on Build GRCh38
rs749348133 Chr.9: 137256084-137256084 on Build GRCh38
rs758320426 Chr.9: 137255934-137255934 on Build GRCh38
rs760254481 Chr.9: 137256020-137256020 on Build GRCh38
rs997628738 Chr.9: 137256217-137256217 on Build GRCh38
rs915810772 Chr.9: 137256276-137256276 on Build GRCh38
rs750601280 Chr.9: 137256060-137256060 on Build GRCh38
rs141153620 Chr.9: 137255975-137255975 on Build GRCh38
rs571323342 Chr.9: 137256150-137256150 on Build GRCh38
rs752724218 Chr.9: 137256054-137256054 on Build GRCh38
rs371374510 Chr.9: 137255863-137255863 on Build GRCh38
rs975173379 Chr.9: 137255928-137255928 on Build GRCh38
rs893092929 Chr.9: 137256138-137256138 on Build GRCh38
rs374064545 Chr.9: 137256103-137256103 on Build GRCh38
rs776164150 Chr.9: 137255909-137255910 on Build GRCh38
rs377724847 Chr.9: 137255892-137255892 on Build GRCh38
rs780132790 Chr.9: 137256065-137256065 on Build GRCh38
rs143781103 Chr.9: 137256062-137256062 on Build GRCh38
rs755088818 Chr.9: 137256085-137256085 on Build GRCh38
rs530611986 Chr.9: 137256187-137256187 on Build GRCh38
rs962608814 Chr.9: 137256236-137256236 on Build GRCh38
rs752566675 Chr.9: 137255932-137255932 on Build GRCh38
rs962682168 Chr.9: 137256260-137256260 on Build GRCh38
rs146836979 Chr.9: 137255961-137255961 on Build GRCh38
rs763854337 Chr.9: 137256059-137256059 on Build GRCh38
rs887001744 Chr.9: 137256215-137256215 on Build GRCh38
rs765245757 Chr.9: 137256050-137256050 on Build GRCh38
rs1032368452 Chr.9: 137255844-137255844 on Build GRCh38
rs763427607 Chr.9: 137255861-137255861 on Build GRCh38
rs540900399 Chr.9: 137256002-137256002 on Build GRCh38
rs987033312 Chr.9: 137256047-137256047 on Build GRCh38
rs1034922907 Chr.9: 137255949-137255949 on Build GRCh38
rs756200034 Chr.9: 137256255-137256255 on Build GRCh38
rs924887419 Chr.9: 137256083-137256083 on Build GRCh38
rs774727395 Chr.9: 137255867-137255867 on Build GRCh38
rs1044858702 Chr.9: 137256132-137256132 on Build GRCh38
rs1004087794 Chr.9: 137256224-137256224 on Build GRCh38
rs955441690 Chr.9: 137256036-137256036 on Build GRCh38
rs781536430 Chr.9: 137255941-137255941 on Build GRCh38
rs904888610 Chr.9: 137255898-137255898 on Build GRCh38
rs879085918 Chr.9: 137256242-137256242 on Build GRCh38
rs974023095 Chr.9: 137256267-137256267 on Build GRCh38
rs143088410 Chr.9: 137255909-137255909 on Build GRCh38
rs201110859 Chr.9: 137256100-137256100 on Build GRCh38
rs373011909 Chr.9: 137255995-137255995 on Build GRCh38
rs776039069 Chr.9: 137256046-137256046 on Build GRCh38
rs201644989 Chr.9: 137255954-137255954 on Build GRCh38
rs772903966 Chr.9: 137256014-137256014 on Build GRCh38
rs780011873 Chr.9: 137255980-137255980 on Build GRCh38
rs374548242 Chr.9: 137255857-137255857 on Build GRCh38
rs758265148 Chr.9: 137256056-137256056 on Build GRCh38
rs78469234 Chr.9: 137255836-137255836 on Build GRCh38
rs150365696 Chr.9: 137255996-137255996 on Build GRCh38
rs141479053 Chr.9: 137256147-137256147 on Build GRCh38
rs745983012 Chr.9: 137256228-137256228 on Build GRCh38
rs139208158 Chr.9: 137256024-137256024 on Build GRCh38
rs769146711 Chr.9: 137256066-137256067 on Build GRCh38
rs765088216 Chr.9: 137255929-137255929 on Build GRCh38
rs377263698 Chr.9: 137256118-137256118 on Build GRCh38
rs767676021 Chr.9: 137256229-137256229 on Build GRCh38
rs750081628 Chr.9: 137255887-137255887 on Build GRCh38
rs1028431846 Chr.9: 137256270-137256270 on Build GRCh38
rs571518027 Chr.9: 137256122-137256122 on Build GRCh38
rs748479169 Chr.9: 137255999-137255999 on Build GRCh38
rs767132619 Chr.9: 137255876-137255876 on Build GRCh38
rs781659478 Chr.9: 137256182-137256182 on Build GRCh38
rs370265308 Chr.9: 137255831-137255831 on Build GRCh38
rs774798184 Chr.9: 137256105-137256111 on Build GRCh38

View Full Product Details