Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137266962-137267489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELFB
Entrez Gene ID
25920
Gene Name
negative elongation factor complex member B
Gene Aliases
COBRA1, NELF-B
UniGene
Hs.521809
Gene Chromosome Location
Chr.9: 137255300-137273548 on Build GRCh38
Transcripts
NM_015456.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1107099 c.1201G>T p.V401F Chr.9: 137267049-137267049 on Build GRCh38
3089157 c.1206A>G p.S402S Chr.9: 137267054-137267054 on Build GRCh38
3089159 c.1255C>T p.R419C Chr.9: 137267256-137267256 on Build GRCh38
6244348 c.1285G>T p.V429F Chr.9: 137267286-137267286 on Build GRCh38
1624791 c.1203C>T p.V401V Chr.9: 137267051-137267051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376239133 Chr.9: 137267178-137267178 on Build GRCh38
rs758099741 Chr.9: 137267121-137267121 on Build GRCh38
rs201404739 Chr.9: 137267339-137267339 on Build GRCh38
rs932886618 Chr.9: 137267391-137267391 on Build GRCh38
rs376605112 Chr.9: 137267324-137267324 on Build GRCh38
rs749624326 Chr.9: 137267015-137267015 on Build GRCh38
rs752588264 Chr.9: 137267228-137267228 on Build GRCh38
rs750105991 Chr.9: 137267388-137267388 on Build GRCh38
rs766447333 Chr.9: 137267202-137267202 on Build GRCh38
rs765874438 Chr.9: 137267105-137267121 on Build GRCh38
rs375292360 Chr.9: 137267106-137267106 on Build GRCh38
rs780787107 Chr.9: 137267236-137267236 on Build GRCh38
rs767700674 Chr.9: 137267137-137267137 on Build GRCh38
rs143716036 Chr.9: 137267029-137267029 on Build GRCh38
rs758151464 Chr.9: 137267313-137267315 on Build GRCh38
rs33923321 Chr.9: 137267212-137267222 on Build GRCh38
rs11531963 Chr.9: 137267282-137267282 on Build GRCh38
rs764505828 Chr.9: 137267374-137267374 on Build GRCh38
rs758899047 Chr.9: 137267228-137267228 on Build GRCh38
rs765131512 Chr.9: 137267199-137267200 on Build GRCh38
rs987826905 Chr.9: 137267070-137267070 on Build GRCh38
rs763585545 Chr.9: 137267368-137267368 on Build GRCh38
rs1034231023 Chr.9: 137267147-137267147 on Build GRCh38
rs753072520 Chr.9: 137267227-137267227 on Build GRCh38
rs200489694 Chr.9: 137267206-137267206 on Build GRCh38
rs775380380 Chr.9: 137267130-137267130 on Build GRCh38
rs747704646 Chr.9: 137267120-137267120 on Build GRCh38
rs199828573 Chr.9: 137267285-137267285 on Build GRCh38
rs749919876 Chr.9: 137267327-137267327 on Build GRCh38
rs754256529 Chr.9: 137267216-137267216 on Build GRCh38
rs375169738 Chr.9: 137267221-137267221 on Build GRCh38
rs925998277 Chr.9: 137267253-137267253 on Build GRCh38
rs1038388000 Chr.9: 137267225-137267225 on Build GRCh38
rs373004598 Chr.9: 137267126-137267126 on Build GRCh38
rs186762786 Chr.9: 137267397-137267397 on Build GRCh38
rs745745289 Chr.9: 137267239-137267239 on Build GRCh38
rs896933400 Chr.9: 137267084-137267084 on Build GRCh38
rs776748331 Chr.9: 137267292-137267292 on Build GRCh38
rs768500766 Chr.9: 137267261-137267261 on Build GRCh38
rs374699224 Chr.9: 137267190-137267190 on Build GRCh38
rs750484303 Chr.9: 137267088-137267088 on Build GRCh38
rs780589463 Chr.9: 137267012-137267012 on Build GRCh38
rs767197101 Chr.9: 137267386-137267386 on Build GRCh38
rs77836500 Chr.9: 137267030-137267030 on Build GRCh38
rs1020565570 Chr.9: 137267431-137267431 on Build GRCh38
rs550728380 Chr.9: 137267180-137267201 on Build GRCh38
rs780031795 Chr.9: 137267332-137267332 on Build GRCh38
rs908712144 Chr.9: 137267461-137267461 on Build GRCh38
rs770624719 Chr.9: 137267123-137267123 on Build GRCh38
rs543108147 Chr.9: 137267190-137267195 on Build GRCh38
rs138777545 Chr.9: 137267346-137267346 on Build GRCh38
rs78649617 Chr.9: 137267074-137267074 on Build GRCh38
rs774371265 Chr.9: 137267073-137267073 on Build GRCh38
rs751951501 Chr.9: 137267381-137267381 on Build GRCh38
rs544060553 Chr.9: 137267183-137267187 on Build GRCh38
rs377190551 Chr.9: 137267212-137267213 on Build GRCh38
rs772869877 Chr.9: 137267051-137267051 on Build GRCh38
rs779362092 Chr.9: 137267105-137267105 on Build GRCh38
rs199634156 Chr.9: 137267125-137267125 on Build GRCh38
rs993102900 Chr.9: 137267176-137267176 on Build GRCh38
rs775999597 Chr.9: 137267364-137267364 on Build GRCh38
rs112225955 Chr.9: 137267172-137267172 on Build GRCh38
rs767539647 Chr.9: 137267077-137267077 on Build GRCh38
rs375155339 Chr.9: 137267210-137267211 on Build GRCh38
rs545057518 Chr.9: 137267191-137267201 on Build GRCh38
rs765711879 Chr.9: 137267226-137267226 on Build GRCh38
rs376711918 Chr.9: 137267090-137267090 on Build GRCh38
rs760331280 Chr.9: 137267005-137267006 on Build GRCh38
rs887194420 Chr.9: 137267212-137267212 on Build GRCh38
rs200528050 Chr.9: 137267235-137267235 on Build GRCh38
rs376737086 Chr.9: 137267337-137267337 on Build GRCh38
rs762708328 Chr.9: 137267132-137267132 on Build GRCh38
rs146930259 Chr.9: 137266994-137266994 on Build GRCh38
rs751219523 Chr.9: 137267320-137267320 on Build GRCh38
rs3936177 Chr.9: 137267184-137267184 on Build GRCh38
rs552363247 Chr.9: 137267072-137267072 on Build GRCh38
rs753662525 Chr.9: 137266989-137266989 on Build GRCh38
rs1008357334 Chr.9: 137267275-137267275 on Build GRCh38
rs759573717 Chr.9: 137267295-137267295 on Build GRCh38
rs759512202 Chr.9: 137267184-137267195 on Build GRCh38
rs9802698 Chr.9: 137267457-137267457 on Build GRCh38
rs755226429 Chr.9: 137267102-137267102 on Build GRCh38
rs1026634211 Chr.9: 137267091-137267091 on Build GRCh38
rs372893987 Chr.9: 137267286-137267286 on Build GRCh38
rs374507529 Chr.9: 137267396-137267396 on Build GRCh38
rs760686506 Chr.9: 137267193-137267193 on Build GRCh38
rs141797959 Chr.9: 137267180-137267190 on Build GRCh38
rs746566888 Chr.9: 137267052-137267052 on Build GRCh38
rs370010361 Chr.9: 137267101-137267101 on Build GRCh38
rs751868850 Chr.9: 137266996-137266996 on Build GRCh38
rs777585712 Chr.9: 137267345-137267345 on Build GRCh38
rs1018609523 Chr.9: 137267278-137267278 on Build GRCh38
rs137937150 Chr.9: 137267004-137267004 on Build GRCh38
rs751060036 Chr.9: 137267011-137267011 on Build GRCh38
rs769776598 Chr.9: 137267363-137267363 on Build GRCh38
rs746513563 Chr.9: 137267122-137267122 on Build GRCh38
rs370204762 Chr.9: 137267362-137267362 on Build GRCh38
rs9802270 Chr.9: 137267419-137267419 on Build GRCh38
rs766870901 Chr.9: 137267095-137267095 on Build GRCh38
rs759539875 Chr.9: 137267190-137267206 on Build GRCh38
rs777553601 Chr.9: 137267231-137267231 on Build GRCh38
rs762311059 Chr.9: 137267382-137267382 on Build GRCh38
rs369149566 Chr.9: 137267257-137267257 on Build GRCh38
rs771439450 Chr.9: 137267048-137267048 on Build GRCh38
rs554518196 Chr.9: 137267256-137267256 on Build GRCh38
rs757226119 Chr.9: 137267003-137267003 on Build GRCh38
rs756994375 Chr.9: 137267321-137267321 on Build GRCh38
rs891379882 Chr.9: 137267408-137267408 on Build GRCh38
rs759414082 Chr.9: 137266990-137266990 on Build GRCh38
rs746673399 Chr.9: 137267233-137267233 on Build GRCh38
rs776783803 Chr.9: 137267358-137267358 on Build GRCh38
rs747982652 Chr.9: 137267340-137267340 on Build GRCh38
rs1009618275 Chr.9: 137267289-137267289 on Build GRCh38
rs371489270 Chr.9: 137267059-137267059 on Build GRCh38
rs913061152 Chr.9: 137267183-137267183 on Build GRCh38
rs115604241 Chr.9: 137267195-137267195 on Build GRCh38
rs773352529 Chr.9: 137267267-137267267 on Build GRCh38
rs759240621 Chr.9: 137267067-137267067 on Build GRCh38
rs765769006 Chr.9: 137267318-137267318 on Build GRCh38
rs776774439 Chr.9: 137267063-137267063 on Build GRCh38
rs146204017 Chr.9: 137267277-137267277 on Build GRCh38
rs867625403 Chr.9: 137267425-137267425 on Build GRCh38
rs376425261 Chr.9: 137267010-137267010 on Build GRCh38
rs755651433 Chr.9: 137267328-137267328 on Build GRCh38
rs540023967 Chr.9: 137267347-137267347 on Build GRCh38

View Full Product Details