Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124878207-124878701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA1
Entrez Gene ID
2800
Gene Name
golgin A1
Gene Aliases
golgin-97
UniGene
Hs.133469
Gene Chromosome Location
Chr.9: 124878294-124941114 on Build GRCh38
Transcripts
NM_002077.3 XM_005251929.3 XM_006717062.3 XM_006717063.3

SNP Detail

SNP ID Chromosome Location
rs769124310 Chr.9: 124878409-124878409 on Build GRCh38
rs965005056 Chr.9: 124878243-124878243 on Build GRCh38
rs574766493 Chr.9: 124878481-124878481 on Build GRCh38
rs752059747 Chr.9: 124878561-124878561 on Build GRCh38
rs532548983 Chr.9: 124878664-124878664 on Build GRCh38
rs947286765 Chr.9: 124878251-124878251 on Build GRCh38
rs77474434 Chr.9: 124878475-124878475 on Build GRCh38
rs992683537 Chr.9: 124878550-124878550 on Build GRCh38
rs1028002983 Chr.9: 124878590-124878590 on Build GRCh38
rs958778126 Chr.9: 124878525-124878525 on Build GRCh38
rs942201627 Chr.9: 124878385-124878385 on Build GRCh38
rs1020689024 Chr.9: 124878233-124878233 on Build GRCh38
rs941054635 Chr.9: 124878674-124878674 on Build GRCh38
rs750752473 Chr.9: 124878486-124878486 on Build GRCh38
rs574529057 Chr.9: 124878584-124878584 on Build GRCh38
rs548421524 Chr.9: 124878301-124878301 on Build GRCh38
rs1037914172 Chr.9: 124878681-124878681 on Build GRCh38
rs997774385 Chr.9: 124878515-124878515 on Build GRCh38
rs978565147 Chr.9: 124878265-124878265 on Build GRCh38
rs986676479 Chr.9: 124878302-124878302 on Build GRCh38
rs965103383 Chr.9: 124878514-124878514 on Build GRCh38
rs909401941 Chr.9: 124878345-124878345 on Build GRCh38
rs953735159 Chr.9: 124878621-124878621 on Build GRCh38
rs897948467 Chr.9: 124878404-124878404 on Build GRCh38
rs545836405 Chr.9: 124878665-124878665 on Build GRCh38
rs996398361 Chr.9: 124878451-124878451 on Build GRCh38
rs565039053 Chr.9: 124878673-124878673 on Build GRCh38
rs779253872 Chr.9: 124878325-124878326 on Build GRCh38
rs916948079 Chr.9: 124878586-124878586 on Build GRCh38
rs138401811 Chr.9: 124878312-124878312 on Build GRCh38
rs569653774 Chr.9: 124878407-124878407 on Build GRCh38
rs937183353 Chr.9: 124878283-124878283 on Build GRCh38
rs1016498690 Chr.9: 124878401-124878401 on Build GRCh38
rs886669592 Chr.9: 124878338-124878339 on Build GRCh38
rs968736323 Chr.9: 124878498-124878498 on Build GRCh38
rs112061540 Chr.9: 124878654-124878654 on Build GRCh38
rs1046730302 Chr.9: 124878331-124878331 on Build GRCh38
rs557337854 Chr.9: 124878380-124878380 on Build GRCh38
rs1036437250 Chr.9: 124878395-124878395 on Build GRCh38
rs191133642 Chr.9: 124878491-124878491 on Build GRCh38
rs543713749 Chr.9: 124878611-124878611 on Build GRCh38
rs528558806 Chr.9: 124878252-124878252 on Build GRCh38
rs949324933 Chr.9: 124878406-124878406 on Build GRCh38
rs538208960 Chr.9: 124878444-124878444 on Build GRCh38
rs1058555 Chr.9: 124878487-124878487 on Build GRCh38
rs929598585 Chr.9: 124878326-124878326 on Build GRCh38
rs1020657921 Chr.9: 124878509-124878509 on Build GRCh38
rs923480302 Chr.9: 124878268-124878268 on Build GRCh38
rs988860881 Chr.9: 124878240-124878240 on Build GRCh38
rs766867578 Chr.9: 124878490-124878490 on Build GRCh38
rs1030822243 Chr.9: 124878471-124878471 on Build GRCh38
rs913117740 Chr.9: 124878247-124878248 on Build GRCh38
rs117916599 Chr.9: 124878335-124878335 on Build GRCh38
rs149575615 Chr.9: 124878503-124878503 on Build GRCh38
rs559321972 Chr.9: 124878678-124878678 on Build GRCh38

View Full Product Details