Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124881668-124882161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA1
Entrez Gene ID
2800
Gene Name
golgin A1
Gene Aliases
golgin-97
UniGene
Hs.133469
Gene Chromosome Location
Chr.9: 124878294-124941114 on Build GRCh38
Transcripts
NM_002077.3 XM_005251929.3 XM_006717062.3 XM_006717063.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5063601 c.2127C>T p.R709R Chr.9: 124881793-124881793 on Build GRCh38
3324774 c.2071C>T p.R691C Chr.9: 124881849-124881849 on Build GRCh38
4188624 c.2054C>G p.T685S Chr.9: 124881866-124881866 on Build GRCh38
7264566 c.2000G>A p.R667Q Chr.9: 124881920-124881920 on Build GRCh38
1105106 c.2045C>T p.T682M Chr.9: 124881875-124881875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938655718 Chr.9: 124881689-124881689 on Build GRCh38
rs776086099 Chr.9: 124881776-124881776 on Build GRCh38
rs781112925 Chr.9: 124881984-124881984 on Build GRCh38
rs764334959 Chr.9: 124881928-124881928 on Build GRCh38
rs757460981 Chr.9: 124881942-124881942 on Build GRCh38
rs890185095 Chr.9: 124882030-124882030 on Build GRCh38
rs112456952 Chr.9: 124882095-124882095 on Build GRCh38
rs536815126 Chr.9: 124881848-124881848 on Build GRCh38
rs200355170 Chr.9: 124881743-124881743 on Build GRCh38
rs957960389 Chr.9: 124881692-124881693 on Build GRCh38
rs779786911 Chr.9: 124881880-124881880 on Build GRCh38
rs758147086 Chr.9: 124881849-124881849 on Build GRCh38
rs374280907 Chr.9: 124881947-124881947 on Build GRCh38
rs370857750 Chr.9: 124881764-124881764 on Build GRCh38
rs935391444 Chr.9: 124882121-124882121 on Build GRCh38
rs750487000 Chr.9: 124882016-124882016 on Build GRCh38
rs765370842 Chr.9: 124881943-124881943 on Build GRCh38
rs200131402 Chr.9: 124881900-124881900 on Build GRCh38
rs754112483 Chr.9: 124881933-124881933 on Build GRCh38
rs554280928 Chr.9: 124881744-124881744 on Build GRCh38
rs890205262 Chr.9: 124881879-124881879 on Build GRCh38
rs753111539 Chr.9: 124881800-124881800 on Build GRCh38
rs1038808734 Chr.9: 124882039-124882039 on Build GRCh38
rs539958942 Chr.9: 124882006-124882006 on Build GRCh38
rs571690365 Chr.9: 124882123-124882123 on Build GRCh38
rs1053371078 Chr.9: 124882027-124882027 on Build GRCh38
rs780312941 Chr.9: 124882127-124882127 on Build GRCh38
rs557307369 Chr.9: 124881985-124881985 on Build GRCh38
rs201725287 Chr.9: 124881884-124881884 on Build GRCh38
rs900157649 Chr.9: 124882041-124882041 on Build GRCh38
rs750562605 Chr.9: 124881703-124881704 on Build GRCh38
rs778079374 Chr.9: 124881847-124881847 on Build GRCh38
rs370616926 Chr.9: 124882119-124882119 on Build GRCh38
rs776373071 Chr.9: 124881886-124881886 on Build GRCh38
rs756202300 Chr.9: 124881990-124881991 on Build GRCh38
rs925912107 Chr.9: 124882009-124882009 on Build GRCh38
rs765162039 Chr.9: 124881756-124881756 on Build GRCh38
rs778945030 Chr.9: 124881874-124881874 on Build GRCh38
rs138765093 Chr.9: 124881920-124881920 on Build GRCh38
rs531668496 Chr.9: 124882098-124882098 on Build GRCh38
rs145763272 Chr.9: 124881885-124881885 on Build GRCh38
rs370812366 Chr.9: 124881986-124881986 on Build GRCh38
rs1052870501 Chr.9: 124881849-124881849 on Build GRCh38
rs754020272 Chr.9: 124881850-124881850 on Build GRCh38
rs139231269 Chr.9: 124881704-124881705 on Build GRCh38
rs948750179 Chr.9: 124882001-124882001 on Build GRCh38
rs750656302 Chr.9: 124882004-124882005 on Build GRCh38
rs760096663 Chr.9: 124881793-124881793 on Build GRCh38
rs769477508 Chr.9: 124881899-124881899 on Build GRCh38
rs768084674 Chr.9: 124881916-124881916 on Build GRCh38
rs377727059 Chr.9: 124881795-124881795 on Build GRCh38
rs756377921 Chr.9: 124881811-124881811 on Build GRCh38
rs752475423 Chr.9: 124881820-124881821 on Build GRCh38
rs182177127 Chr.9: 124882046-124882046 on Build GRCh38
rs752151566 Chr.9: 124881792-124881792 on Build GRCh38
rs757371259 Chr.9: 124881866-124881866 on Build GRCh38
rs1027198000 Chr.9: 124882052-124882052 on Build GRCh38
rs911497321 Chr.9: 124882087-124882087 on Build GRCh38
rs202227499 Chr.9: 124881940-124881940 on Build GRCh38
rs758349685 Chr.9: 124881962-124881962 on Build GRCh38
rs1010805909 Chr.9: 124881934-124881934 on Build GRCh38
rs141961382 Chr.9: 124881794-124881794 on Build GRCh38
rs771994948 Chr.9: 124881877-124881877 on Build GRCh38
rs780060409 Chr.9: 124881976-124881976 on Build GRCh38
rs760313990 Chr.9: 124881745-124881745 on Build GRCh38
rs908657344 Chr.9: 124881688-124881688 on Build GRCh38
rs761426461 Chr.9: 124881778-124881778 on Build GRCh38
rs2304956 Chr.9: 124882017-124882017 on Build GRCh38
rs188750707 Chr.9: 124881983-124881983 on Build GRCh38
rs981883937 Chr.9: 124881717-124881717 on Build GRCh38
rs888707005 Chr.9: 124882056-124882056 on Build GRCh38
rs769564188 Chr.9: 124881992-124881992 on Build GRCh38
rs761222606 Chr.9: 124881875-124881875 on Build GRCh38
rs746877287 Chr.9: 124881980-124881980 on Build GRCh38
rs556471997 Chr.9: 124881887-124881887 on Build GRCh38
rs982949183 Chr.9: 124881918-124881918 on Build GRCh38
rs114403273 Chr.9: 124882031-124882031 on Build GRCh38
rs761134317 Chr.9: 124881927-124881927 on Build GRCh38
rs764795922 Chr.9: 124881787-124881787 on Build GRCh38
rs989690588 Chr.9: 124881697-124881697 on Build GRCh38
rs3214725 Chr.9: 124881710-124881711 on Build GRCh38
rs559693123 Chr.9: 124882078-124882078 on Build GRCh38
rs1041235194 Chr.9: 124882105-124882105 on Build GRCh38
rs1004353870 Chr.9: 124882068-124882068 on Build GRCh38

View Full Product Details