Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 124890300-124890802 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA1
Entrez Gene ID
2800
Gene Name
golgin A1
Gene Aliases
golgin-97
UniGene
Hs.133469
Gene Chromosome Location
Chr.9: 124878294-124941114 on Build GRCh38
Transcripts
NM_002077.3 XM_005251929.3 XM_006717062.3 XM_006717063.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3703479 c.1452A>C p.Q484H Chr.9: 124890434-124890434 on Build GRCh38
1105116 c.1447G>A p.A483T Chr.9: 124890439-124890439 on Build GRCh38
3675230 c.1478G>T p.R493M Chr.9: 124890408-124890408 on Build GRCh38
1105115 c.1448C>T p.A483V Chr.9: 124890438-124890438 on Build GRCh38
1105117 c.1411G>T p.E471* Chr.9: 124890475-124890475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966424084 Chr.9: 124890712-124890712 on Build GRCh38
rs1006225986 Chr.9: 124890583-124890583 on Build GRCh38
rs150023063 Chr.9: 124890471-124890471 on Build GRCh38
rs199773776 Chr.9: 124890361-124890361 on Build GRCh38
rs943572676 Chr.9: 124890482-124890482 on Build GRCh38
rs777133957 Chr.9: 124890485-124890485 on Build GRCh38
rs1010775294 Chr.9: 124890664-124890664 on Build GRCh38
rs943533048 Chr.9: 124890764-124890764 on Build GRCh38
rs184162785 Chr.9: 124890550-124890550 on Build GRCh38
rs774901878 Chr.9: 124890321-124890321 on Build GRCh38
rs886403794 Chr.9: 124890578-124890578 on Build GRCh38
rs769387512 Chr.9: 124890408-124890408 on Build GRCh38
rs938189361 Chr.9: 124890328-124890328 on Build GRCh38
rs1003425074 Chr.9: 124890626-124890626 on Build GRCh38
rs753929411 Chr.9: 124890363-124890363 on Build GRCh38
rs746697142 Chr.9: 124890502-124890502 on Build GRCh38
rs549333887 Chr.9: 124890614-124890614 on Build GRCh38
rs955216876 Chr.9: 124890731-124890731 on Build GRCh38
rs562412524 Chr.9: 124890560-124890560 on Build GRCh38
rs377268563 Chr.9: 124890428-124890428 on Build GRCh38
rs906535827 Chr.9: 124890589-124890589 on Build GRCh38
rs78804812 Chr.9: 124890448-124890448 on Build GRCh38
rs549828659 Chr.9: 124890660-124890660 on Build GRCh38
rs139716668 Chr.9: 124890372-124890372 on Build GRCh38
rs747682330 Chr.9: 124890401-124890401 on Build GRCh38
rs622280 Chr.9: 124890342-124890342 on Build GRCh38
rs776017837 Chr.9: 124890508-124890508 on Build GRCh38
rs779970639 Chr.9: 124890442-124890442 on Build GRCh38
rs1003883682 Chr.9: 124890352-124890352 on Build GRCh38
rs752244 Chr.9: 124890726-124890726 on Build GRCh38
rs145680781 Chr.9: 124890562-124890562 on Build GRCh38
rs764323375 Chr.9: 124890418-124890418 on Build GRCh38
rs79327267 Chr.9: 124890720-124890720 on Build GRCh38
rs543719133 Chr.9: 124890343-124890343 on Build GRCh38
rs1019999206 Chr.9: 124890682-124890682 on Build GRCh38
rs368593273 Chr.9: 124890379-124890379 on Build GRCh38
rs774953646 Chr.9: 124890526-124890526 on Build GRCh38
rs754627379 Chr.9: 124890354-124890354 on Build GRCh38
rs762013178 Chr.9: 124890529-124890529 on Build GRCh38
rs761062293 Chr.9: 124890517-124890517 on Build GRCh38
rs949496793 Chr.9: 124890553-124890553 on Build GRCh38
rs773702787 Chr.9: 124890417-124890417 on Build GRCh38
rs112448305 Chr.9: 124890532-124890532 on Build GRCh38
rs374921841 Chr.9: 124890758-124890758 on Build GRCh38
rs1034055399 Chr.9: 124890645-124890645 on Build GRCh38
rs140097195 Chr.9: 124890410-124890410 on Build GRCh38
rs1037741578 Chr.9: 124890522-124890522 on Build GRCh38
rs767741513 Chr.9: 124890346-124890346 on Build GRCh38
rs192906152 Chr.9: 124890449-124890449 on Build GRCh38
rs771438568 Chr.9: 124890509-124890509 on Build GRCh38
rs991126400 Chr.9: 124890726-124890726 on Build GRCh38
rs760939520 Chr.9: 124890710-124890710 on Build GRCh38
rs1022034070 Chr.9: 124890677-124890677 on Build GRCh38
rs200187423 Chr.9: 124890390-124890390 on Build GRCh38
rs968317289 Chr.9: 124890699-124890699 on Build GRCh38
rs774962630 Chr.9: 124890409-124890409 on Build GRCh38
rs748765014 Chr.9: 124890498-124890498 on Build GRCh38
rs61733258 Chr.9: 124890457-124890457 on Build GRCh38
rs753011394 Chr.9: 124890360-124890360 on Build GRCh38
rs528641619 Chr.9: 124890540-124890540 on Build GRCh38
rs949585789 Chr.9: 124890775-124890775 on Build GRCh38
rs768981360 Chr.9: 124890520-124890520 on Build GRCh38
rs142324555 Chr.9: 124890413-124890413 on Build GRCh38
rs1052454417 Chr.9: 124890459-124890459 on Build GRCh38
rs772470633 Chr.9: 124890503-124890503 on Build GRCh38
rs529318977 Chr.9: 124890581-124890581 on Build GRCh38
rs769394758 Chr.9: 124890484-124890484 on Build GRCh38
rs746738929 Chr.9: 124890385-124890385 on Build GRCh38
rs770420839 Chr.9: 124890521-124890522 on Build GRCh38
rs1047692735 Chr.9: 124890564-124890564 on Build GRCh38
rs756732016 Chr.9: 124890373-124890373 on Build GRCh38
rs768248656 Chr.9: 124890362-124890362 on Build GRCh38
rs757281555 Chr.9: 124890364-124890364 on Build GRCh38
rs767004250 Chr.9: 124890340-124890340 on Build GRCh38
rs775722194 Chr.9: 124890416-124890416 on Build GRCh38
rs978416920 Chr.9: 124890705-124890705 on Build GRCh38
rs768425423 Chr.9: 124890388-124890388 on Build GRCh38
rs201455650 Chr.9: 124890381-124890381 on Build GRCh38
rs765213836 Chr.9: 124890421-124890421 on Build GRCh38
rs750436692 Chr.9: 124890422-124890422 on Build GRCh38
rs900435613 Chr.9: 124890616-124890616 on Build GRCh38
rs565867185 Chr.9: 124890676-124890676 on Build GRCh38
rs1026171722 Chr.9: 124890320-124890320 on Build GRCh38
rs987737066 Chr.9: 124890739-124890739 on Build GRCh38
rs1010394679 Chr.9: 124890680-124890680 on Build GRCh38
rs756995687 Chr.9: 124890736-124890736 on Build GRCh38
rs754022523 Chr.9: 124890420-124890420 on Build GRCh38
rs139362538 Chr.9: 124890467-124890467 on Build GRCh38

View Full Product Details