Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 125302471-125302967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAPVD1
Entrez Gene ID
26130
Gene Name
GTPase activating protein and VPS9 domains 1
Gene Aliases
GAPEX5, GAPex-5, RAP6
UniGene
Hs.495134
Gene Chromosome Location
Chr.9: 125261794-125365011 on Build GRCh38
Transcripts
NM_001282679.1 NM_001282680.1 NM_001282681.1 NM_015635.3 XM_005251901.3 XM_005251904.3 XM_006717044.3 XM_011518499.2 XM_011518500.2 XM_011518502.2 XM_011518506.2 XM_011518507.2 XM_017014600.1 XM_017014601.1 XM_017014602.1 XM_017014603.1 XM_017014604.1 XM_017014605.1 XM_017014606.1 XM_017014607.1 XM_017014608.1 XM_017014609.1 XM_017014610.1 XM_017014611.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3952337 c.722A>G p.D241G Chr.9: 125302519-125302519 on Build GRCh38
4949605 c.820T>C p.Y274H Chr.9: 125302617-125302617 on Build GRCh38
6669296 c.1012C>T p.R338* Chr.9: 125302809-125302809 on Build GRCh38
7099073 c.1023G>C p.L341L Chr.9: 125302820-125302820 on Build GRCh38
5633237 c.997A>G p.I333V Chr.9: 125302794-125302794 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749152514 Chr.9: 125302646-125302646 on Build GRCh38
rs1039098240 Chr.9: 125302902-125302902 on Build GRCh38
rs773295739 Chr.9: 125302541-125302541 on Build GRCh38
rs4437748 Chr.9: 125302569-125302569 on Build GRCh38
rs753020065 Chr.9: 125302596-125302596 on Build GRCh38
rs200449709 Chr.9: 125302784-125302784 on Build GRCh38
rs901966713 Chr.9: 125302929-125302929 on Build GRCh38
rs752392994 Chr.9: 125302772-125302772 on Build GRCh38
rs181982027 Chr.9: 125302548-125302548 on Build GRCh38
rs764441838 Chr.9: 125302718-125302718 on Build GRCh38
rs138640842 Chr.9: 125302818-125302818 on Build GRCh38
rs151076290 Chr.9: 125302778-125302778 on Build GRCh38
rs200721068 Chr.9: 125302600-125302600 on Build GRCh38
rs372739784 Chr.9: 125302517-125302517 on Build GRCh38
rs762362496 Chr.9: 125302750-125302750 on Build GRCh38
rs61733785 Chr.9: 125302676-125302676 on Build GRCh38
rs753338033 Chr.9: 125302759-125302759 on Build GRCh38
rs745967169 Chr.9: 125302692-125302692 on Build GRCh38
rs770821838 Chr.9: 125302677-125302677 on Build GRCh38
rs531525173 Chr.9: 125302582-125302582 on Build GRCh38
rs769870989 Chr.9: 125302820-125302820 on Build GRCh38
rs768254622 Chr.9: 125302523-125302523 on Build GRCh38
rs779510473 Chr.9: 125302518-125302518 on Build GRCh38
rs953198730 Chr.9: 125302924-125302924 on Build GRCh38
rs768812788 Chr.9: 125302833-125302833 on Build GRCh38
rs915667077 Chr.9: 125302752-125302752 on Build GRCh38
rs776630852 Chr.9: 125302575-125302575 on Build GRCh38
rs768005190 Chr.9: 125302852-125302852 on Build GRCh38
rs777495342 Chr.9: 125302667-125302667 on Build GRCh38
rs542250668 Chr.9: 125302535-125302535 on Build GRCh38
rs775618648 Chr.9: 125302830-125302830 on Build GRCh38
rs147633449 Chr.9: 125302606-125302606 on Build GRCh38
rs1012994439 Chr.9: 125302616-125302616 on Build GRCh38
rs935942659 Chr.9: 125302553-125302553 on Build GRCh38
rs778284557 Chr.9: 125302511-125302511 on Build GRCh38
rs749854037 Chr.9: 125302512-125302512 on Build GRCh38
rs764531486 Chr.9: 125302619-125302619 on Build GRCh38
rs763240757 Chr.9: 125302711-125302711 on Build GRCh38
rs778815867 Chr.9: 125302528-125302528 on Build GRCh38
rs757180474 Chr.9: 125302785-125302785 on Build GRCh38
rs776742076 Chr.9: 125302689-125302689 on Build GRCh38
rs757773363 Chr.9: 125302634-125302634 on Build GRCh38
rs747985352 Chr.9: 125302531-125302531 on Build GRCh38
rs770645803 Chr.9: 125302558-125302558 on Build GRCh38
rs778725908 Chr.9: 125302649-125302649 on Build GRCh38
rs764914655 Chr.9: 125302766-125302766 on Build GRCh38
rs55779102 Chr.9: 125302647-125302647 on Build GRCh38
rs758271475 Chr.9: 125302666-125302666 on Build GRCh38
rs985811969 Chr.9: 125302940-125302940 on Build GRCh38
rs752115913 Chr.9: 125302632-125302632 on Build GRCh38
rs774638407 Chr.9: 125302749-125302749 on Build GRCh38
rs370781614 Chr.9: 125302832-125302832 on Build GRCh38
rs762103980 Chr.9: 125302851-125302851 on Build GRCh38
rs764168229 Chr.9: 125302733-125302733 on Build GRCh38
rs997675645 Chr.9: 125302930-125302930 on Build GRCh38
rs767937930 Chr.9: 125302753-125302753 on Build GRCh38
rs748967334 Chr.9: 125302520-125302520 on Build GRCh38
rs1045575465 Chr.9: 125302809-125302809 on Build GRCh38
rs765485724 Chr.9: 125302593-125302593 on Build GRCh38
rs550220093 Chr.9: 125302893-125302893 on Build GRCh38
rs1054441369 Chr.9: 125302566-125302566 on Build GRCh38
rs547560589 Chr.9: 125302693-125302693 on Build GRCh38
rs199552778 Chr.9: 125302856-125302856 on Build GRCh38
rs759279783 Chr.9: 125302839-125302841 on Build GRCh38
rs570328296 Chr.9: 125302935-125302935 on Build GRCh38
rs943357493 Chr.9: 125302886-125302886 on Build GRCh38
rs781013665 Chr.9: 125302790-125302790 on Build GRCh38
rs774733930 Chr.9: 125302849-125302849 on Build GRCh38
rs369013199 Chr.9: 125302493-125302493 on Build GRCh38
rs754545159 Chr.9: 125302763-125302763 on Build GRCh38
rs747890761 Chr.9: 125302658-125302658 on Build GRCh38

View Full Product Details