Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 125323707-125324207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAPVD1
Entrez Gene ID
26130
Gene Name
GTPase activating protein and VPS9 domains 1
Gene Aliases
GAPEX5, GAPex-5, RAP6
UniGene
Hs.495134
Gene Chromosome Location
Chr.9: 125261794-125365011 on Build GRCh38
Transcripts
NM_001282679.1 NM_001282680.1 NM_001282681.1 NM_015635.3 XM_005251901.3 XM_005251904.3 XM_006717044.3 XM_011518499.2 XM_011518500.2 XM_011518502.2 XM_011518506.2 XM_011518507.2 XM_017014600.1 XM_017014601.1 XM_017014602.1 XM_017014603.1 XM_017014604.1 XM_017014605.1 XM_017014606.1 XM_017014607.1 XM_017014608.1 XM_017014609.1 XM_017014610.1 XM_017014611.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7353238 c.1757T>C p.V586A Chr.9: 125323822-125323822 on Build GRCh38
3382420 c.1741A>T p.N581Y Chr.9: 125323806-125323806 on Build GRCh38
4957758 c.1845A>T p.L615F Chr.9: 125323910-125323910 on Build GRCh38
7135909 c.1739C>T p.S580L Chr.9: 125323804-125323804 on Build GRCh38
7137386 c.1774G>C p.E592Q Chr.9: 125323839-125323839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375861526 Chr.9: 125324074-125324074 on Build GRCh38
rs768124214 Chr.9: 125323913-125323913 on Build GRCh38
rs1018958687 Chr.9: 125324148-125324148 on Build GRCh38
rs976053233 Chr.9: 125324180-125324180 on Build GRCh38
rs984252186 Chr.9: 125324088-125324088 on Build GRCh38
rs1010022643 Chr.9: 125323888-125323888 on Build GRCh38
rs766333634 Chr.9: 125323776-125323776 on Build GRCh38
rs768941829 Chr.9: 125323764-125323766 on Build GRCh38
rs777129072 Chr.9: 125323853-125323853 on Build GRCh38
rs771517562 Chr.9: 125323882-125323882 on Build GRCh38
rs774813812 Chr.9: 125323894-125323894 on Build GRCh38
rs953430406 Chr.9: 125323739-125323739 on Build GRCh38
rs972186607 Chr.9: 125323849-125323849 on Build GRCh38
rs1028940453 Chr.9: 125324133-125324133 on Build GRCh38
rs112254230 Chr.9: 125323895-125323895 on Build GRCh38
rs748860611 Chr.9: 125323903-125323903 on Build GRCh38
rs41316946 Chr.9: 125324040-125324040 on Build GRCh38
rs774003860 Chr.9: 125323915-125323915 on Build GRCh38
rs1000504083 Chr.9: 125324099-125324099 on Build GRCh38
rs904406286 Chr.9: 125323941-125323941 on Build GRCh38
rs567468837 Chr.9: 125323856-125323856 on Build GRCh38
rs771692854 Chr.9: 125323908-125323908 on Build GRCh38
rs368394108 Chr.9: 125323783-125323783 on Build GRCh38
rs904397823 Chr.9: 125324080-125324080 on Build GRCh38
rs1034293359 Chr.9: 125323992-125323992 on Build GRCh38
rs926932920 Chr.9: 125323954-125323954 on Build GRCh38
rs201287198 Chr.9: 125323959-125323959 on Build GRCh38
rs889045337 Chr.9: 125324135-125324135 on Build GRCh38
rs372630662 Chr.9: 125323810-125323810 on Build GRCh38
rs964302974 Chr.9: 125324156-125324156 on Build GRCh38
rs900183029 Chr.9: 125323802-125323802 on Build GRCh38
rs753979382 Chr.9: 125323782-125323782 on Build GRCh38
rs981084712 Chr.9: 125323950-125323950 on Build GRCh38
rs76944979 Chr.9: 125323754-125323754 on Build GRCh38
rs750359976 Chr.9: 125323772-125323772 on Build GRCh38
rs367596260 Chr.9: 125323974-125323974 on Build GRCh38
rs572438187 Chr.9: 125324174-125324174 on Build GRCh38
rs778606692 Chr.9: 125323782-125323782 on Build GRCh38
rs146812220 Chr.9: 125324094-125324094 on Build GRCh38
rs766375230 Chr.9: 125323961-125323961 on Build GRCh38
rs1025731512 Chr.9: 125324079-125324082 on Build GRCh38
rs769111202 Chr.9: 125323850-125323850 on Build GRCh38
rs780436212 Chr.9: 125323809-125323809 on Build GRCh38
rs41305469 Chr.9: 125323933-125323933 on Build GRCh38
rs116983971 Chr.9: 125323997-125323997 on Build GRCh38
rs755103690 Chr.9: 125323806-125323806 on Build GRCh38
rs41305471 Chr.9: 125323822-125323822 on Build GRCh38
rs111454673 Chr.9: 125324021-125324021 on Build GRCh38
rs761508180 Chr.9: 125323920-125323920 on Build GRCh38
rs750725903 Chr.9: 125323987-125323988 on Build GRCh38
rs559130356 Chr.9: 125324142-125324142 on Build GRCh38
rs374411640 Chr.9: 125323757-125323757 on Build GRCh38
rs778349706 Chr.9: 125323836-125323836 on Build GRCh38
rs747488572 Chr.9: 125323861-125323861 on Build GRCh38
rs767292054 Chr.9: 125323931-125323931 on Build GRCh38
rs1014298345 Chr.9: 125324078-125324078 on Build GRCh38
rs10118820 Chr.9: 125324064-125324064 on Build GRCh38

View Full Product Details