Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127088035-127088530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL2
Entrez Gene ID
23452
Gene Name
angiopoietin like 2
Gene Aliases
ARP2, HARP
UniGene
Hs.653262
Gene Chromosome Location
Chr.9: 127087349-127122803 on Build GRCh38
Transcripts
NM_012098.2 NM_001190728.1 NM_001190729.1 NM_001190730.1 NM_001322320.1 NM_001322321.1 NM_001322322.1 NM_001322323.1 NM_001322324.1 NM_001322325.1 NM_014636.2 XM_006717328.3 XM_011519225.1 XM_011519226.1 XM_011519227.1 XM_011519228.1 XM_011519229.1 XM_011519230.2 XM_011519231.1 XM_011519232.1 XM_011519233.1 XM_011519234.2 XM_011519235.2 XM_011519236.1 XM_011519238.2 XM_017015342.1 XM_017015343.1 XM_017015344.1 XM_017015345.1 XM_017015346.1 XM_017015347.1 XM_017015348.1 XM_017015349.1 XM_017015350.1

Gene Symbol
RALGPS1
Entrez Gene ID
9649
Gene Name
Ral GEF with PH domain and SH3 binding motif 1
Gene Aliases
RALGEF2, RALGPS1A
UniGene
Hs.432842
Gene Chromosome Location
Chr.9: 126914774-127223166 on Build GRCh38
Transcripts
NM_012098.2 NM_001190728.1 NM_001190729.1 NM_001190730.1 NM_001322320.1 NM_001322321.1 NM_001322322.1 NM_001322323.1 NM_001322324.1 NM_001322325.1 NM_014636.2 XM_006717328.3 XM_011519225.1 XM_011519226.1 XM_011519227.1 XM_011519228.1 XM_011519229.1 XM_011519230.2 XM_011519231.1 XM_011519232.1 XM_011519233.1 XM_011519234.2 XM_011519235.2 XM_011519236.1 XM_011519238.2 XM_017015342.1 XM_017015343.1 XM_017015344.1 XM_017015345.1 XM_017015346.1 XM_017015347.1 XM_017015348.1 XM_017015349.1 XM_017015350.1

SNP Detail

SNP ID Chromosome Location
rs183676873 Chr.9: 127088468-127088468 on Build GRCh38
rs893994717 Chr.9: 127088323-127088323 on Build GRCh38
rs1053055759 Chr.9: 127088300-127088300 on Build GRCh38
rs909705206 Chr.9: 127088108-127088108 on Build GRCh38
rs922849401 Chr.9: 127088279-127088279 on Build GRCh38
rs557692235 Chr.9: 127088362-127088362 on Build GRCh38
rs942953305 Chr.9: 127088254-127088254 on Build GRCh38
rs1052626835 Chr.9: 127088095-127088095 on Build GRCh38
rs868011382 Chr.9: 127088315-127088315 on Build GRCh38
rs1022077332 Chr.9: 127088121-127088121 on Build GRCh38
rs750966650 Chr.9: 127088374-127088374 on Build GRCh38
rs991423136 Chr.9: 127088144-127088144 on Build GRCh38
rs1039414892 Chr.9: 127088064-127088064 on Build GRCh38
rs1010696979 Chr.9: 127088110-127088110 on Build GRCh38
rs948042899 Chr.9: 127088221-127088221 on Build GRCh38
rs1043565306 Chr.9: 127088364-127088364 on Build GRCh38
rs2787592 Chr.9: 127088124-127088124 on Build GRCh38
rs1026153413 Chr.9: 127088227-127088227 on Build GRCh38
rs933659734 Chr.9: 127088281-127088281 on Build GRCh38
rs189408102 Chr.9: 127088081-127088081 on Build GRCh38
rs1016671799 Chr.9: 127088435-127088435 on Build GRCh38
rs886889978 Chr.9: 127088399-127088399 on Build GRCh38
rs765716343 Chr.9: 127088350-127088350 on Build GRCh38
rs574948941 Chr.9: 127088382-127088382 on Build GRCh38
rs373222674 Chr.9: 127088251-127088251 on Build GRCh38
rs754439219 Chr.9: 127088454-127088454 on Build GRCh38
rs537992120 Chr.9: 127088337-127088337 on Build GRCh38
rs191429062 Chr.9: 127088420-127088420 on Build GRCh38
rs951162964 Chr.9: 127088071-127088071 on Build GRCh38
rs896366539 Chr.9: 127088466-127088466 on Build GRCh38
rs971336692 Chr.9: 127088486-127088486 on Build GRCh38
rs376167034 Chr.9: 127088276-127088276 on Build GRCh38
rs991288664 Chr.9: 127088460-127088460 on Build GRCh38
rs983058413 Chr.9: 127088086-127088086 on Build GRCh38
rs766773997 Chr.9: 127088490-127088490 on Build GRCh38
rs7023554 Chr.9: 127088249-127088249 on Build GRCh38
rs1029827248 Chr.9: 127088070-127088070 on Build GRCh38
rs758793342 Chr.9: 127088415-127088415 on Build GRCh38
rs141796573 Chr.9: 127088216-127088216 on Build GRCh38
rs892180254 Chr.9: 127088107-127088107 on Build GRCh38
rs975678264 Chr.9: 127088278-127088278 on Build GRCh38
rs960329470 Chr.9: 127088195-127088195 on Build GRCh38
rs567292408 Chr.9: 127088282-127088282 on Build GRCh38
rs537154836 Chr.9: 127088191-127088191 on Build GRCh38
rs577615905 Chr.9: 127088375-127088375 on Build GRCh38
rs951442015 Chr.9: 127088241-127088241 on Build GRCh38
rs1001964927 Chr.9: 127088431-127088431 on Build GRCh38
rs760227706 Chr.9: 127088346-127088346 on Build GRCh38

View Full Product Details