Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127713882-127714385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP157
Entrez Gene ID
286207
Gene Name
cilia and flagella associated protein 157
Gene Aliases
C9orf117
UniGene
Hs.652519
Gene Chromosome Location
Chr.9: 127706261-127716002 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014630.1 NM_001002913.1 XM_006716955.3 XM_017014276.1 XM_017014277.1 XM_017014278.1

Gene Symbol
PTRH1
Entrez Gene ID
138428
Gene Name
peptidyl-tRNA hydrolase 1 homolog
Gene Aliases
C9orf115, PTH1
UniGene
Hs.653265
Gene Chromosome Location
Chr.9: 127695024-127716714 on Build GRCh38
Transcripts
NM_001012502.2 XM_006717064.3 XM_011518559.2 XM_017014630.1 NM_001002913.1 XM_006716955.3 XM_017014276.1 XM_017014277.1 XM_017014278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7386427 c.517C>G p.Q173E Chr.9: 127714228-127714228 on Build GRCh38
1460108 c.589G>A p.D197N Chr.9: 127714156-127714156 on Build GRCh38
7225545 c.641C>T p.P214L Chr.9: 127714104-127714104 on Build GRCh38
7213934 c.481G>T p.V161L Chr.9: 127714264-127714264 on Build GRCh38
1187817 c.479G>A p.R160Q Chr.9: 127714266-127714266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs950241233 Chr.9: 127713994-127713994 on Build GRCh38
rs767878836 Chr.9: 127714256-127714256 on Build GRCh38
rs201042035 Chr.9: 127714233-127714233 on Build GRCh38
rs370341445 Chr.9: 127714362-127714362 on Build GRCh38
rs757696659 Chr.9: 127714060-127714060 on Build GRCh38
rs760754685 Chr.9: 127713928-127713928 on Build GRCh38
rs769978661 Chr.9: 127714097-127714097 on Build GRCh38
rs369609759 Chr.9: 127714334-127714334 on Build GRCh38
rs997494620 Chr.9: 127714279-127714279 on Build GRCh38
rs926989419 Chr.9: 127713910-127713910 on Build GRCh38
rs750188015 Chr.9: 127714235-127714235 on Build GRCh38
rs373886001 Chr.9: 127714330-127714330 on Build GRCh38
rs201242648 Chr.9: 127713903-127713903 on Build GRCh38
rs766802204 Chr.9: 127714212-127714212 on Build GRCh38
rs10851 Chr.9: 127714232-127714232 on Build GRCh38
rs557298488 Chr.9: 127714256-127714256 on Build GRCh38
rs372888029 Chr.9: 127714093-127714093 on Build GRCh38
rs376325151 Chr.9: 127713936-127713936 on Build GRCh38
rs1047881788 Chr.9: 127714182-127714182 on Build GRCh38
rs561739237 Chr.9: 127714342-127714342 on Build GRCh38
rs771263623 Chr.9: 127714346-127714346 on Build GRCh38
rs745641656 Chr.9: 127714002-127714002 on Build GRCh38
rs768046728 Chr.9: 127714064-127714064 on Build GRCh38
rs772492545 Chr.9: 127714200-127714200 on Build GRCh38
rs560855459 Chr.9: 127714040-127714040 on Build GRCh38
rs773501750 Chr.9: 127714260-127714260 on Build GRCh38
rs374142343 Chr.9: 127714317-127714317 on Build GRCh38
rs11540273 Chr.9: 127714252-127714252 on Build GRCh38
rs771007236 Chr.9: 127713929-127713929 on Build GRCh38
rs750972481 Chr.9: 127714125-127714125 on Build GRCh38
rs749361456 Chr.9: 127714172-127714172 on Build GRCh38
rs151211174 Chr.9: 127714103-127714103 on Build GRCh38
rs541795955 Chr.9: 127714027-127714027 on Build GRCh38
rs1006461792 Chr.9: 127714244-127714244 on Build GRCh38
rs759710036 Chr.9: 127713935-127713935 on Build GRCh38
rs539609593 Chr.9: 127714303-127714303 on Build GRCh38
rs762418487 Chr.9: 127714222-127714222 on Build GRCh38
rs763754948 Chr.9: 127714287-127714287 on Build GRCh38
rs762138186 Chr.9: 127714187-127714187 on Build GRCh38
rs143528313 Chr.9: 127713957-127713957 on Build GRCh38
rs776811044 Chr.9: 127713934-127713934 on Build GRCh38
rs903402449 Chr.9: 127714020-127714020 on Build GRCh38
rs568809556 Chr.9: 127714228-127714228 on Build GRCh38
rs767993669 Chr.9: 127714119-127714119 on Build GRCh38
rs557520001 Chr.9: 127713908-127713908 on Build GRCh38
rs572914315 Chr.9: 127713996-127713996 on Build GRCh38
rs980009614 Chr.9: 127714108-127714108 on Build GRCh38
rs117880987 Chr.9: 127714104-127714104 on Build GRCh38
rs757157562 Chr.9: 127714152-127714152 on Build GRCh38
rs1054742320 Chr.9: 127714254-127714254 on Build GRCh38
rs949171676 Chr.9: 127714183-127714183 on Build GRCh38
rs764224561 Chr.9: 127714107-127714107 on Build GRCh38
rs746777512 Chr.9: 127714147-127714147 on Build GRCh38
rs34215199 Chr.9: 127714199-127714200 on Build GRCh38
rs374423214 Chr.9: 127714030-127714030 on Build GRCh38
rs763807558 Chr.9: 127714223-127714223 on Build GRCh38
rs369413361 Chr.9: 127714293-127714293 on Build GRCh38
rs145355499 Chr.9: 127714277-127714277 on Build GRCh38
rs762515935 Chr.9: 127714282-127714282 on Build GRCh38
rs553457756 Chr.9: 127714318-127714318 on Build GRCh38
rs548167098 Chr.9: 127714175-127714175 on Build GRCh38
rs762432020 Chr.9: 127714122-127714122 on Build GRCh38
rs751931992 Chr.9: 127714057-127714057 on Build GRCh38
rs761489230 Chr.9: 127714296-127714296 on Build GRCh38
rs756036042 Chr.9: 127714245-127714245 on Build GRCh38
rs774608645 Chr.9: 127714181-127714181 on Build GRCh38
rs752199939 Chr.9: 127714217-127714217 on Build GRCh38
rs770941801 Chr.9: 127714090-127714090 on Build GRCh38
rs14033 Chr.9: 127714250-127714250 on Build GRCh38
rs141664618 Chr.9: 127714249-127714249 on Build GRCh38
rs753299888 Chr.9: 127714065-127714065 on Build GRCh38
rs375475837 Chr.9: 127714135-127714135 on Build GRCh38
rs1006511045 Chr.9: 127714056-127714056 on Build GRCh38
rs774283636 Chr.9: 127714115-127714116 on Build GRCh38
rs370579403 Chr.9: 127713937-127713937 on Build GRCh38
rs750185535 Chr.9: 127714305-127714305 on Build GRCh38
rs1045894299 Chr.9: 127714203-127714203 on Build GRCh38
rs867137147 Chr.9: 127714255-127714255 on Build GRCh38
rs377023908 Chr.9: 127714349-127714349 on Build GRCh38
rs1017610892 Chr.9: 127714292-127714292 on Build GRCh38
rs956329857 Chr.9: 127714088-127714088 on Build GRCh38
rs1033011487 Chr.9: 127714028-127714028 on Build GRCh38
rs888025441 Chr.9: 127714049-127714049 on Build GRCh38
rs966166403 Chr.9: 127714084-127714084 on Build GRCh38
rs773019870 Chr.9: 127713912-127713912 on Build GRCh38
rs565746487 Chr.9: 127714128-127714128 on Build GRCh38
rs758116458 Chr.9: 127714070-127714070 on Build GRCh38
rs755913569 Chr.9: 127714308-127714308 on Build GRCh38
rs773696109 Chr.9: 127714202-127714202 on Build GRCh38
rs778399562 Chr.9: 127714068-127714068 on Build GRCh38
rs747365184 Chr.9: 127714344-127714344 on Build GRCh38
rs552233791 Chr.9: 127714116-127714116 on Build GRCh38
rs144597665 Chr.9: 127714157-127714157 on Build GRCh38
rs546254841 Chr.9: 127713941-127713941 on Build GRCh38
rs768925730 Chr.9: 127713949-127713950 on Build GRCh38
rs746828810 Chr.9: 127714089-127714089 on Build GRCh38
rs529899427 Chr.9: 127714059-127714059 on Build GRCh38
rs758004584 Chr.9: 127714134-127714134 on Build GRCh38
rs761187751 Chr.9: 127714209-127714209 on Build GRCh38
rs147912974 Chr.9: 127714164-127714164 on Build GRCh38
rs755909480 Chr.9: 127714031-127714031 on Build GRCh38
rs893178147 Chr.9: 127714038-127714038 on Build GRCh38
rs779880867 Chr.9: 127714247-127714247 on Build GRCh38
rs777041822 Chr.9: 127714280-127714280 on Build GRCh38
rs752164335 Chr.9: 127714129-127714129 on Build GRCh38
rs150422298 Chr.9: 127714261-127714261 on Build GRCh38
rs762191160 Chr.9: 127714118-127714118 on Build GRCh38
rs549752136 Chr.9: 127714071-127714071 on Build GRCh38
rs781266875 Chr.9: 127714156-127714156 on Build GRCh38
rs374364179 Chr.9: 127714335-127714335 on Build GRCh38
rs367619523 Chr.9: 127714248-127714248 on Build GRCh38
rs148037738 Chr.9: 127714319-127714319 on Build GRCh38
rs754570027 Chr.9: 127714067-127714067 on Build GRCh38
rs111378450 Chr.9: 127714251-127714251 on Build GRCh38

View Full Product Details