Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127808179-127808722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FPGS
Entrez Gene ID
2356
Gene Name
folylpolyglutamate synthase
Gene Aliases
-
UniGene
Hs.335084
Gene Chromosome Location
Chr.9: 127802858-127814520 on Build GRCh38
Transcripts
NM_001018078.2 NM_001288803.1 NM_004957.5 NR_110170.1 XM_005251864.3 XM_011518439.2 XM_017014565.1 XM_017014566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685506 c.963C>T p.D321D Chr.9: 127808698-127808698 on Build GRCh38
5786649 c.902A>T p.H301L Chr.9: 127808637-127808637 on Build GRCh38
3929812 c.855C>G p.A285A Chr.9: 127808590-127808590 on Build GRCh38
4886983 c.823-1G>T p.? Chr.9: 127808557-127808557 on Build GRCh38
1460140 c.882C>T p.T294T Chr.9: 127808617-127808617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021153306 Chr.9: 127808207-127808207 on Build GRCh38
rs902528643 Chr.9: 127808323-127808323 on Build GRCh38
rs201728522 Chr.9: 127808536-127808536 on Build GRCh38
rs376937555 Chr.9: 127808690-127808690 on Build GRCh38
rs749391689 Chr.9: 127808589-127808589 on Build GRCh38
rs113550574 Chr.9: 127808462-127808462 on Build GRCh38
rs374522185 Chr.9: 127808345-127808345 on Build GRCh38
rs758923980 Chr.9: 127808213-127808213 on Build GRCh38
rs762305751 Chr.9: 127808565-127808565 on Build GRCh38
rs139705653 Chr.9: 127808513-127808513 on Build GRCh38
rs1019438405 Chr.9: 127808372-127808372 on Build GRCh38
rs973085448 Chr.9: 127808411-127808411 on Build GRCh38
rs770023397 Chr.9: 127808535-127808535 on Build GRCh38
rs370916627 Chr.9: 127808336-127808336 on Build GRCh38
rs763326521 Chr.9: 127808537-127808537 on Build GRCh38
rs749901700 Chr.9: 127808220-127808220 on Build GRCh38
rs780082215 Chr.9: 127808218-127808220 on Build GRCh38
rs41307463 Chr.9: 127808359-127808359 on Build GRCh38
rs759152324 Chr.9: 127808577-127808577 on Build GRCh38
rs764618351 Chr.9: 127808337-127808337 on Build GRCh38
rs780528787 Chr.9: 127808360-127808360 on Build GRCh38
rs1038958808 Chr.9: 127808387-127808387 on Build GRCh38
rs553384159 Chr.9: 127808281-127808281 on Build GRCh38
rs774688472 Chr.9: 127808632-127808632 on Build GRCh38
rs373230782 Chr.9: 127808653-127808653 on Build GRCh38
rs965203643 Chr.9: 127808643-127808643 on Build GRCh38
rs748096485 Chr.9: 127808251-127808251 on Build GRCh38
rs865879157 Chr.9: 127808445-127808445 on Build GRCh38
rs1001400806 Chr.9: 127808470-127808470 on Build GRCh38
rs781407616 Chr.9: 127808523-127808523 on Build GRCh38
rs758155866 Chr.9: 127808580-127808580 on Build GRCh38
rs541013836 Chr.9: 127808294-127808294 on Build GRCh38
rs763406812 Chr.9: 127808335-127808335 on Build GRCh38
rs959361425 Chr.9: 127808287-127808287 on Build GRCh38
rs902728453 Chr.9: 127808542-127808542 on Build GRCh38
rs904026321 Chr.9: 127808463-127808463 on Build GRCh38
rs532279179 Chr.9: 127808431-127808431 on Build GRCh38
rs149509788 Chr.9: 127808593-127808593 on Build GRCh38
rs753209026 Chr.9: 127808212-127808212 on Build GRCh38
rs111530564 Chr.9: 127808425-127808425 on Build GRCh38
rs372883379 Chr.9: 127808594-127808594 on Build GRCh38
rs941830153 Chr.9: 127808381-127808381 on Build GRCh38
rs768512887 Chr.9: 127808654-127808654 on Build GRCh38
rs779629685 Chr.9: 127808228-127808228 on Build GRCh38
rs769926008 Chr.9: 127808608-127808608 on Build GRCh38
rs778768325 Chr.9: 127808238-127808238 on Build GRCh38
rs756148731 Chr.9: 127808693-127808693 on Build GRCh38
rs776150249 Chr.9: 127808644-127808644 on Build GRCh38
rs10987744 Chr.9: 127808453-127808453 on Build GRCh38
rs753558472 Chr.9: 127808348-127808348 on Build GRCh38
rs772024551 Chr.9: 127808260-127808260 on Build GRCh38
rs200345662 Chr.9: 127808322-127808322 on Build GRCh38
rs151191184 Chr.9: 127808509-127808509 on Build GRCh38
rs768686209 Chr.9: 127808599-127808600 on Build GRCh38
rs764029343 Chr.9: 127808668-127808668 on Build GRCh38
rs1001276226 Chr.9: 127808557-127808557 on Build GRCh38
rs751362851 Chr.9: 127808210-127808213 on Build GRCh38
rs375100718 Chr.9: 127808605-127808605 on Build GRCh38
rs769079401 Chr.9: 127808622-127808622 on Build GRCh38
rs921813967 Chr.9: 127808389-127808389 on Build GRCh38
rs747047353 Chr.9: 127808288-127808288 on Build GRCh38
rs745917370 Chr.9: 127808607-127808607 on Build GRCh38
rs148686790 Chr.9: 127808650-127808650 on Build GRCh38
rs771301592 Chr.9: 127808295-127808295 on Build GRCh38
rs765935827 Chr.9: 127808568-127808568 on Build GRCh38
rs747088361 Chr.9: 127808510-127808510 on Build GRCh38
rs370634067 Chr.9: 127808206-127808206 on Build GRCh38
rs760063021 Chr.9: 127808407-127808407 on Build GRCh38
rs373153022 Chr.9: 127808432-127808432 on Build GRCh38
rs749079922 Chr.9: 127808606-127808606 on Build GRCh38
rs757288134 Chr.9: 127808677-127808677 on Build GRCh38
rs778622701 Chr.9: 127808357-127808357 on Build GRCh38
rs118102203 Chr.9: 127808330-127808330 on Build GRCh38
rs779995758 Chr.9: 127808696-127808696 on Build GRCh38
rs762360733 Chr.9: 127808633-127808633 on Build GRCh38
rs754665279 Chr.9: 127808236-127808236 on Build GRCh38
rs755791295 Chr.9: 127808226-127808226 on Build GRCh38
rs756432070 Chr.9: 127808209-127808210 on Build GRCh38
rs760116355 Chr.9: 127808344-127808344 on Build GRCh38
rs774776300 Chr.9: 127808546-127808546 on Build GRCh38
rs749955008 Chr.9: 127808338-127808338 on Build GRCh38
rs539980676 Chr.9: 127808675-127808675 on Build GRCh38
rs559365787 Chr.9: 127808434-127808434 on Build GRCh38
rs763542427 Chr.9: 127808430-127808430 on Build GRCh38
rs780261977 Chr.9: 127808611-127808611 on Build GRCh38
rs759825513 Chr.9: 127808328-127808328 on Build GRCh38
rs144175156 Chr.9: 127808647-127808647 on Build GRCh38
rs142234881 Chr.9: 127808375-127808375 on Build GRCh38
rs753327063 Chr.9: 127808574-127808574 on Build GRCh38
rs563131884 Chr.9: 127808419-127808419 on Build GRCh38
rs574368125 Chr.9: 127808362-127808362 on Build GRCh38
rs115912190 Chr.9: 127808231-127808231 on Build GRCh38
rs749584122 Chr.9: 127808699-127808699 on Build GRCh38
rs754649891 Chr.9: 127808355-127808355 on Build GRCh38
rs775901209 Chr.9: 127808332-127808332 on Build GRCh38
rs772537574 Chr.9: 127808642-127808642 on Build GRCh38
rs755174848 Chr.9: 127808700-127808700 on Build GRCh38
rs775491551 Chr.9: 127808597-127808597 on Build GRCh38
rs2230270 Chr.9: 127808615-127808615 on Build GRCh38
rs752458887 Chr.9: 127808578-127808578 on Build GRCh38
rs750479758 Chr.9: 127808691-127808691 on Build GRCh38
rs746125350 Chr.9: 127808526-127808526 on Build GRCh38

View Full Product Details