Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127814933-127815431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENG
Entrez Gene ID
2022
Gene Name
endoglin
Gene Aliases
END, HHT1, ORW1
UniGene
Hs.76753
Gene Chromosome Location
Chr.9: 127815012-127854773 on Build GRCh38
Transcripts
NM_000118.3 NM_001114753.2 NM_001278138.1

SNP Detail

SNP ID Chromosome Location
rs941181607 Chr.9: 127815011-127815011 on Build GRCh38
rs41514846 Chr.9: 127815214-127815217 on Build GRCh38
rs770303011 Chr.9: 127815333-127815333 on Build GRCh38
rs1015027303 Chr.9: 127814971-127814971 on Build GRCh38
rs745464567 Chr.9: 127815396-127815396 on Build GRCh38
rs569243404 Chr.9: 127815338-127815338 on Build GRCh38
rs971311800 Chr.9: 127815306-127815306 on Build GRCh38
rs549011956 Chr.9: 127815251-127815251 on Build GRCh38
rs41478248 Chr.9: 127815362-127815362 on Build GRCh38
rs1054104291 Chr.9: 127815147-127815147 on Build GRCh38
rs893984357 Chr.9: 127815148-127815148 on Build GRCh38
rs953652893 Chr.9: 127815340-127815340 on Build GRCh38
rs763238389 Chr.9: 127815289-127815289 on Build GRCh38
rs1029698984 Chr.9: 127815231-127815231 on Build GRCh38
rs779334540 Chr.9: 127815259-127815259 on Build GRCh38
rs991371974 Chr.9: 127815294-127815294 on Build GRCh38
rs576252697 Chr.9: 127814998-127814998 on Build GRCh38
rs190129922 Chr.9: 127815191-127815191 on Build GRCh38
rs746493910 Chr.9: 127815316-127815316 on Build GRCh38
rs143099696 Chr.9: 127815124-127815124 on Build GRCh38
rs780329814 Chr.9: 127815051-127815051 on Build GRCh38
rs141125778 Chr.9: 127815279-127815279 on Build GRCh38
rs529337078 Chr.9: 127815233-127815233 on Build GRCh38
rs1045413208 Chr.9: 127815096-127815096 on Build GRCh38
rs544896906 Chr.9: 127815118-127815118 on Build GRCh38
rs962297713 Chr.9: 127814972-127814972 on Build GRCh38
rs995059042 Chr.9: 127815333-127815334 on Build GRCh38
rs1004320905 Chr.9: 127815245-127815245 on Build GRCh38
rs894147346 Chr.9: 127815228-127815228 on Build GRCh38
rs991818524 Chr.9: 127815018-127815018 on Build GRCh38
rs760085561 Chr.9: 127815169-127815169 on Build GRCh38
rs148192743 Chr.9: 127815190-127815190 on Build GRCh38
rs953469902 Chr.9: 127815040-127815040 on Build GRCh38
rs981311217 Chr.9: 127815336-127815336 on Build GRCh38
rs971160067 Chr.9: 127814962-127814962 on Build GRCh38
rs773530203 Chr.9: 127815037-127815039 on Build GRCh38
rs569902440 Chr.9: 127814955-127814955 on Build GRCh38
rs138178682 Chr.9: 127815409-127815409 on Build GRCh38
rs34691816 Chr.9: 127815380-127815381 on Build GRCh38
rs902837678 Chr.9: 127815101-127815101 on Build GRCh38
rs1018935323 Chr.9: 127815368-127815368 on Build GRCh38
rs72616667 Chr.9: 127815159-127815159 on Build GRCh38
rs934304856 Chr.9: 127815356-127815356 on Build GRCh38
rs10264 Chr.9: 127815041-127815041 on Build GRCh38
rs973595276 Chr.9: 127814987-127814987 on Build GRCh38
rs950002211 Chr.9: 127815068-127815068 on Build GRCh38
rs915796790 Chr.9: 127815053-127815053 on Build GRCh38
rs1003706334 Chr.9: 127814969-127814969 on Build GRCh38
rs1018947844 Chr.9: 127815166-127815166 on Build GRCh38
rs915728475 Chr.9: 127815295-127815295 on Build GRCh38

View Full Product Details