Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127866403-127866931 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK1
Entrez Gene ID
203
Gene Name
adenylate kinase 1
Gene Aliases
HTL-S-58j
UniGene
Hs.175473
Gene Chromosome Location
Chr.9: 127866480-127886749 on Build GRCh38
Transcripts
NM_000476.2 NM_001318121.1 NM_001318122.1

SNP Detail

SNP ID Chromosome Location
rs886975727 Chr.9: 127866650-127866650 on Build GRCh38
rs182437286 Chr.9: 127866570-127866570 on Build GRCh38
rs551949292 Chr.9: 127866781-127866781 on Build GRCh38
rs913987 Chr.9: 127866860-127866860 on Build GRCh38
rs867890792 Chr.9: 127866505-127866505 on Build GRCh38
rs931744654 Chr.9: 127866627-127866627 on Build GRCh38
rs1037273092 Chr.9: 127866528-127866528 on Build GRCh38
rs563017433 Chr.9: 127866445-127866445 on Build GRCh38
rs1029305631 Chr.9: 127866856-127866856 on Build GRCh38
rs538398363 Chr.9: 127866780-127866780 on Build GRCh38
rs978653678 Chr.9: 127866840-127866840 on Build GRCh38
rs565533878 Chr.9: 127866503-127866503 on Build GRCh38
rs764962840 Chr.9: 127866836-127866836 on Build GRCh38
rs895887136 Chr.9: 127866783-127866783 on Build GRCh38
rs560200284 Chr.9: 127866785-127866785 on Build GRCh38
rs569421825 Chr.9: 127866754-127866754 on Build GRCh38
rs988636212 Chr.9: 127866571-127866571 on Build GRCh38
rs541017076 Chr.9: 127866769-127866769 on Build GRCh38
rs913430216 Chr.9: 127866575-127866575 on Build GRCh38
rs1043326781 Chr.9: 127866582-127866582 on Build GRCh38
rs540192824 Chr.9: 127866572-127866572 on Build GRCh38
rs1037157374 Chr.9: 127866724-127866724 on Build GRCh38
rs1048394435 Chr.9: 127866759-127866759 on Build GRCh38
rs536667729 Chr.9: 127866890-127866890 on Build GRCh38
rs1020097537 Chr.9: 127866835-127866835 on Build GRCh38
rs911682801 Chr.9: 127866887-127866887 on Build GRCh38
rs911615355 Chr.9: 127866638-127866638 on Build GRCh38
rs987533381 Chr.9: 127866882-127866882 on Build GRCh38
rs921571688 Chr.9: 127866610-127866610 on Build GRCh38
rs962058344 Chr.9: 127866909-127866909 on Build GRCh38
rs946209931 Chr.9: 127866577-127866577 on Build GRCh38
rs746399990 Chr.9: 127866502-127866502 on Build GRCh38
rs963928274 Chr.9: 127866719-127866719 on Build GRCh38
rs545952308 Chr.9: 127866605-127866606 on Build GRCh38
rs1055771863 Chr.9: 127866617-127866617 on Build GRCh38
rs944183361 Chr.9: 127866475-127866475 on Build GRCh38
rs534596319 Chr.9: 127866820-127866820 on Build GRCh38
rs753649922 Chr.9: 127866635-127866635 on Build GRCh38
rs529721048 Chr.9: 127866600-127866600 on Build GRCh38
rs968586068 Chr.9: 127866592-127866592 on Build GRCh38
rs987260239 Chr.9: 127866628-127866628 on Build GRCh38
rs531909283 Chr.9: 127866448-127866448 on Build GRCh38
rs975072022 Chr.9: 127866520-127866520 on Build GRCh38
rs905950657 Chr.9: 127866560-127866560 on Build GRCh38
rs762490 Chr.9: 127866460-127866460 on Build GRCh38
rs187001682 Chr.9: 127866821-127866821 on Build GRCh38
rs1056433559 Chr.9: 127866859-127866859 on Build GRCh38
rs978868244 Chr.9: 127866789-127866790 on Build GRCh38
rs866078332 Chr.9: 127866720-127866720 on Build GRCh38
rs978434190 Chr.9: 127866593-127866593 on Build GRCh38
rs1016868533 Chr.9: 127866704-127866704 on Build GRCh38
rs528971918 Chr.9: 127866446-127866447 on Build GRCh38
rs77787745 Chr.9: 127866776-127866776 on Build GRCh38
rs73669434 Chr.9: 127866452-127866452 on Build GRCh38
rs897402601 Chr.9: 127866547-127866547 on Build GRCh38
rs995505204 Chr.9: 127866551-127866551 on Build GRCh38
rs1036166386 Chr.9: 127866569-127866569 on Build GRCh38
rs548111211 Chr.9: 127866562-127866562 on Build GRCh38
rs913649497 Chr.9: 127866777-127866777 on Build GRCh38
rs1005387142 Chr.9: 127866669-127866669 on Build GRCh38
rs897414057 Chr.9: 127866728-127866728 on Build GRCh38
rs950088062 Chr.9: 127866876-127866876 on Build GRCh38
rs926223836 Chr.9: 127866801-127866801 on Build GRCh38
rs996933905 Chr.9: 127866721-127866721 on Build GRCh38
rs1050362815 Chr.9: 127866459-127866459 on Build GRCh38
rs768038369 Chr.9: 127866658-127866658 on Build GRCh38
rs189027138 Chr.9: 127866557-127866557 on Build GRCh38
rs1026136971 Chr.9: 127866647-127866647 on Build GRCh38
rs549576089 Chr.9: 127866743-127866743 on Build GRCh38

View Full Product Details