Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127871681-127872099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK1
Entrez Gene ID
203
Gene Name
adenylate kinase 1
Gene Aliases
HTL-S-58j
UniGene
Hs.175473
Gene Chromosome Location
Chr.9: 127866480-127886749 on Build GRCh38
Transcripts
NM_000476.2 NM_001318121.1 NM_001318122.1 XM_017014427.1 XM_017014428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1105427 c.323G>C p.R108P Chr.9: 127871824-127871824 on Build GRCh38
4138834 c.316G>A p.E106K Chr.9: 127871831-127871831 on Build GRCh38
3326164 c.296T>G p.V99G Chr.9: 127871851-127871851 on Build GRCh38
3326165 c.289C>T p.R97W Chr.9: 127871858-127871858 on Build GRCh38
3699437 c.323G>T p.R108L Chr.9: 127871824-127871824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779559662 Chr.9: 127871953-127871953 on Build GRCh38
rs751629167 Chr.9: 127871807-127871807 on Build GRCh38
rs368912370 Chr.9: 127871928-127871928 on Build GRCh38
rs1055659277 Chr.9: 127871755-127871755 on Build GRCh38
rs139396377 Chr.9: 127871776-127871776 on Build GRCh38
rs968392001 Chr.9: 127871735-127871735 on Build GRCh38
rs746592337 Chr.9: 127871881-127871881 on Build GRCh38
rs762906771 Chr.9: 127871801-127871801 on Build GRCh38
rs767658405 Chr.9: 127871910-127871910 on Build GRCh38
rs77423462 Chr.9: 127871941-127871941 on Build GRCh38
rs73669436 Chr.9: 127871901-127871901 on Build GRCh38
rs913440524 Chr.9: 127871951-127871951 on Build GRCh38
rs895691863 Chr.9: 127872074-127872074 on Build GRCh38
rs904218627 Chr.9: 127872000-127872000 on Build GRCh38
rs745471497 Chr.9: 127871894-127871894 on Build GRCh38
rs931317223 Chr.9: 127871859-127871859 on Build GRCh38
rs768549785 Chr.9: 127871962-127871962 on Build GRCh38
rs774424327 Chr.9: 127871907-127871907 on Build GRCh38
rs999860360 Chr.9: 127872050-127872050 on Build GRCh38
rs751000982 Chr.9: 127871973-127871973 on Build GRCh38
rs979146671 Chr.9: 127871960-127871960 on Build GRCh38
rs368918727 Chr.9: 127872063-127872063 on Build GRCh38
rs761886813 Chr.9: 127871860-127871860 on Build GRCh38
rs371668364 Chr.9: 127871943-127871943 on Build GRCh38
rs767491704 Chr.9: 127871933-127871934 on Build GRCh38
rs757397992 Chr.9: 127871824-127871824 on Build GRCh38
rs558447135 Chr.9: 127871729-127871729 on Build GRCh38
rs780427267 Chr.9: 127871852-127871852 on Build GRCh38
rs755873143 Chr.9: 127872053-127872053 on Build GRCh38
rs944267738 Chr.9: 127871780-127871780 on Build GRCh38
rs750087402 Chr.9: 127871914-127871914 on Build GRCh38
rs368949848 Chr.9: 127871856-127871856 on Build GRCh38
rs1028689944 Chr.9: 127871756-127871756 on Build GRCh38
rs937438628 Chr.9: 127872025-127872025 on Build GRCh38
rs774452787 Chr.9: 127871965-127871965 on Build GRCh38
rs375122190 Chr.9: 127871778-127871778 on Build GRCh38
rs104894102 Chr.9: 127871828-127871828 on Build GRCh38
rs749173887 Chr.9: 127871956-127871956 on Build GRCh38
rs1020346247 Chr.9: 127871705-127871705 on Build GRCh38
rs375921903 Chr.9: 127871917-127871917 on Build GRCh38
rs1009937 Chr.9: 127872005-127872005 on Build GRCh38
rs144171533 Chr.9: 127871899-127871899 on Build GRCh38
rs779381063 Chr.9: 127871860-127871860 on Build GRCh38
rs756778564 Chr.9: 127871942-127871942 on Build GRCh38
rs563055475 Chr.9: 127872048-127872048 on Build GRCh38
rs201062806 Chr.9: 127871948-127871948 on Build GRCh38
rs375653403 Chr.9: 127871855-127871855 on Build GRCh38
rs769429363 Chr.9: 127871952-127871952 on Build GRCh38
rs750642408 Chr.9: 127871911-127871911 on Build GRCh38
rs764274408 Chr.9: 127871802-127871802 on Build GRCh38
rs1055881600 Chr.9: 127872070-127872070 on Build GRCh38
rs768770372 Chr.9: 127871906-127871906 on Build GRCh38
rs987683798 Chr.9: 127871931-127871931 on Build GRCh38
rs753177103 Chr.9: 127871939-127871939 on Build GRCh38
rs978945513 Chr.9: 127871736-127871736 on Build GRCh38
rs886782745 Chr.9: 127871810-127871810 on Build GRCh38
rs147616730 Chr.9: 127871893-127871893 on Build GRCh38
rs745510283 Chr.9: 127871945-127871945 on Build GRCh38
rs879454091 Chr.9: 127872004-127872004 on Build GRCh38
rs779439692 Chr.9: 127871929-127871929 on Build GRCh38
rs751605963 Chr.9: 127871776-127871777 on Build GRCh38
rs766671097 Chr.9: 127871918-127871918 on Build GRCh38
rs937206860 Chr.9: 127871745-127871745 on Build GRCh38
rs755458590 Chr.9: 127871858-127871858 on Build GRCh38
rs770394256 Chr.9: 127871890-127871890 on Build GRCh38
rs765054939 Chr.9: 127871786-127871786 on Build GRCh38
rs954249292 Chr.9: 127871761-127871761 on Build GRCh38
rs761804052 Chr.9: 127871988-127871988 on Build GRCh38
rs767893823 Chr.9: 127871827-127871827 on Build GRCh38
rs780711229 Chr.9: 127871944-127871944 on Build GRCh38
rs201099728 Chr.9: 127871796-127871796 on Build GRCh38
rs372248359 Chr.9: 127871909-127871909 on Build GRCh38
rs769503890 Chr.9: 127871895-127871895 on Build GRCh38
rs372237345 Chr.9: 127871829-127871829 on Build GRCh38

View Full Product Details