Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127877684-127878227 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AK1
Entrez Gene ID
203
Gene Name
adenylate kinase 1
Gene Aliases
HTL-S-58j
UniGene
Hs.175473
Gene Chromosome Location
Chr.9: 127866480-127886749 on Build GRCh38
Transcripts
NM_000476.2 NM_001318121.1 XM_017014427.1 XM_017014428.1

SNP Detail

SNP ID Chromosome Location
rs768847194 Chr.9: 127878177-127878177 on Build GRCh38
rs779611078 Chr.9: 127878170-127878170 on Build GRCh38
rs543245168 Chr.9: 127878084-127878084 on Build GRCh38
rs1005930441 Chr.9: 127878124-127878124 on Build GRCh38
rs912279129 Chr.9: 127878165-127878165 on Build GRCh38
rs925656489 Chr.9: 127877919-127877919 on Build GRCh38
rs1051286574 Chr.9: 127877736-127877736 on Build GRCh38
rs971282509 Chr.9: 127877885-127877885 on Build GRCh38
rs569938131 Chr.9: 127877758-127877758 on Build GRCh38
rs952680473 Chr.9: 127877772-127877772 on Build GRCh38
rs984259610 Chr.9: 127877805-127877805 on Build GRCh38
rs994452784 Chr.9: 127878048-127878048 on Build GRCh38
rs920980025 Chr.9: 127878038-127878038 on Build GRCh38
rs944844344 Chr.9: 127877771-127877771 on Build GRCh38
rs35466810 Chr.9: 127877818-127877818 on Build GRCh38
rs1041588777 Chr.9: 127878027-127878027 on Build GRCh38
rs114398415 Chr.9: 127878203-127878203 on Build GRCh38
rs13290745 Chr.9: 127877812-127877812 on Build GRCh38
rs1041920533 Chr.9: 127878194-127878194 on Build GRCh38
rs1034807488 Chr.9: 127877809-127877809 on Build GRCh38
rs1015354559 Chr.9: 127877938-127877938 on Build GRCh38
rs534267399 Chr.9: 127877916-127877916 on Build GRCh38
rs756088401 Chr.9: 127877933-127877933 on Build GRCh38
rs79093871 Chr.9: 127877876-127877876 on Build GRCh38
rs935663583 Chr.9: 127877926-127877926 on Build GRCh38
rs187948230 Chr.9: 127878155-127878155 on Build GRCh38
rs986597103 Chr.9: 127878134-127878134 on Build GRCh38
rs993040650 Chr.9: 127877834-127877834 on Build GRCh38
rs533790398 Chr.9: 127877763-127877763 on Build GRCh38
rs556840867 Chr.9: 127878093-127878093 on Build GRCh38
rs903455953 Chr.9: 127877811-127877811 on Build GRCh38
rs13290737 Chr.9: 127877797-127877797 on Build GRCh38
rs574244308 Chr.9: 127878082-127878082 on Build GRCh38
rs901397391 Chr.9: 127877743-127877743 on Build GRCh38
rs553755979 Chr.9: 127878135-127878135 on Build GRCh38
rs754001213 Chr.9: 127878032-127878032 on Build GRCh38
rs13291054 Chr.9: 127877875-127877875 on Build GRCh38
rs13291025 Chr.9: 127877796-127877796 on Build GRCh38
rs910261652 Chr.9: 127877967-127877967 on Build GRCh38
rs917182820 Chr.9: 127877849-127877849 on Build GRCh38
rs1024120629 Chr.9: 127878173-127878173 on Build GRCh38
rs538594958 Chr.9: 127877808-127877808 on Build GRCh38
rs530601015 Chr.9: 127878080-127878080 on Build GRCh38
rs2774265 Chr.9: 127878166-127878166 on Build GRCh38
rs959017896 Chr.9: 127878143-127878143 on Build GRCh38
rs894425767 Chr.9: 127877854-127877854 on Build GRCh38
rs558114757 Chr.9: 127877820-127877820 on Build GRCh38
rs780237268 Chr.9: 127878026-127878026 on Build GRCh38
rs888449474 Chr.9: 127878106-127878106 on Build GRCh38
rs967290363 Chr.9: 127877858-127877858 on Build GRCh38
rs994190755 Chr.9: 127877746-127877746 on Build GRCh38

View Full Product Details