Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127935398-127935900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPM2
Entrez Gene ID
8818
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit
Gene Aliases
CDG1U
UniGene
Hs.108973
Gene Chromosome Location
Chr.9: 127935095-127938484 on Build GRCh38
Transcripts
NM_003863.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5505039 c.213T>C p.Y71Y Chr.9: 127935764-127935764 on Build GRCh38
6648876 c.231G>A p.K77K Chr.9: 127935746-127935746 on Build GRCh38
3952354 c.225G>A p.K75K Chr.9: 127935752-127935752 on Build GRCh38
3982740 c.227C>G p.T76S Chr.9: 127935750-127935750 on Build GRCh38
5948902 c.254G>T p.*85L Chr.9: 127935723-127935723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866378101 Chr.9: 127935693-127935693 on Build GRCh38
rs1009546130 Chr.9: 127935436-127935436 on Build GRCh38
rs773384426 Chr.9: 127935854-127935854 on Build GRCh38
rs753375016 Chr.9: 127935721-127935721 on Build GRCh38
rs568108561 Chr.9: 127935737-127935737 on Build GRCh38
rs780096109 Chr.9: 127935432-127935432 on Build GRCh38
rs867390491 Chr.9: 127935502-127935502 on Build GRCh38
rs889404680 Chr.9: 127935433-127935433 on Build GRCh38
rs1020885625 Chr.9: 127935875-127935875 on Build GRCh38
rs6781 Chr.9: 127935764-127935764 on Build GRCh38
rs759597928 Chr.9: 127935769-127935769 on Build GRCh38
rs1002680153 Chr.9: 127935800-127935800 on Build GRCh38
rs935700038 Chr.9: 127935503-127935503 on Build GRCh38
rs34734482 Chr.9: 127935856-127935857 on Build GRCh38
rs962270657 Chr.9: 127935596-127935596 on Build GRCh38
rs953468134 Chr.9: 127935679-127935679 on Build GRCh38
rs925041268 Chr.9: 127935606-127935606 on Build GRCh38
rs1003690875 Chr.9: 127935592-127935592 on Build GRCh38
rs754599759 Chr.9: 127935725-127935725 on Build GRCh38
rs776591740 Chr.9: 127935765-127935765 on Build GRCh38
rs894257268 Chr.9: 127935531-127935531 on Build GRCh38
rs952108629 Chr.9: 127935485-127935485 on Build GRCh38
rs924499723 Chr.9: 127935463-127935463 on Build GRCh38
rs775806368 Chr.9: 127935796-127935796 on Build GRCh38
rs886063482 Chr.9: 127935785-127935785 on Build GRCh38
rs965861975 Chr.9: 127935763-127935763 on Build GRCh38
rs994877531 Chr.9: 127935614-127935614 on Build GRCh38
rs1033554698 Chr.9: 127935827-127935827 on Build GRCh38
rs747964051 Chr.9: 127935731-127935731 on Build GRCh38
rs932364068 Chr.9: 127935658-127935658 on Build GRCh38
rs991958222 Chr.9: 127935863-127935863 on Build GRCh38
rs1015198471 Chr.9: 127935593-127935593 on Build GRCh38
rs751967535 Chr.9: 127935809-127935809 on Build GRCh38
rs960679134 Chr.9: 127935533-127935533 on Build GRCh38
rs764595439 Chr.9: 127935804-127935804 on Build GRCh38
rs201974273 Chr.9: 127935714-127935714 on Build GRCh38
rs566391229 Chr.9: 127935535-127935535 on Build GRCh38
rs534573021 Chr.9: 127935587-127935587 on Build GRCh38
rs769557610 Chr.9: 127935427-127935427 on Build GRCh38
rs763247539 Chr.9: 127935801-127935801 on Build GRCh38
rs867450362 Chr.9: 127935546-127935546 on Build GRCh38
rs148371368 Chr.9: 127935773-127935773 on Build GRCh38
rs960812152 Chr.9: 127935856-127935856 on Build GRCh38
rs1033916308 Chr.9: 127935525-127935525 on Build GRCh38
rs866992869 Chr.9: 127935545-127935545 on Build GRCh38
rs768897006 Chr.9: 127935715-127935715 on Build GRCh38
rs935919419 Chr.9: 127935785-127935785 on Build GRCh38
rs372162940 Chr.9: 127935468-127935468 on Build GRCh38
rs897822609 Chr.9: 127935445-127935445 on Build GRCh38
rs7997 Chr.9: 127935750-127935750 on Build GRCh38
rs774687931 Chr.9: 127935741-127935741 on Build GRCh38
rs1018940950 Chr.9: 127935444-127935444 on Build GRCh38
rs992481008 Chr.9: 127935551-127935551 on Build GRCh38
rs1050640242 Chr.9: 127935703-127935703 on Build GRCh38
rs376186668 Chr.9: 127935821-127935821 on Build GRCh38
rs564981501 Chr.9: 127935504-127935504 on Build GRCh38
rs945031872 Chr.9: 127935560-127935560 on Build GRCh38
rs763370572 Chr.9: 127935699-127935699 on Build GRCh38
rs778557558 Chr.9: 127935729-127935729 on Build GRCh38
rs886063481 Chr.9: 127935673-127935673 on Build GRCh38
rs34405428 Chr.9: 127935714-127935715 on Build GRCh38
rs913601751 Chr.9: 127935556-127935556 on Build GRCh38
rs993427887 Chr.9: 127935451-127935451 on Build GRCh38

View Full Product Details