Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127938033-127938545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPM2
Entrez Gene ID
8818
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 2, regulatory subunit
Gene Aliases
CDG1U
UniGene
Hs.108973
Gene Chromosome Location
Chr.9: 127935095-127938484 on Build GRCh38
Transcripts
NM_003863.3

SNP Detail

SNP ID Chromosome Location
rs559768401 Chr.9: 127938091-127938091 on Build GRCh38
rs566910080 Chr.9: 127938407-127938407 on Build GRCh38
rs1017161856 Chr.9: 127938193-127938193 on Build GRCh38
rs3758327 Chr.9: 127938448-127938448 on Build GRCh38
rs185263072 Chr.9: 127938344-127938344 on Build GRCh38
rs1029721171 Chr.9: 127938142-127938142 on Build GRCh38
rs984396634 Chr.9: 127938198-127938198 on Build GRCh38
rs1011556760 Chr.9: 127938119-127938119 on Build GRCh38
rs557999431 Chr.9: 127938505-127938505 on Build GRCh38
rs992817674 Chr.9: 127938151-127938151 on Build GRCh38
rs1052639568 Chr.9: 127938337-127938337 on Build GRCh38
rs571582553 Chr.9: 127938250-127938250 on Build GRCh38
rs900070590 Chr.9: 127938370-127938370 on Build GRCh38
rs1004704382 Chr.9: 127938432-127938432 on Build GRCh38
rs998154622 Chr.9: 127938380-127938380 on Build GRCh38
rs886063486 Chr.9: 127938139-127938139 on Build GRCh38
rs145442130 Chr.9: 127938437-127938440 on Build GRCh38
rs547777622 Chr.9: 127938510-127938510 on Build GRCh38
rs947022375 Chr.9: 127938352-127938352 on Build GRCh38
rs962872503 Chr.9: 127938451-127938451 on Build GRCh38
rs982608840 Chr.9: 127938178-127938178 on Build GRCh38
rs971272091 Chr.9: 127938506-127938506 on Build GRCh38
rs924367795 Chr.9: 127938321-127938321 on Build GRCh38
rs962698694 Chr.9: 127938199-127938199 on Build GRCh38
rs922510404 Chr.9: 127938261-127938261 on Build GRCh38
rs935502735 Chr.9: 127938060-127938060 on Build GRCh38
rs575777173 Chr.9: 127938488-127938488 on Build GRCh38
rs555607108 Chr.9: 127938466-127938466 on Build GRCh38
rs1034632132 Chr.9: 127938426-127938426 on Build GRCh38
rs1001794505 Chr.9: 127938112-127938112 on Build GRCh38
rs942503523 Chr.9: 127938232-127938232 on Build GRCh38
rs746237087 Chr.9: 127938322-127938322 on Build GRCh38
rs375582451 Chr.9: 127938266-127938266 on Build GRCh38
rs899836755 Chr.9: 127938133-127938133 on Build GRCh38
rs951493665 Chr.9: 127938187-127938187 on Build GRCh38
rs545477837 Chr.9: 127938140-127938140 on Build GRCh38
rs759478797 Chr.9: 127938193-127938194 on Build GRCh38
rs949705217 Chr.9: 127938256-127938256 on Build GRCh38
rs1053002123 Chr.9: 127938088-127938088 on Build GRCh38
rs998107942 Chr.9: 127938139-127938139 on Build GRCh38
rs1052046144 Chr.9: 127938331-127938331 on Build GRCh38
rs112813338 Chr.9: 127938202-127938202 on Build GRCh38
rs1040368266 Chr.9: 127938369-127938369 on Build GRCh38
rs548845259 Chr.9: 127938166-127938166 on Build GRCh38
rs1014304720 Chr.9: 127938434-127938434 on Build GRCh38
rs981077896 Chr.9: 127938299-127938299 on Build GRCh38
rs886063487 Chr.9: 127938432-127938432 on Build GRCh38
rs531435619 Chr.9: 127938214-127938214 on Build GRCh38
rs190075729 Chr.9: 127938398-127938398 on Build GRCh38
rs1025643123 Chr.9: 127938440-127938441 on Build GRCh38
rs892241910 Chr.9: 127938340-127938340 on Build GRCh38
rs192275912 Chr.9: 127938249-127938249 on Build GRCh38
rs951439372 Chr.9: 127938153-127938153 on Build GRCh38
rs951425710 Chr.9: 127938443-127938446 on Build GRCh38
rs771081688 Chr.9: 127938231-127938231 on Build GRCh38
rs894085027 Chr.9: 127938104-127938104 on Build GRCh38
rs1010662067 Chr.9: 127938343-127938343 on Build GRCh38
rs528993503 Chr.9: 127938129-127938129 on Build GRCh38

View Full Product Details