Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127942412-127942910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102A
Entrez Gene ID
399665
Gene Name
family with sequence similarity 102 member A
Gene Aliases
C9orf132, EEIG1, SYM-3A, bA203J24.7
UniGene
Hs.535972
Gene Chromosome Location
Chr.9: 127940582-127980533 on Build GRCh38
Transcripts
NM_001035254.2 NM_203305.2

SNP Detail

SNP ID Chromosome Location
rs754215279 Chr.9: 127942705-127942705 on Build GRCh38
rs957881383 Chr.9: 127942884-127942884 on Build GRCh38
rs552889075 Chr.9: 127942494-127942494 on Build GRCh38
rs1053418203 Chr.9: 127942442-127942442 on Build GRCh38
rs989357985 Chr.9: 127942691-127942691 on Build GRCh38
rs901909501 Chr.9: 127942699-127942699 on Build GRCh38
rs554933923 Chr.9: 127942887-127942887 on Build GRCh38
rs2296704 Chr.9: 127942728-127942728 on Build GRCh38
rs952190078 Chr.9: 127942833-127942833 on Build GRCh38
rs971201578 Chr.9: 127942626-127942626 on Build GRCh38
rs560105451 Chr.9: 127942504-127942504 on Build GRCh38
rs34658008 Chr.9: 127942500-127942501 on Build GRCh38
rs930904455 Chr.9: 127942621-127942622 on Build GRCh38
rs1025392172 Chr.9: 127942825-127942825 on Build GRCh38
rs990796409 Chr.9: 127942612-127942612 on Build GRCh38
rs959362845 Chr.9: 127942597-127942597 on Build GRCh38
rs1006354996 Chr.9: 127942732-127942732 on Build GRCh38
rs1022647555 Chr.9: 127942551-127942551 on Build GRCh38
rs943457683 Chr.9: 127942472-127942472 on Build GRCh38
rs943346128 Chr.9: 127942669-127942669 on Build GRCh38
rs780597625 Chr.9: 127942622-127942623 on Build GRCh38
rs36092398 Chr.9: 127942457-127942457 on Build GRCh38
rs1026882714 Chr.9: 127942563-127942563 on Build GRCh38
rs951038306 Chr.9: 127942572-127942572 on Build GRCh38
rs3818510 Chr.9: 127942696-127942696 on Build GRCh38
rs929714761 Chr.9: 127942710-127942710 on Build GRCh38
rs984967711 Chr.9: 127942635-127942638 on Build GRCh38
rs998686696 Chr.9: 127942544-127942545 on Build GRCh38
rs879302263 Chr.9: 127942729-127942730 on Build GRCh38
rs750529163 Chr.9: 127942613-127942613 on Build GRCh38
rs889455779 Chr.9: 127942478-127942478 on Build GRCh38
rs539851592 Chr.9: 127942550-127942550 on Build GRCh38
rs530621980 Chr.9: 127942680-127942680 on Build GRCh38
rs1016491961 Chr.9: 127942596-127942596 on Build GRCh38
rs895386275 Chr.9: 127942792-127942792 on Build GRCh38
rs537642445 Chr.9: 127942757-127942757 on Build GRCh38
rs893058836 Chr.9: 127942545-127942545 on Build GRCh38
rs1052952434 Chr.9: 127942704-127942706 on Build GRCh38
rs1012276460 Chr.9: 127942806-127942806 on Build GRCh38
rs745379088 Chr.9: 127942846-127942846 on Build GRCh38
rs936473268 Chr.9: 127942690-127942690 on Build GRCh38
rs568025002 Chr.9: 127942576-127942576 on Build GRCh38
rs780266615 Chr.9: 127942700-127942700 on Build GRCh38
rs1043060 Chr.9: 127942841-127942841 on Build GRCh38
rs1016544259 Chr.9: 127942756-127942756 on Build GRCh38
rs911111516 Chr.9: 127942694-127942694 on Build GRCh38
rs779142107 Chr.9: 127942869-127942869 on Build GRCh38
rs41308910 Chr.9: 127942561-127942561 on Build GRCh38
rs1011283935 Chr.9: 127942547-127942547 on Build GRCh38
rs146143264 Chr.9: 127942726-127942726 on Build GRCh38
rs748653375 Chr.9: 127942774-127942774 on Build GRCh38
rs1002522598 Chr.9: 127942743-127942743 on Build GRCh38
rs1025085813 Chr.9: 127942620-127942620 on Build GRCh38
rs934528387 Chr.9: 127942703-127942704 on Build GRCh38
rs1040437863 Chr.9: 127942528-127942528 on Build GRCh38
rs373081704 Chr.9: 127942841-127942841 on Build GRCh38
rs374595997 Chr.9: 127942733-127942733 on Build GRCh38
rs767533107 Chr.9: 127942449-127942449 on Build GRCh38
rs989242271 Chr.9: 127942885-127942885 on Build GRCh38
rs910570037 Chr.9: 127942665-127942668 on Build GRCh38
rs972254211 Chr.9: 127942598-127942598 on Build GRCh38
rs900892798 Chr.9: 127942530-127942530 on Build GRCh38
rs923761284 Chr.9: 127942653-127942653 on Build GRCh38
rs977989712 Chr.9: 127942642-127942642 on Build GRCh38
rs769049077 Chr.9: 127942885-127942886 on Build GRCh38
rs1031044382 Chr.9: 127942878-127942878 on Build GRCh38
rs570294608 Chr.9: 127942708-127942708 on Build GRCh38
rs1032030515 Chr.9: 127942545-127942545 on Build GRCh38
rs114873837 Chr.9: 127942837-127942837 on Build GRCh38
rs548173436 Chr.9: 127942564-127942564 on Build GRCh38
rs749775399 Chr.9: 127942763-127942763 on Build GRCh38

View Full Product Details