Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127942809-127943301 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102A
Entrez Gene ID
399665
Gene Name
family with sequence similarity 102 member A
Gene Aliases
C9orf132, EEIG1, SYM-3A, bA203J24.7
UniGene
Hs.535972
Gene Chromosome Location
Chr.9: 127940582-127980533 on Build GRCh38
Transcripts
NM_001035254.2 NM_203305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3779648 c.1115C>G p.S372C Chr.9: 127943232-127943232 on Build GRCh38
4925714 c.1139T>C p.V380A Chr.9: 127943208-127943208 on Build GRCh38
5410457 c.1113C>T p.V371V Chr.9: 127943234-127943234 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs952243697 Chr.9: 127943076-127943076 on Build GRCh38
rs374998532 Chr.9: 127943021-127943021 on Build GRCh38
rs775065514 Chr.9: 127943019-127943019 on Build GRCh38
rs753669130 Chr.9: 127943159-127943159 on Build GRCh38
rs957881383 Chr.9: 127942884-127942884 on Build GRCh38
rs140195314 Chr.9: 127943024-127943024 on Build GRCh38
rs113478202 Chr.9: 127943067-127943067 on Build GRCh38
rs554933923 Chr.9: 127942887-127942887 on Build GRCh38
rs952190078 Chr.9: 127942833-127942833 on Build GRCh38
rs999507518 Chr.9: 127943032-127943032 on Build GRCh38
rs1006328547 Chr.9: 127943080-127943081 on Build GRCh38
rs376528865 Chr.9: 127943179-127943179 on Build GRCh38
rs371451560 Chr.9: 127943138-127943138 on Build GRCh38
rs199943878 Chr.9: 127943150-127943150 on Build GRCh38
rs557530704 Chr.9: 127942939-127942939 on Build GRCh38
rs146858971 Chr.9: 127943256-127943256 on Build GRCh38
rs888194691 Chr.9: 127943264-127943265 on Build GRCh38
rs963966171 Chr.9: 127943139-127943139 on Build GRCh38
rs532935303 Chr.9: 127943188-127943188 on Build GRCh38
rs1031515261 Chr.9: 127943041-127943041 on Build GRCh38
rs988909680 Chr.9: 127943158-127943158 on Build GRCh38
rs377367528 Chr.9: 127943254-127943254 on Build GRCh38
rs764632306 Chr.9: 127943272-127943272 on Build GRCh38
rs773276236 Chr.9: 127943161-127943161 on Build GRCh38
rs927084572 Chr.9: 127943182-127943182 on Build GRCh38
rs191650091 Chr.9: 127943165-127943165 on Build GRCh38
rs769516529 Chr.9: 127942896-127942896 on Build GRCh38
rs964983553 Chr.9: 127943097-127943097 on Build GRCh38
rs976709093 Chr.9: 127943104-127943104 on Build GRCh38
rs528795656 Chr.9: 127943091-127943091 on Build GRCh38
rs942189909 Chr.9: 127942980-127942980 on Build GRCh38
rs1056807220 Chr.9: 127943049-127943049 on Build GRCh38
rs185677323 Chr.9: 127943186-127943186 on Build GRCh38
rs1026662036 Chr.9: 127943072-127943072 on Build GRCh38
rs976485884 Chr.9: 127942924-127942924 on Build GRCh38
rs771984430 Chr.9: 127943229-127943229 on Build GRCh38
rs947188573 Chr.9: 127942938-127942938 on Build GRCh38
rs183227175 Chr.9: 127943178-127943178 on Build GRCh38
rs1052370129 Chr.9: 127943269-127943269 on Build GRCh38
rs922255650 Chr.9: 127942927-127942927 on Build GRCh38
rs373563289 Chr.9: 127943277-127943277 on Build GRCh38
rs1017851431 Chr.9: 127943083-127943083 on Build GRCh38
rs745379088 Chr.9: 127942846-127942846 on Build GRCh38
rs779719327 Chr.9: 127943204-127943204 on Build GRCh38
rs911168710 Chr.9: 127942899-127942899 on Build GRCh38
rs766563592 Chr.9: 127943170-127943170 on Build GRCh38
rs201003697 Chr.9: 127943248-127943248 on Build GRCh38
rs941085356 Chr.9: 127943197-127943197 on Build GRCh38
rs370939573 Chr.9: 127942928-127942928 on Build GRCh38
rs1043060 Chr.9: 127942841-127942841 on Build GRCh38
rs888174123 Chr.9: 127942954-127942954 on Build GRCh38
rs779142107 Chr.9: 127942869-127942869 on Build GRCh38
rs923302090 Chr.9: 127943144-127943144 on Build GRCh38
rs574693308 Chr.9: 127942923-127942923 on Build GRCh38
rs1006379483 Chr.9: 127943273-127943273 on Build GRCh38
rs1037855341 Chr.9: 127943008-127943008 on Build GRCh38
rs1053575317 Chr.9: 127942930-127942938 on Build GRCh38
rs574769217 Chr.9: 127942902-127942902 on Build GRCh38
rs113049857 Chr.9: 127942930-127942930 on Build GRCh38
rs754156192 Chr.9: 127943177-127943177 on Build GRCh38
rs754793970 Chr.9: 127943188-127943190 on Build GRCh38
rs755729316 Chr.9: 127943199-127943199 on Build GRCh38
rs749057845 Chr.9: 127943219-127943219 on Build GRCh38
rs1044356905 Chr.9: 127942992-127942992 on Build GRCh38
rs1012495578 Chr.9: 127943023-127943023 on Build GRCh38
rs373081704 Chr.9: 127942841-127942841 on Build GRCh38
rs758969060 Chr.9: 127943184-127943184 on Build GRCh38
rs370734681 Chr.9: 127943271-127943271 on Build GRCh38
rs201869221 Chr.9: 127943155-127943155 on Build GRCh38
rs1018063304 Chr.9: 127943124-127943124 on Build GRCh38
rs759647556 Chr.9: 127943263-127943263 on Build GRCh38
rs989242271 Chr.9: 127942885-127942885 on Build GRCh38
rs748146104 Chr.9: 127943225-127943225 on Build GRCh38
rs906527690 Chr.9: 127943026-127943026 on Build GRCh38
rs1031044382 Chr.9: 127942878-127942878 on Build GRCh38
rs769049077 Chr.9: 127942885-127942886 on Build GRCh38
rs577472869 Chr.9: 127943005-127943005 on Build GRCh38
rs942605506 Chr.9: 127942915-127942915 on Build GRCh38
rs145965684 Chr.9: 127943224-127943224 on Build GRCh38
rs773177408 Chr.9: 127943244-127943244 on Build GRCh38
rs114873837 Chr.9: 127942837-127942837 on Build GRCh38
rs143780472 Chr.9: 127943059-127943059 on Build GRCh38
rs553926062 Chr.9: 127943255-127943255 on Build GRCh38
rs774215483 Chr.9: 127943212-127943212 on Build GRCh38
rs772174892 Chr.9: 127943160-127943160 on Build GRCh38
rs765764338 Chr.9: 127943265-127943265 on Build GRCh38
rs533837423 Chr.9: 127943215-127943215 on Build GRCh38

View Full Product Details