Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 127950332-127950826 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM102A
Entrez Gene ID
399665
Gene Name
family with sequence similarity 102 member A
Gene Aliases
C9orf132, EEIG1, SYM-3A, bA203J24.7
UniGene
Hs.535972
Gene Chromosome Location
Chr.9: 127940582-127980533 on Build GRCh38
Transcripts
NM_001035254.2 NM_203305.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658024 c.366C>T p.L122L Chr.9: 127950471-127950471 on Build GRCh38
3847717 c.326A>T p.E109V Chr.9: 127950511-127950511 on Build GRCh38
3847718 c.325G>T p.E109* Chr.9: 127950512-127950512 on Build GRCh38
6476217 c.332C>T p.A111V Chr.9: 127950505-127950505 on Build GRCh38
1643749 c.338C>T p.S113L Chr.9: 127950499-127950499 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369396063 Chr.9: 127950558-127950558 on Build GRCh38
rs1045839925 Chr.9: 127950561-127950561 on Build GRCh38
rs775489876 Chr.9: 127950581-127950581 on Build GRCh38
rs945572979 Chr.9: 127950583-127950583 on Build GRCh38
rs953898089 Chr.9: 127950719-127950719 on Build GRCh38
rs187249615 Chr.9: 127950399-127950399 on Build GRCh38
rs376775888 Chr.9: 127950557-127950557 on Build GRCh38
rs961532192 Chr.9: 127950453-127950453 on Build GRCh38
rs928308519 Chr.9: 127950518-127950518 on Build GRCh38
rs768219961 Chr.9: 127950489-127950489 on Build GRCh38
rs967040002 Chr.9: 127950783-127950783 on Build GRCh38
rs1008218675 Chr.9: 127950718-127950718 on Build GRCh38
rs750877121 Chr.9: 127950545-127950545 on Build GRCh38
rs986633751 Chr.9: 127950744-127950744 on Build GRCh38
rs766425248 Chr.9: 127950407-127950407 on Build GRCh38
rs1054841702 Chr.9: 127950799-127950799 on Build GRCh38
rs778558216 Chr.9: 127950548-127950548 on Build GRCh38
rs143563525 Chr.9: 127950534-127950534 on Build GRCh38
rs772055725 Chr.9: 127950495-127950495 on Build GRCh38
rs776680535 Chr.9: 127950568-127950568 on Build GRCh38
rs747768221 Chr.9: 127950494-127950494 on Build GRCh38
rs778107111 Chr.9: 127950471-127950471 on Build GRCh38
rs747870543 Chr.9: 127950554-127950554 on Build GRCh38
rs148109002 Chr.9: 127950618-127950618 on Build GRCh38
rs891145428 Chr.9: 127950676-127950676 on Build GRCh38
rs753895286 Chr.9: 127950411-127950411 on Build GRCh38
rs770797425 Chr.9: 127950565-127950565 on Build GRCh38
rs764457999 Chr.9: 127950528-127950528 on Build GRCh38
rs1016029583 Chr.9: 127950661-127950661 on Build GRCh38
rs752120354 Chr.9: 127950477-127950477 on Build GRCh38
rs199620865 Chr.9: 127950379-127950379 on Build GRCh38
rs573794413 Chr.9: 127950791-127950791 on Build GRCh38
rs369369408 Chr.9: 127950457-127950457 on Build GRCh38
rs752984958 Chr.9: 127950512-127950512 on Build GRCh38
rs978009363 Chr.9: 127950789-127950789 on Build GRCh38
rs748795367 Chr.9: 127950487-127950487 on Build GRCh38
rs763149931 Chr.9: 127950513-127950513 on Build GRCh38
rs142892270 Chr.9: 127950646-127950646 on Build GRCh38
rs200517841 Chr.9: 127950562-127950562 on Build GRCh38
rs778762388 Chr.9: 127950490-127950490 on Build GRCh38
rs997102787 Chr.9: 127950638-127950638 on Build GRCh38
rs759680391 Chr.9: 127950450-127950450 on Build GRCh38
rs148868201 Chr.9: 127950415-127950415 on Build GRCh38
rs767087163 Chr.9: 127950504-127950504 on Build GRCh38
rs533401870 Chr.9: 127950366-127950366 on Build GRCh38
rs995620800 Chr.9: 127950671-127950671 on Build GRCh38
rs1015649358 Chr.9: 127950445-127950445 on Build GRCh38
rs745788228 Chr.9: 127950573-127950573 on Build GRCh38
rs761489487 Chr.9: 127950577-127950577 on Build GRCh38
rs962725033 Chr.9: 127950664-127950664 on Build GRCh38
rs1008333248 Chr.9: 127950416-127950416 on Build GRCh38
rs751887392 Chr.9: 127950531-127950531 on Build GRCh38
rs547384439 Chr.9: 127950567-127950567 on Build GRCh38
rs569137013 Chr.9: 127950485-127950485 on Build GRCh38
rs936326290 Chr.9: 127950792-127950792 on Build GRCh38
rs868258935 Chr.9: 127950505-127950505 on Build GRCh38
rs770832940 Chr.9: 127950502-127950502 on Build GRCh38
rs760540755 Chr.9: 127950499-127950499 on Build GRCh38
rs769918331 Chr.9: 127950578-127950578 on Build GRCh38
rs766387100 Chr.9: 127950793-127950793 on Build GRCh38
rs757726724 Chr.9: 127950484-127950484 on Build GRCh38
rs781691855 Chr.9: 127950544-127950544 on Build GRCh38
rs760654140 Chr.9: 127950405-127950405 on Build GRCh38
rs1043890694 Chr.9: 127950586-127950586 on Build GRCh38
rs756036720 Chr.9: 127950762-127950762 on Build GRCh38
rs137904200 Chr.9: 127950498-127950498 on Build GRCh38
rs202051418 Chr.9: 127950386-127950386 on Build GRCh38
rs773318486 Chr.9: 127950402-127950402 on Build GRCh38
rs553557700 Chr.9: 127950784-127950784 on Build GRCh38
rs191712969 Chr.9: 127950456-127950456 on Build GRCh38
rs754571658 Chr.9: 127950547-127950547 on Build GRCh38
rs1016116966 Chr.9: 127950771-127950771 on Build GRCh38
rs371818022 Chr.9: 127950385-127950385 on Build GRCh38
rs758803670 Chr.9: 127950458-127950458 on Build GRCh38
rs112631852 Chr.9: 127950360-127950360 on Build GRCh38

View Full Product Details