Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128163224-128163719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf16
Entrez Gene ID
79095
Gene Name
chromosome 9 open reading frame 16
Gene Aliases
EST00098
UniGene
Hs.522412
Gene Chromosome Location
Chr.9: 128159425-128163928 on Build GRCh38
Transcripts
NM_024112.3 XM_011519004.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4607223 c.211C>A p.Q71K Chr.9: 128163574-128163574 on Build GRCh38
4538316 c.252G>A p.*84* Chr.9: 128163615-128163615 on Build GRCh38
4163362 c.245G>A p.S82N Chr.9: 128163608-128163608 on Build GRCh38
3326476 c.164G>A p.R55H Chr.9: 128163527-128163527 on Build GRCh38
1460186 c.210C>A p.F70L Chr.9: 128163573-128163573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372584380 Chr.9: 128163421-128163421 on Build GRCh38
rs561823797 Chr.9: 128163696-128163696 on Build GRCh38
rs781483127 Chr.9: 128163595-128163595 on Build GRCh38
rs370967531 Chr.9: 128163402-128163402 on Build GRCh38
rs771888992 Chr.9: 128163400-128163400 on Build GRCh38
rs11547071 Chr.9: 128163485-128163485 on Build GRCh38
rs760145524 Chr.9: 128163526-128163526 on Build GRCh38
rs1010146324 Chr.9: 128163315-128163315 on Build GRCh38
rs752549241 Chr.9: 128163576-128163576 on Build GRCh38
rs757235243 Chr.9: 128163594-128163594 on Build GRCh38
rs541682513 Chr.9: 128163304-128163304 on Build GRCh38
rs912891346 Chr.9: 128163609-128163609 on Build GRCh38
rs764729037 Chr.9: 128163575-128163575 on Build GRCh38
rs765391228 Chr.9: 128163503-128163505 on Build GRCh38
rs766042254 Chr.9: 128163448-128163448 on Build GRCh38
rs540874014 Chr.9: 128163634-128163634 on Build GRCh38
rs759782846 Chr.9: 128163498-128163500 on Build GRCh38
rs376526629 Chr.9: 128163399-128163399 on Build GRCh38
rs750230464 Chr.9: 128163635-128163635 on Build GRCh38
rs760122023 Chr.9: 128163447-128163447 on Build GRCh38
rs532014523 Chr.9: 128163541-128163541 on Build GRCh38
rs771288916 Chr.9: 128163407-128163407 on Build GRCh38
rs373098556 Chr.9: 128163658-128163658 on Build GRCh38
rs148128033 Chr.9: 128163553-128163553 on Build GRCh38
rs756367617 Chr.9: 128163606-128163606 on Build GRCh38
rs758360266 Chr.9: 128163498-128163498 on Build GRCh38
rs770320040 Chr.9: 128163420-128163420 on Build GRCh38
rs752786661 Chr.9: 128163573-128163573 on Build GRCh38
rs1044133770 Chr.9: 128163276-128163276 on Build GRCh38
rs73672436 Chr.9: 128163632-128163632 on Build GRCh38
rs752428154 Chr.9: 128163497-128163497 on Build GRCh38
rs745940671 Chr.9: 128163596-128163596 on Build GRCh38
rs770130189 Chr.9: 128163539-128163539 on Build GRCh38
rs751585836 Chr.9: 128163586-128163586 on Build GRCh38
rs374330051 Chr.9: 128163585-128163585 on Build GRCh38
rs1049825572 Chr.9: 128163695-128163695 on Build GRCh38
rs932826948 Chr.9: 128163685-128163685 on Build GRCh38
rs760008560 Chr.9: 128163417-128163417 on Build GRCh38
rs1052196 Chr.9: 128163487-128163487 on Build GRCh38
rs753449872 Chr.9: 128163451-128163451 on Build GRCh38
rs142709964 Chr.9: 128163314-128163314 on Build GRCh38
rs922853789 Chr.9: 128163472-128163472 on Build GRCh38
rs928275851 Chr.9: 128163277-128163277 on Build GRCh38
rs760211969 Chr.9: 128163562-128163562 on Build GRCh38
rs754595267 Chr.9: 128163465-128163465 on Build GRCh38
rs759249879 Chr.9: 128163572-128163572 on Build GRCh38
rs746145233 Chr.9: 128163527-128163527 on Build GRCh38
rs11547072 Chr.9: 128163554-128163554 on Build GRCh38
rs1053020742 Chr.9: 128163513-128163513 on Build GRCh38
rs375624105 Chr.9: 128163440-128163440 on Build GRCh38
rs758075952 Chr.9: 128163577-128163577 on Build GRCh38
rs1008254960 Chr.9: 128163519-128163519 on Build GRCh38
rs763575935 Chr.9: 128163429-128163429 on Build GRCh38
rs200071704 Chr.9: 128163618-128163618 on Build GRCh38
rs904028799 Chr.9: 128163351-128163351 on Build GRCh38
rs117376966 Chr.9: 128163341-128163341 on Build GRCh38
rs974401009 Chr.9: 128163558-128163558 on Build GRCh38
rs749687008 Chr.9: 128163630-128163630 on Build GRCh38
rs777509724 Chr.9: 128163504-128163504 on Build GRCh38
rs972302520 Chr.9: 128163256-128163256 on Build GRCh38
rs776940283 Chr.9: 128163410-128163410 on Build GRCh38
rs969879259 Chr.9: 128163564-128163564 on Build GRCh38
rs543043015 Chr.9: 128163397-128163397 on Build GRCh38
rs938369480 Chr.9: 128163291-128163291 on Build GRCh38
rs373847523 Chr.9: 128163563-128163563 on Build GRCh38
rs996903370 Chr.9: 128163395-128163395 on Build GRCh38
rs764508683 Chr.9: 128163438-128163438 on Build GRCh38
rs775993586 Chr.9: 128163548-128163548 on Build GRCh38
rs1039894384 Chr.9: 128163404-128163404 on Build GRCh38
rs749958212 Chr.9: 128163441-128163441 on Build GRCh38
rs767299814 Chr.9: 128163657-128163657 on Build GRCh38
rs780084626 Chr.9: 128163610-128163610 on Build GRCh38
rs563256427 Chr.9: 128163523-128163523 on Build GRCh38
rs747104395 Chr.9: 128163522-128163522 on Build GRCh38
rs548026235 Chr.9: 128163646-128163646 on Build GRCh38

View Full Product Details