Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128203497-128203989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
UniGene
Hs.212395
Gene Chromosome Location
Chr.9: 128166065-128204383 on Build GRCh38
Transcripts
NM_001131015.1 NM_012127.2 NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_006716993.1 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014372.1 XM_017014373.1

Gene Symbol
DNM1
Entrez Gene ID
1759
Gene Name
dynamin 1
Gene Aliases
DNM, EIEE31
UniGene
Hs.522413
Gene Chromosome Location
Chr.9: 128203355-128255248 on Build GRCh38
Transcripts
NM_001131015.1 NM_012127.2 NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_006716993.1 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014372.1 XM_017014373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3654557 c.70G>A p.G24S Chr.9: 128203540-128203540 on Build GRCh38
3654558 c.70G>A p.G24S Chr.9: 128203540-128203540 on Build GRCh38
3664119 c.122G>A p.S41N Chr.9: 128203592-128203592 on Build GRCh38
7093427 c.148A>C p.N50H Chr.9: 128203618-128203618 on Build GRCh38
3664118 c.122G>A p.S41N Chr.9: 128203592-128203592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750933399 Chr.9: 128203555-128203555 on Build GRCh38
rs901303026 Chr.9: 128203711-128203711 on Build GRCh38
rs561719219 Chr.9: 128203784-128203784 on Build GRCh38
rs541942261 Chr.9: 128203723-128203723 on Build GRCh38
rs937334479 Chr.9: 128203709-128203709 on Build GRCh38
rs953178364 Chr.9: 128203937-128203937 on Build GRCh38
rs752123322 Chr.9: 128203568-128203568 on Build GRCh38
rs892563960 Chr.9: 128203969-128203969 on Build GRCh38
rs971870821 Chr.9: 128203895-128203895 on Build GRCh38
rs972962445 Chr.9: 128203961-128203961 on Build GRCh38
rs769600214 Chr.9: 128203620-128203620 on Build GRCh38
rs869312702 Chr.9: 128203609-128203609 on Build GRCh38
rs373421556 Chr.9: 128203757-128203757 on Build GRCh38
rs559486724 Chr.9: 128203960-128203960 on Build GRCh38
rs778543086 Chr.9: 128203559-128203559 on Build GRCh38
rs756572497 Chr.9: 128203558-128203558 on Build GRCh38
rs376304839 Chr.9: 128203758-128203758 on Build GRCh38
rs1019884702 Chr.9: 128203947-128203947 on Build GRCh38
rs972860852 Chr.9: 128203578-128203578 on Build GRCh38
rs1036489716 Chr.9: 128203650-128203650 on Build GRCh38
rs771476756 Chr.9: 128203727-128203749 on Build GRCh38
rs1052540193 Chr.9: 128203959-128203959 on Build GRCh38
rs944188764 Chr.9: 128203557-128203557 on Build GRCh38
rs770857641 Chr.9: 128203594-128203594 on Build GRCh38
rs142432944 Chr.9: 128203587-128203587 on Build GRCh38
rs895704653 Chr.9: 128203739-128203739 on Build GRCh38
rs918788217 Chr.9: 128203965-128203965 on Build GRCh38
rs984474001 Chr.9: 128203907-128203907 on Build GRCh38
rs931414082 Chr.9: 128203639-128203639 on Build GRCh38
rs1048463645 Chr.9: 128203647-128203647 on Build GRCh38
rs762189357 Chr.9: 128203552-128203552 on Build GRCh38
rs575427347 Chr.9: 128203789-128203789 on Build GRCh38
rs1028713106 Chr.9: 128203910-128203910 on Build GRCh38
rs781131754 Chr.9: 128203608-128203608 on Build GRCh38
rs758104991 Chr.9: 128203575-128203575 on Build GRCh38
rs998356210 Chr.9: 128203727-128203727 on Build GRCh38
rs919005830 Chr.9: 128203654-128203654 on Build GRCh38
rs775680544 Chr.9: 128203645-128203645 on Build GRCh38
rs61757224 Chr.9: 128203516-128203516 on Build GRCh38
rs540152796 Chr.9: 128203673-128203673 on Build GRCh38
rs111647560 Chr.9: 128203851-128203851 on Build GRCh38
rs577437504 Chr.9: 128203554-128203554 on Build GRCh38
rs774571600 Chr.9: 128203680-128203680 on Build GRCh38
rs1034869470 Chr.9: 128203856-128203856 on Build GRCh38
rs934178360 Chr.9: 128203955-128203955 on Build GRCh38
rs115261347 Chr.9: 128203939-128203939 on Build GRCh38
rs1010890313 Chr.9: 128203759-128203759 on Build GRCh38
rs930348357 Chr.9: 128203903-128203903 on Build GRCh38
rs751741699 Chr.9: 128203518-128203518 on Build GRCh38
rs1010102169 Chr.9: 128203847-128203847 on Build GRCh38
rs952882811 Chr.9: 128203968-128203968 on Build GRCh38
rs745880488 Chr.9: 128203614-128203614 on Build GRCh38
rs533062466 Chr.9: 128203896-128203896 on Build GRCh38
rs901423796 Chr.9: 128203952-128203952 on Build GRCh38
rs1000380960 Chr.9: 128203908-128203908 on Build GRCh38
rs564336358 Chr.9: 128203866-128203866 on Build GRCh38
rs930317245 Chr.9: 128203660-128203660 on Build GRCh38
rs1056979334 Chr.9: 128203738-128203738 on Build GRCh38
rs112665081 Chr.9: 128203795-128203795 on Build GRCh38
rs762975001 Chr.9: 128203667-128203667 on Build GRCh38
rs981059050 Chr.9: 128203837-128203837 on Build GRCh38
rs960860958 Chr.9: 128203877-128203877 on Build GRCh38
rs768734624 Chr.9: 128203674-128203674 on Build GRCh38
rs572915709 Chr.9: 128203705-128203705 on Build GRCh38
rs552678023 Chr.9: 128203692-128203692 on Build GRCh38

View Full Product Details