Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128204106-128204616 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIZ1
Entrez Gene ID
25792
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
LSFR1, NP94, ZNF356
UniGene
Hs.212395
Gene Chromosome Location
Chr.9: 128166065-128204383 on Build GRCh38
Transcripts
NM_001131015.1 NM_012127.2 NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_006716993.1 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014372.1 XM_017014373.1

Gene Symbol
DNM1
Entrez Gene ID
1759
Gene Name
dynamin 1
Gene Aliases
DNM, EIEE31
UniGene
Hs.522413
Gene Chromosome Location
Chr.9: 128203355-128255248 on Build GRCh38
Transcripts
NM_001131015.1 NM_012127.2 NM_001005336.2 NM_001288737.1 NM_001288738.1 NM_001288739.1 NM_004408.3 XM_005251763.2 XM_005251768.2 XM_005251769.2 XM_006716992.2 XM_006716993.1 XM_011518335.2 XM_011518336.2 XM_017014369.1 XM_017014370.1 XM_017014371.1 XM_017014372.1 XM_017014373.1

SNP Detail

SNP ID Chromosome Location
rs888943863 Chr.9: 128204304-128204304 on Build GRCh38
rs1017796468 Chr.9: 128204360-128204360 on Build GRCh38
rs961694259 Chr.9: 128204195-128204195 on Build GRCh38
rs7851473 Chr.9: 128204553-128204553 on Build GRCh38
rs1006520748 Chr.9: 128204459-128204459 on Build GRCh38
rs935971222 Chr.9: 128204526-128204526 on Build GRCh38
rs896453911 Chr.9: 128204462-128204462 on Build GRCh38
rs563844548 Chr.9: 128204436-128204436 on Build GRCh38
rs889545582 Chr.9: 128204433-128204433 on Build GRCh38
rs984505129 Chr.9: 128204139-128204139 on Build GRCh38
rs1006088437 Chr.9: 128204512-128204512 on Build GRCh38
rs1011863798 Chr.9: 128204184-128204184 on Build GRCh38
rs997238063 Chr.9: 128204572-128204572 on Build GRCh38
rs994434948 Chr.9: 128204208-128204208 on Build GRCh38
rs897514118 Chr.9: 128204339-128204339 on Build GRCh38
rs529769489 Chr.9: 128204435-128204435 on Build GRCh38
rs138622590 Chr.9: 128204521-128204521 on Build GRCh38
rs935840097 Chr.9: 128204262-128204262 on Build GRCh38
rs1040149621 Chr.9: 128204555-128204555 on Build GRCh38
rs747775562 Chr.9: 128204389-128204389 on Build GRCh38
rs539733673 Chr.9: 128204227-128204227 on Build GRCh38
rs992883982 Chr.9: 128204455-128204455 on Build GRCh38
rs1040119011 Chr.9: 128204324-128204324 on Build GRCh38
rs571057337 Chr.9: 128204226-128204226 on Build GRCh38
rs925853102 Chr.9: 128204249-128204249 on Build GRCh38
rs113686866 Chr.9: 128204335-128204335 on Build GRCh38
rs977324455 Chr.9: 128204245-128204245 on Build GRCh38
rs938083610 Chr.9: 128204439-128204439 on Build GRCh38
rs146363472 Chr.9: 128204395-128204395 on Build GRCh38
rs577818144 Chr.9: 128204483-128204483 on Build GRCh38
rs955626106 Chr.9: 128204398-128204398 on Build GRCh38
rs896410197 Chr.9: 128204258-128204258 on Build GRCh38
rs1006055679 Chr.9: 128204351-128204351 on Build GRCh38
rs34473764 Chr.9: 128204326-128204326 on Build GRCh38
rs992852968 Chr.9: 128204230-128204230 on Build GRCh38
rs10760549 Chr.9: 128204156-128204156 on Build GRCh38
rs10987928 Chr.9: 128204539-128204539 on Build GRCh38
rs1002644141 Chr.9: 128204228-128204228 on Build GRCh38
rs529551529 Chr.9: 128204479-128204479 on Build GRCh38
rs560675096 Chr.9: 128204225-128204225 on Build GRCh38
rs914150111 Chr.9: 128204433-128204434 on Build GRCh38
rs553674081 Chr.9: 128204234-128204234 on Build GRCh38
rs771747748 Chr.9: 128204507-128204507 on Build GRCh38
rs548146819 Chr.9: 128204591-128204591 on Build GRCh38
rs748677912 Chr.9: 128204213-128204213 on Build GRCh38
rs535398398 Chr.9: 128204281-128204281 on Build GRCh38
rs577597873 Chr.9: 128204350-128204350 on Build GRCh38
rs910399810 Chr.9: 128204550-128204550 on Build GRCh38
rs1030421590 Chr.9: 128204587-128204587 on Build GRCh38
rs910293444 Chr.9: 128204140-128204140 on Build GRCh38
rs976167107 Chr.9: 128204376-128204376 on Build GRCh38
rs891607644 Chr.9: 128204169-128204169 on Build GRCh38
rs1026326664 Chr.9: 128204367-128204367 on Build GRCh38
rs1018647494 Chr.9: 128204165-128204165 on Build GRCh38
rs944468334 Chr.9: 128204308-128204308 on Build GRCh38
rs977414110 Chr.9: 128204509-128204509 on Build GRCh38
rs368852624 Chr.9: 128204418-128204418 on Build GRCh38
rs993375051 Chr.9: 128204300-128204300 on Build GRCh38
rs897546583 Chr.9: 128204588-128204588 on Build GRCh38
rs45596331 Chr.9: 128204463-128204463 on Build GRCh38
rs1019307410 Chr.9: 128204465-128204465 on Build GRCh38
rs575389805 Chr.9: 128204374-128204374 on Build GRCh38
rs573581011 Chr.9: 128204256-128204256 on Build GRCh38
rs768148864 Chr.9: 128204243-128204243 on Build GRCh38
rs946020292 Chr.9: 128204238-128204238 on Build GRCh38
rs958822753 Chr.9: 128204229-128204229 on Build GRCh38
rs1050310620 Chr.9: 128204301-128204301 on Build GRCh38

View Full Product Details