Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128259089-128259595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GOLGA2
Entrez Gene ID
2801
Gene Name
golgin A2
Gene Aliases
GM130
UniGene
Hs.155827
Gene Chromosome Location
Chr.9: 128255829-128276007 on Build GRCh38
Transcripts
NM_004486.4 XM_005251930.1 XM_005251931.1 XM_005251932.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3326645 c.1871C>T p.A624V Chr.9: 128259312-128259312 on Build GRCh38
6380549 c.2008G>T p.E670* Chr.9: 128259175-128259175 on Build GRCh38
3996326 c.2016G>A p.Q672Q Chr.9: 128259167-128259167 on Build GRCh38
3326643 c.1873G>A p.A625T Chr.9: 128259310-128259310 on Build GRCh38
1745189 c.1818_1819delAA p.L608fs*24 Chr.9: 128259364-128259365 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753903300 Chr.9: 128259199-128259199 on Build GRCh38
rs958324954 Chr.9: 128259494-128259494 on Build GRCh38
rs755044065 Chr.9: 128259294-128259294 on Build GRCh38
rs771357451 Chr.9: 128259323-128259323 on Build GRCh38
rs774887378 Chr.9: 128259239-128259239 on Build GRCh38
rs775248808 Chr.9: 128259153-128259153 on Build GRCh38
rs547428164 Chr.9: 128259524-128259524 on Build GRCh38
rs1020482105 Chr.9: 128259454-128259454 on Build GRCh38
rs1043303945 Chr.9: 128259352-128259352 on Build GRCh38
rs143703939 Chr.9: 128259191-128259191 on Build GRCh38
rs747352647 Chr.9: 128259311-128259311 on Build GRCh38
rs372053134 Chr.9: 128259118-128259118 on Build GRCh38
rs376648484 Chr.9: 128259373-128259373 on Build GRCh38
rs760422757 Chr.9: 128259244-128259244 on Build GRCh38
rs768880004 Chr.9: 128259123-128259123 on Build GRCh38
rs149599897 Chr.9: 128259334-128259334 on Build GRCh38
rs878866954 Chr.9: 128259340-128259340 on Build GRCh38
rs766783666 Chr.9: 128259561-128259561 on Build GRCh38
rs774184231 Chr.9: 128259342-128259342 on Build GRCh38
rs144285995 Chr.9: 128259362-128259362 on Build GRCh38
rs371567512 Chr.9: 128259372-128259372 on Build GRCh38
rs747829710 Chr.9: 128259351-128259351 on Build GRCh38
rs776616919 Chr.9: 128259395-128259395 on Build GRCh38
rs759143477 Chr.9: 128259144-128259144 on Build GRCh38
rs760161583 Chr.9: 128259131-128259131 on Build GRCh38
rs907973863 Chr.9: 128259560-128259560 on Build GRCh38
rs748452527 Chr.9: 128259134-128259134 on Build GRCh38
rs779164185 Chr.9: 128259306-128259306 on Build GRCh38
rs111474923 Chr.9: 128259316-128259316 on Build GRCh38
rs552068111 Chr.9: 128259309-128259309 on Build GRCh38
rs752733813 Chr.9: 128259404-128259404 on Build GRCh38
rs999923116 Chr.9: 128259464-128259464 on Build GRCh38
rs780369866 Chr.9: 128259115-128259115 on Build GRCh38
rs369933643 Chr.9: 128259142-128259142 on Build GRCh38
rs1030496309 Chr.9: 128259416-128259416 on Build GRCh38
rs767386561 Chr.9: 128259255-128259255 on Build GRCh38
rs373294606 Chr.9: 128259152-128259152 on Build GRCh38
rs147019511 Chr.9: 128259189-128259189 on Build GRCh38
rs41276658 Chr.9: 128259154-128259154 on Build GRCh38
rs766292407 Chr.9: 128259391-128259391 on Build GRCh38
rs139831210 Chr.9: 128259245-128259245 on Build GRCh38
rs753866907 Chr.9: 128259278-128259278 on Build GRCh38
rs748878589 Chr.9: 128259330-128259330 on Build GRCh38
rs760549190 Chr.9: 128259271-128259271 on Build GRCh38
rs532305915 Chr.9: 128259273-128259273 on Build GRCh38
rs949538957 Chr.9: 128259262-128259262 on Build GRCh38
rs750189320 Chr.9: 128259266-128259266 on Build GRCh38
rs772411440 Chr.9: 128259206-128259206 on Build GRCh38
rs368942802 Chr.9: 128259367-128259367 on Build GRCh38
rs758803761 Chr.9: 128259310-128259310 on Build GRCh38
rs761761265 Chr.9: 128259253-128259253 on Build GRCh38
rs761312204 Chr.9: 128259539-128259539 on Build GRCh38
rs780026282 Chr.9: 128259195-128259195 on Build GRCh38
rs765270591 Chr.9: 128259401-128259401 on Build GRCh38
rs1008724631 Chr.9: 128259432-128259432 on Build GRCh38
rs540983831 Chr.9: 128259138-128259138 on Build GRCh38
rs1032691529 Chr.9: 128259482-128259482 on Build GRCh38
rs763793851 Chr.9: 128259160-128259160 on Build GRCh38
rs147458263 Chr.9: 128259238-128259238 on Build GRCh38
rs563364974 Chr.9: 128259272-128259272 on Build GRCh38
rs747443265 Chr.9: 128259212-128259212 on Build GRCh38
rs1015353496 Chr.9: 128259540-128259540 on Build GRCh38
rs368066409 Chr.9: 128259411-128259411 on Build GRCh38
rs369066256 Chr.9: 128259200-128259200 on Build GRCh38
rs778302995 Chr.9: 128259207-128259207 on Build GRCh38
rs751415915 Chr.9: 128259170-128259170 on Build GRCh38
rs759660404 Chr.9: 128259396-128259396 on Build GRCh38
rs138214998 Chr.9: 128259190-128259190 on Build GRCh38
rs748579135 Chr.9: 128259202-128259202 on Build GRCh38
rs376341495 Chr.9: 128259132-128259132 on Build GRCh38
rs1012506823 Chr.9: 128259500-128259500 on Build GRCh38
rs954996828 Chr.9: 128259512-128259512 on Build GRCh38
rs781676920 Chr.9: 128259329-128259329 on Build GRCh38
rs750976647 Chr.9: 128259463-128259463 on Build GRCh38

View Full Product Details