Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128434022-128434511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CERCAM
Entrez Gene ID
51148
Gene Name
cerebral endothelial cell adhesion molecule
Gene Aliases
CEECAM1, GLT25D3
UniGene
Hs.495230
Gene Chromosome Location
Chr.9: 128405993-128437351 on Build GRCh38
Transcripts
NM_001286760.1 NM_016174.4 XM_005252035.1 XM_011518762.1 XM_011518763.2 XM_017014794.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200848 c.1127A>G p.E376G Chr.9: 128434439-128434439 on Build GRCh38
4671773 c.1042C>T p.R348W Chr.9: 128434174-128434174 on Build GRCh38
6627296 c.1157G>A p.G386D Chr.9: 128434469-128434469 on Build GRCh38
3904440 c.1018C>T p.R340C Chr.9: 128434150-128434150 on Build GRCh38
1248115 c.1013A>G p.D338G Chr.9: 128434145-128434145 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1002163981 Chr.9: 128434293-128434293 on Build GRCh38
rs774446216 Chr.9: 128434431-128434431 on Build GRCh38
rs770587623 Chr.9: 128434417-128434417 on Build GRCh38
rs367921792 Chr.9: 128434448-128434448 on Build GRCh38
rs747314330 Chr.9: 128434102-128434102 on Build GRCh38
rs371084815 Chr.9: 128434053-128434053 on Build GRCh38
rs372180473 Chr.9: 128434183-128434183 on Build GRCh38
rs757564903 Chr.9: 128434175-128434175 on Build GRCh38
rs750444153 Chr.9: 128434439-128434441 on Build GRCh38
rs144344654 Chr.9: 128434452-128434452 on Build GRCh38
rs776859847 Chr.9: 128434335-128434335 on Build GRCh38
rs756658692 Chr.9: 128434275-128434275 on Build GRCh38
rs187973790 Chr.9: 128434343-128434343 on Build GRCh38
rs756919056 Chr.9: 128434466-128434466 on Build GRCh38
rs551849112 Chr.9: 128434112-128434112 on Build GRCh38
rs776156593 Chr.9: 128434123-128434123 on Build GRCh38
rs141407199 Chr.9: 128434151-128434151 on Build GRCh38
rs1004209976 Chr.9: 128434309-128434309 on Build GRCh38
rs375319496 Chr.9: 128434391-128434391 on Build GRCh38
rs370255320 Chr.9: 128434276-128434276 on Build GRCh38
rs756615888 Chr.9: 128434481-128434487 on Build GRCh38
rs995610983 Chr.9: 128434347-128434347 on Build GRCh38
rs2072398 Chr.9: 128434055-128434055 on Build GRCh38
rs766217072 Chr.9: 128434067-128434067 on Build GRCh38
rs771784043 Chr.9: 128434222-128434222 on Build GRCh38
rs71497697 Chr.9: 128434201-128434201 on Build GRCh38
rs147009468 Chr.9: 128434124-128434124 on Build GRCh38
rs766133803 Chr.9: 128434147-128434147 on Build GRCh38
rs767657041 Chr.9: 128434226-128434228 on Build GRCh38
rs762924138 Chr.9: 128434428-128434428 on Build GRCh38
rs776448337 Chr.9: 128434249-128434249 on Build GRCh38
rs576374091 Chr.9: 128434314-128434314 on Build GRCh38
rs769918619 Chr.9: 128434421-128434421 on Build GRCh38
rs1051009217 Chr.9: 128434267-128434267 on Build GRCh38
rs939010036 Chr.9: 128434161-128434161 on Build GRCh38
rs144224831 Chr.9: 128434251-128434251 on Build GRCh38
rs559248980 Chr.9: 128434215-128434215 on Build GRCh38
rs763904400 Chr.9: 128434299-128434299 on Build GRCh38
rs757840914 Chr.9: 128434105-128434105 on Build GRCh38
rs768167210 Chr.9: 128434113-128434113 on Build GRCh38
rs762044222 Chr.9: 128434268-128434268 on Build GRCh38
rs367716860 Chr.9: 128434079-128434079 on Build GRCh38
rs760334710 Chr.9: 128434068-128434068 on Build GRCh38
rs764208324 Chr.9: 128434171-128434171 on Build GRCh38
rs564900292 Chr.9: 128434297-128434297 on Build GRCh38
rs777290474 Chr.9: 128434410-128434410 on Build GRCh38
rs747630906 Chr.9: 128434213-128434213 on Build GRCh38
rs375926053 Chr.9: 128434255-128434255 on Build GRCh38
rs752793645 Chr.9: 128434163-128434163 on Build GRCh38
rs112392544 Chr.9: 128434296-128434296 on Build GRCh38
rs758655136 Chr.9: 128434081-128434081 on Build GRCh38
rs374673314 Chr.9: 128434174-128434174 on Build GRCh38
rs746618410 Chr.9: 128434235-128434235 on Build GRCh38
rs1021037494 Chr.9: 128434104-128434104 on Build GRCh38
rs760387211 Chr.9: 128434146-128434146 on Build GRCh38
rs570001214 Chr.9: 128434078-128434078 on Build GRCh38
rs2072397 Chr.9: 128434057-128434057 on Build GRCh38
rs773799994 Chr.9: 128434115-128434115 on Build GRCh38
rs754280026 Chr.9: 128434279-128434279 on Build GRCh38
rs114084202 Chr.9: 128434184-128434184 on Build GRCh38
rs7258 Chr.9: 128434416-128434416 on Build GRCh38
rs769832976 Chr.9: 128434252-128434252 on Build GRCh38
rs201652666 Chr.9: 128434472-128434472 on Build GRCh38
rs529782145 Chr.9: 128434420-128434420 on Build GRCh38
rs974205300 Chr.9: 128434090-128434090 on Build GRCh38
rs7259 Chr.9: 128434425-128434425 on Build GRCh38
rs545248656 Chr.9: 128434332-128434332 on Build GRCh38
rs754367815 Chr.9: 128434453-128434453 on Build GRCh38
rs376824941 Chr.9: 128434225-128434225 on Build GRCh38
rs532440285 Chr.9: 128434096-128434096 on Build GRCh38
rs1011032767 Chr.9: 128434085-128434085 on Build GRCh38
rs941771419 Chr.9: 128434218-128434218 on Build GRCh38
rs778431750 Chr.9: 128434189-128434189 on Build GRCh38
rs182483042 Chr.9: 128434400-128434400 on Build GRCh38
rs112916291 Chr.9: 128434467-128434467 on Build GRCh38
rs367870752 Chr.9: 128434050-128434050 on Build GRCh38
rs148628855 Chr.9: 128434150-128434150 on Build GRCh38
rs375543369 Chr.9: 128434066-128434066 on Build GRCh38
rs373555879 Chr.9: 128434261-128434261 on Build GRCh38
rs376962573 Chr.9: 128434262-128434262 on Build GRCh38
rs534312335 Chr.9: 128434160-128434160 on Build GRCh38
rs553943859 Chr.9: 128434168-128434168 on Build GRCh38
rs376071086 Chr.9: 128434270-128434270 on Build GRCh38
rs368478455 Chr.9: 128434242-128434242 on Build GRCh38
rs530636470 Chr.9: 128434337-128434337 on Build GRCh38
rs752991899 Chr.9: 128434080-128434080 on Build GRCh38
rs761035980 Chr.9: 128434449-128434449 on Build GRCh38
rs781545820 Chr.9: 128434109-128434109 on Build GRCh38
rs777189294 Chr.9: 128434457-128434457 on Build GRCh38
rs761911544 Chr.9: 128434055-128434056 on Build GRCh38
rs750751766 Chr.9: 128434271-128434271 on Build GRCh38
rs369784853 Chr.9: 128434429-128434429 on Build GRCh38
rs386738721 Chr.9: 128434055-128434057 on Build GRCh38
rs140126170 Chr.9: 128434479-128434479 on Build GRCh38
rs751060949 Chr.9: 128434464-128434464 on Build GRCh38
rs772506698 Chr.9: 128434049-128434049 on Build GRCh38

View Full Product Details