Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128508703-128509203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLE1
Entrez Gene ID
2733
Gene Name
GLE1, RNA export mediator
Gene Aliases
GLE1L, LCCS, LCCS1, hGLE1
UniGene
Hs.522418
Gene Chromosome Location
Chr.9: 128504692-128542301 on Build GRCh38
Transcripts
NM_001003722.1 NM_001499.2 XM_006717059.2 XM_006717060.2 XM_011518549.1 XM_011518550.1 XM_011518551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4830715 c.230C>G p.S77* Chr.9: 128509006-128509006 on Build GRCh38
3847750 c.137C>T p.S46F Chr.9: 128508913-128508913 on Build GRCh38
1460269 c.164A>G p.H55R Chr.9: 128508940-128508940 on Build GRCh38
4761403 c.236T>C p.L79P Chr.9: 128509012-128509012 on Build GRCh38
166066 c.242delA p.Q81fs*61 Chr.9: 128509018-128509018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766058424 Chr.9: 128509002-128509002 on Build GRCh38
rs371811161 Chr.9: 128508775-128508775 on Build GRCh38
rs758174530 Chr.9: 128509126-128509126 on Build GRCh38
rs764539366 Chr.9: 128509056-128509056 on Build GRCh38
rs973529054 Chr.9: 128508810-128508810 on Build GRCh38
rs201042972 Chr.9: 128509006-128509006 on Build GRCh38
rs886043020 Chr.9: 128508954-128508954 on Build GRCh38
rs1041997835 Chr.9: 128509135-128509135 on Build GRCh38
rs773171034 Chr.9: 128508952-128508952 on Build GRCh38
rs543589909 Chr.9: 128509086-128509086 on Build GRCh38
rs760226671 Chr.9: 128509085-128509085 on Build GRCh38
rs749111923 Chr.9: 128508940-128508940 on Build GRCh38
rs375373350 Chr.9: 128508971-128508971 on Build GRCh38
rs867535211 Chr.9: 128508822-128508822 on Build GRCh38
rs950188082 Chr.9: 128508777-128508777 on Build GRCh38
rs912934898 Chr.9: 128508978-128508978 on Build GRCh38
rs969620233 Chr.9: 128508842-128508842 on Build GRCh38
rs927485122 Chr.9: 128508793-128508793 on Build GRCh38
rs1016877456 Chr.9: 128508956-128508956 on Build GRCh38
rs548471917 Chr.9: 128509097-128509097 on Build GRCh38
rs369339566 Chr.9: 128508977-128508977 on Build GRCh38
rs772765696 Chr.9: 128509017-128509017 on Build GRCh38
rs779590845 Chr.9: 128508730-128508730 on Build GRCh38
rs141903692 Chr.9: 128509036-128509036 on Build GRCh38
rs773542362 Chr.9: 128508843-128508843 on Build GRCh38
rs749876906 Chr.9: 128508903-128508903 on Build GRCh38
rs772263557 Chr.9: 128508950-128508950 on Build GRCh38
rs777044751 Chr.9: 128509046-128509046 on Build GRCh38
rs897878197 Chr.9: 128509074-128509074 on Build GRCh38
rs757473697 Chr.9: 128509145-128509145 on Build GRCh38
rs761142109 Chr.9: 128509026-128509026 on Build GRCh38
rs759935136 Chr.9: 128508891-128508891 on Build GRCh38
rs754826368 Chr.9: 128509013-128509013 on Build GRCh38
rs780559879 Chr.9: 128508898-128508898 on Build GRCh38
rs990788723 Chr.9: 128509061-128509061 on Build GRCh38
rs138871311 Chr.9: 128508892-128508892 on Build GRCh38
rs774891003 Chr.9: 128509066-128509066 on Build GRCh38
rs904405577 Chr.9: 128509000-128509000 on Build GRCh38
rs771305251 Chr.9: 128508968-128508968 on Build GRCh38
rs774230809 Chr.9: 128508836-128508836 on Build GRCh38
rs1054727057 Chr.9: 128508807-128508807 on Build GRCh38
rs748023893 Chr.9: 128508948-128508948 on Build GRCh38
rs760751321 Chr.9: 128508955-128508955 on Build GRCh38
rs754271392 Chr.9: 128508853-128508853 on Build GRCh38
rs771547725 Chr.9: 128508997-128508997 on Build GRCh38
rs753226987 Chr.9: 128508893-128508893 on Build GRCh38
rs760021172 Chr.9: 128508981-128508981 on Build GRCh38
rs760803447 Chr.9: 128508848-128508848 on Build GRCh38
rs764813834 Chr.9: 128509106-128509106 on Build GRCh38
rs375383415 Chr.9: 128508840-128508840 on Build GRCh38
rs994955066 Chr.9: 128509114-128509114 on Build GRCh38
rs758282188 Chr.9: 128509021-128509021 on Build GRCh38
rs918716099 Chr.9: 128508758-128508758 on Build GRCh38
rs764600069 Chr.9: 128508994-128508994 on Build GRCh38
rs1015391374 Chr.9: 128508792-128508792 on Build GRCh38
rs756846701 Chr.9: 128508896-128508896 on Build GRCh38
rs777613862 Chr.9: 128509129-128509129 on Build GRCh38
rs752458769 Chr.9: 128509110-128509110 on Build GRCh38
rs753465803 Chr.9: 128509009-128509009 on Build GRCh38
rs972795807 Chr.9: 128508735-128508735 on Build GRCh38
rs374779242 Chr.9: 128509052-128509052 on Build GRCh38
rs766557201 Chr.9: 128508852-128508852 on Build GRCh38
rs542899467 Chr.9: 128508747-128508747 on Build GRCh38
rs550698804 Chr.9: 128508926-128508926 on Build GRCh38
rs775726336 Chr.9: 128509049-128509049 on Build GRCh38
rs962137630 Chr.9: 128508802-128508802 on Build GRCh38
rs969574533 Chr.9: 128509132-128509132 on Build GRCh38
rs201830047 Chr.9: 128508942-128508942 on Build GRCh38
rs770504824 Chr.9: 128509133-128509133 on Build GRCh38
rs769230078 Chr.9: 128508890-128508891 on Build GRCh38
rs377411601 Chr.9: 128509047-128509047 on Build GRCh38
rs781421905 Chr.9: 128509147-128509147 on Build GRCh38
rs754630031 Chr.9: 128509101-128509101 on Build GRCh38
rs1035658346 Chr.9: 128509178-128509178 on Build GRCh38
rs768406713 Chr.9: 128508941-128508941 on Build GRCh38
rs533223796 Chr.9: 128508998-128508998 on Build GRCh38
rs755731392 Chr.9: 128508907-128508907 on Build GRCh38
rs368997996 Chr.9: 128508841-128508841 on Build GRCh38
rs867034826 Chr.9: 128509063-128509063 on Build GRCh38
rs763372143 Chr.9: 128508993-128508993 on Build GRCh38
rs893449421 Chr.9: 128508818-128508818 on Build GRCh38
rs765678276 Chr.9: 128508986-128508986 on Build GRCh38

View Full Product Details