Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128522331-128522838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLE1
Entrez Gene ID
2733
Gene Name
GLE1, RNA export mediator
Gene Aliases
GLE1L, LCCS, LCCS1, hGLE1
UniGene
Hs.522418
Gene Chromosome Location
Chr.9: 128504692-128542301 on Build GRCh38
Transcripts
NM_001003722.1 NM_001499.2 XM_006717059.2 XM_006717060.2 XM_011518549.1 XM_011518550.1 XM_011518551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6193984 c.470A>G p.K157R Chr.9: 128522705-128522705 on Build GRCh38
6193983 c.470A>G p.K157R Chr.9: 128522705-128522705 on Build GRCh38
6671447 c.563G>A p.R188Q Chr.9: 128522798-128522798 on Build GRCh38
3664122 c.468G>A p.R156R Chr.9: 128522703-128522703 on Build GRCh38
3748979 c.473T>G p.V158G Chr.9: 128522708-128522708 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001661177 Chr.9: 128522442-128522442 on Build GRCh38
rs142600956 Chr.9: 128522382-128522382 on Build GRCh38
rs752621456 Chr.9: 128522636-128522639 on Build GRCh38
rs777593978 Chr.9: 128522653-128522654 on Build GRCh38
rs904595258 Chr.9: 128522659-128522659 on Build GRCh38
rs750079562 Chr.9: 128522618-128522619 on Build GRCh38
rs1031873575 Chr.9: 128522457-128522457 on Build GRCh38
rs967327227 Chr.9: 128522522-128522522 on Build GRCh38
rs953419055 Chr.9: 128522576-128522576 on Build GRCh38
rs919508931 Chr.9: 128522611-128522611 on Build GRCh38
rs763753969 Chr.9: 128522637-128522638 on Build GRCh38
rs956258832 Chr.9: 128522505-128522505 on Build GRCh38
rs944836828 Chr.9: 128522593-128522593 on Build GRCh38
rs912020444 Chr.9: 128522378-128522378 on Build GRCh38
rs767900688 Chr.9: 128522618-128522621 on Build GRCh38
rs749444057 Chr.9: 128522776-128522777 on Build GRCh38
rs953464975 Chr.9: 128522782-128522782 on Build GRCh38
rs886043001 Chr.9: 128522655-128522658 on Build GRCh38
rs141181146 Chr.9: 128522690-128522690 on Build GRCh38
rs752926934 Chr.9: 128522374-128522374 on Build GRCh38
rs371902708 Chr.9: 128522771-128522771 on Build GRCh38
rs546962600 Chr.9: 128522399-128522399 on Build GRCh38
rs925971843 Chr.9: 128522556-128522556 on Build GRCh38
rs117846436 Chr.9: 128522383-128522383 on Build GRCh38
rs773154466 Chr.9: 128522706-128522706 on Build GRCh38
rs1048039054 Chr.9: 128522648-128522648 on Build GRCh38
rs866859481 Chr.9: 128522689-128522690 on Build GRCh38
rs768122055 Chr.9: 128522798-128522798 on Build GRCh38
rs756407395 Chr.9: 128522638-128522638 on Build GRCh38
rs778110977 Chr.9: 128522676-128522676 on Build GRCh38
rs754974982 Chr.9: 128522651-128522651 on Build GRCh38
rs758577981 Chr.9: 128522375-128522375 on Build GRCh38
rs747796840 Chr.9: 128522702-128522702 on Build GRCh38
rs773846928 Chr.9: 128522807-128522807 on Build GRCh38
rs1020205073 Chr.9: 128522511-128522511 on Build GRCh38
rs368484174 Chr.9: 128522797-128522797 on Build GRCh38
rs753975204 Chr.9: 128522649-128522649 on Build GRCh38
rs978895619 Chr.9: 128522437-128522437 on Build GRCh38
rs780012131 Chr.9: 128522694-128522695 on Build GRCh38
rs761390183 Chr.9: 128522636-128522638 on Build GRCh38
rs566762820 Chr.9: 128522749-128522749 on Build GRCh38
rs759714264 Chr.9: 128522721-128522721 on Build GRCh38
rs960102415 Chr.9: 128522467-128522467 on Build GRCh38
rs386833693 Chr.9: 128522658-128522658 on Build GRCh38
rs758822493 Chr.9: 128522672-128522672 on Build GRCh38
rs1034120152 Chr.9: 128522448-128522448 on Build GRCh38
rs144164277 Chr.9: 128522810-128522810 on Build GRCh38
rs533725212 Chr.9: 128522379-128522379 on Build GRCh38
rs761361807 Chr.9: 128522429-128522429 on Build GRCh38
rs888179656 Chr.9: 128522426-128522426 on Build GRCh38
rs746377265 Chr.9: 128522722-128522722 on Build GRCh38
rs895599693 Chr.9: 128522657-128522657 on Build GRCh38
rs978738769 Chr.9: 128522524-128522524 on Build GRCh38
rs931092100 Chr.9: 128522578-128522578 on Build GRCh38
rs774204660 Chr.9: 128522696-128522696 on Build GRCh38
rs553148909 Chr.9: 128522580-128522580 on Build GRCh38
rs1014094634 Chr.9: 128522470-128522470 on Build GRCh38
rs993789355 Chr.9: 128522395-128522395 on Build GRCh38
rs34787152 Chr.9: 128522428-128522429 on Build GRCh38
rs911984786 Chr.9: 128522592-128522592 on Build GRCh38
rs201267934 Chr.9: 128522808-128522808 on Build GRCh38
rs886042471 Chr.9: 128522656-128522658 on Build GRCh38
rs766170630 Chr.9: 128522638-128522638 on Build GRCh38
rs374581579 Chr.9: 128522791-128522791 on Build GRCh38
rs903089056 Chr.9: 128522694-128522694 on Build GRCh38
rs1000085560 Chr.9: 128522716-128522716 on Build GRCh38
rs537504118 Chr.9: 128522674-128522674 on Build GRCh38
rs771873782 Chr.9: 128522806-128522806 on Build GRCh38
rs60154464 Chr.9: 128522658-128522658 on Build GRCh38
rs139301061 Chr.9: 128522730-128522730 on Build GRCh38
rs773349220 Chr.9: 128522636-128522636 on Build GRCh38
rs559370225 Chr.9: 128522799-128522799 on Build GRCh38
rs548972583 Chr.9: 128522587-128522587 on Build GRCh38
rs148220489 Chr.9: 128522682-128522682 on Build GRCh38
rs1023015901 Chr.9: 128522405-128522405 on Build GRCh38
rs778891636 Chr.9: 128522635-128522636 on Build GRCh38
rs986157254 Chr.9: 128522787-128522787 on Build GRCh38
rs766373222 Chr.9: 128522712-128522712 on Build GRCh38
rs1000306035 Chr.9: 128522661-128522661 on Build GRCh38
rs760331142 Chr.9: 128522634-128522663 on Build GRCh38
rs143856051 Chr.9: 128522679-128522679 on Build GRCh38
rs765290778 Chr.9: 128522728-128522728 on Build GRCh38
rs1048235268 Chr.9: 128522354-128522354 on Build GRCh38
rs376866571 Chr.9: 128522406-128522406 on Build GRCh38
rs765485576 Chr.9: 128522652-128522652 on Build GRCh38
rs920991775 Chr.9: 128522574-128522574 on Build GRCh38
rs760338426 Chr.9: 128522708-128522708 on Build GRCh38
rs150781935 Chr.9: 128522720-128522720 on Build GRCh38
rs1019578110 Chr.9: 128522777-128522777 on Build GRCh38
rs535460595 Chr.9: 128522423-128522423 on Build GRCh38
rs761746960 Chr.9: 128522625-128522625 on Build GRCh38
rs757866530 Chr.9: 128522410-128522410 on Build GRCh38
rs571350843 Chr.9: 128522687-128522687 on Build GRCh38
rs750691188 Chr.9: 128522612-128522612 on Build GRCh38
rs765826569 Chr.9: 128522636-128522637 on Build GRCh38
rs939943613 Chr.9: 128522614-128522614 on Build GRCh38
rs751981354 Chr.9: 128522766-128522766 on Build GRCh38
rs781773792 Chr.9: 128522544-128522544 on Build GRCh38
rs758627953 Chr.9: 128522746-128522746 on Build GRCh38
rs534129844 Chr.9: 128522793-128522793 on Build GRCh38
rs767215607 Chr.9: 128522432-128522432 on Build GRCh38
rs1055916088 Chr.9: 128522655-128522655 on Build GRCh38
rs752897261 Chr.9: 128522653-128522653 on Build GRCh38
rs776690956 Chr.9: 128522811-128522811 on Build GRCh38
rs767405041 Chr.9: 128522634-128522634 on Build GRCh38
rs756312366 Chr.9: 128522658-128522659 on Build GRCh38
rs937024001 Chr.9: 128522654-128522654 on Build GRCh38
rs146578060 Chr.9: 128522603-128522603 on Build GRCh38
rs760664675 Chr.9: 128522637-128522637 on Build GRCh38
rs746108894 Chr.9: 128522659-128522659 on Build GRCh38
rs74481329 Chr.9: 128522693-128522693 on Build GRCh38

View Full Product Details