Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128822740-128823237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPOUT1
Entrez Gene ID
51490
Gene Name
SPOUT domain containing methyltransferase 1
Gene Aliases
C9orf114, CENP-32, HSPC109
UniGene
Hs.224137
Gene Chromosome Location
Chr.9: 128819651-128833309 on Build GRCh38
Transcripts
NM_016390.3 XM_017014804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4163394 c.1106T>C p.I369T Chr.9: 128822790-128822790 on Build GRCh38
3327748 c.1087G>A p.A363T Chr.9: 128822809-128822809 on Build GRCh38
5017836 c.1084delG p.A362fs*>15 Chr.9: 128822812-128822812 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756577151 Chr.9: 128823126-128823126 on Build GRCh38
rs932412482 Chr.9: 128823197-128823197 on Build GRCh38
rs914831925 Chr.9: 128823184-128823184 on Build GRCh38
rs374594356 Chr.9: 128823121-128823131 on Build GRCh38
rs1003795098 Chr.9: 128823165-128823165 on Build GRCh38
rs533166916 Chr.9: 128822845-128822845 on Build GRCh38
rs868777763 Chr.9: 128822767-128822767 on Build GRCh38
rs1026092957 Chr.9: 128822850-128822850 on Build GRCh38
rs138635517 Chr.9: 128823119-128823129 on Build GRCh38
rs768391359 Chr.9: 128822773-128822773 on Build GRCh38
rs763248167 Chr.9: 128822877-128822877 on Build GRCh38
rs142430618 Chr.9: 128822919-128822919 on Build GRCh38
rs890216924 Chr.9: 128823044-128823044 on Build GRCh38
rs190887939 Chr.9: 128823201-128823201 on Build GRCh38
rs890791128 Chr.9: 128823205-128823205 on Build GRCh38
rs570960661 Chr.9: 128822809-128822809 on Build GRCh38
rs781376531 Chr.9: 128822846-128822846 on Build GRCh38
rs770733505 Chr.9: 128822785-128822785 on Build GRCh38
rs771583877 Chr.9: 128822882-128822882 on Build GRCh38
rs974340116 Chr.9: 128823069-128823069 on Build GRCh38
rs752129005 Chr.9: 128822824-128822824 on Build GRCh38
rs893296469 Chr.9: 128822887-128822887 on Build GRCh38
rs982984841 Chr.9: 128822778-128822778 on Build GRCh38
rs4425853 Chr.9: 128823196-128823196 on Build GRCh38
rs35677895 Chr.9: 128822807-128822807 on Build GRCh38
rs974184433 Chr.9: 128822810-128822810 on Build GRCh38
rs2977994 Chr.9: 128822937-128822937 on Build GRCh38
rs1007157532 Chr.9: 128823057-128823057 on Build GRCh38
rs759647522 Chr.9: 128822796-128822796 on Build GRCh38
rs368048038 Chr.9: 128822872-128822872 on Build GRCh38
rs557551564 Chr.9: 128823058-128823058 on Build GRCh38
rs150443788 Chr.9: 128822776-128822776 on Build GRCh38
rs1004126732 Chr.9: 128822998-128822998 on Build GRCh38
rs555507893 Chr.9: 128822938-128822938 on Build GRCh38
rs959736999 Chr.9: 128823173-128823173 on Build GRCh38
rs2280843 Chr.9: 128822790-128822790 on Build GRCh38
rs962559085 Chr.9: 128823034-128823034 on Build GRCh38
rs989074039 Chr.9: 128823134-128823134 on Build GRCh38
rs765545425 Chr.9: 128822797-128822797 on Build GRCh38
rs527330652 Chr.9: 128822849-128822849 on Build GRCh38
rs928847434 Chr.9: 128822867-128822867 on Build GRCh38
rs1012198065 Chr.9: 128822888-128822888 on Build GRCh38
rs928626683 Chr.9: 128823127-128823127 on Build GRCh38
rs569132609 Chr.9: 128822955-128822955 on Build GRCh38
rs374931053 Chr.9: 128822775-128822775 on Build GRCh38
rs750951664 Chr.9: 128823037-128823037 on Build GRCh38
rs764207968 Chr.9: 128822899-128822899 on Build GRCh38
rs947550896 Chr.9: 128823186-128823186 on Build GRCh38
rs2997919 Chr.9: 128822926-128822926 on Build GRCh38
rs542957645 Chr.9: 128823204-128823204 on Build GRCh38
rs746978372 Chr.9: 128822884-128822884 on Build GRCh38
rs773137464 Chr.9: 128822784-128822784 on Build GRCh38
rs4595231 Chr.9: 128823216-128823216 on Build GRCh38
rs771988854 Chr.9: 128822783-128822783 on Build GRCh38
rs752706355 Chr.9: 128822803-128822803 on Build GRCh38
rs553729318 Chr.9: 128823198-128823198 on Build GRCh38
rs376972609 Chr.9: 128823133-128823133 on Build GRCh38
rs748918864 Chr.9: 128822768-128822768 on Build GRCh38
rs867821327 Chr.9: 128822823-128822823 on Build GRCh38
rs557467725 Chr.9: 128823158-128823158 on Build GRCh38
rs1004311903 Chr.9: 128823212-128823212 on Build GRCh38
rs1051490981 Chr.9: 128823009-128823009 on Build GRCh38
rs934149707 Chr.9: 128823207-128823207 on Build GRCh38
rs970147118 Chr.9: 128823093-128823093 on Build GRCh38
rs754569361 Chr.9: 128822860-128822860 on Build GRCh38
rs1023025467 Chr.9: 128823070-128823070 on Build GRCh38
rs747589955 Chr.9: 128822878-128822878 on Build GRCh38
rs538459995 Chr.9: 128822984-128822984 on Build GRCh38
rs1035081665 Chr.9: 128823168-128823168 on Build GRCh38
rs752030128 Chr.9: 128822916-128822916 on Build GRCh38
rs763084971 Chr.9: 128823195-128823195 on Build GRCh38
rs1015967967 Chr.9: 128823007-128823007 on Build GRCh38
rs994707082 Chr.9: 128823096-128823096 on Build GRCh38
rs777635976 Chr.9: 128822883-128822883 on Build GRCh38
rs946931539 Chr.9: 128822927-128822927 on Build GRCh38
rs577708312 Chr.9: 128823123-128823123 on Build GRCh38
rs934690998 Chr.9: 128823130-128823130 on Build GRCh38

View Full Product Details