Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 128945965-128946461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOLK
Entrez Gene ID
22845
Gene Name
dolichol kinase
Gene Aliases
CDG1M, DK, DK1, SEC59, TMEM15
UniGene
Hs.531563
Gene Chromosome Location
Chr.9: 128945530-128947733 on Build GRCh38
Transcripts
NM_014908.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5618031 c.1079A>G p.Y360C Chr.9: 128946225-128946225 on Build GRCh38
7365309 c.976G>A p.E326K Chr.9: 128946328-128946328 on Build GRCh38
383612 c.916C>A p.L306M Chr.9: 128946388-128946388 on Build GRCh38
1460360 c.871C>A p.L291I Chr.9: 128946433-128946433 on Build GRCh38
752595 c.1326C>T p.A442A Chr.9: 128945978-128945978 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747285047 Chr.9: 128946145-128946145 on Build GRCh38
rs140983401 Chr.9: 128946016-128946016 on Build GRCh38
rs866644738 Chr.9: 128946125-128946125 on Build GRCh38
rs748455807 Chr.9: 128946386-128946386 on Build GRCh38
rs777991465 Chr.9: 128946140-128946140 on Build GRCh38
rs759025031 Chr.9: 128946294-128946294 on Build GRCh38
rs757321804 Chr.9: 128946146-128946146 on Build GRCh38
rs201984428 Chr.9: 128946138-128946138 on Build GRCh38
rs778808864 Chr.9: 128946034-128946034 on Build GRCh38
rs768213501 Chr.9: 128946417-128946418 on Build GRCh38
rs770126479 Chr.9: 128946080-128946080 on Build GRCh38
rs142287416 Chr.9: 128946065-128946065 on Build GRCh38
rs775228530 Chr.9: 128946077-128946077 on Build GRCh38
rs927774250 Chr.9: 128946385-128946385 on Build GRCh38
rs759148554 Chr.9: 128946197-128946197 on Build GRCh38
rs769241947 Chr.9: 128946298-128946298 on Build GRCh38
rs745571625 Chr.9: 128946398-128946398 on Build GRCh38
rs1026201752 Chr.9: 128946316-128946316 on Build GRCh38
rs909238356 Chr.9: 128946240-128946240 on Build GRCh38
rs138962748 Chr.9: 128946047-128946047 on Build GRCh38
rs375939944 Chr.9: 128946293-128946293 on Build GRCh38
rs755691113 Chr.9: 128946014-128946014 on Build GRCh38
rs760442336 Chr.9: 128946181-128946181 on Build GRCh38
rs777885775 Chr.9: 128946049-128946049 on Build GRCh38
rs753795845 Chr.9: 128946114-128946114 on Build GRCh38
rs771007659 Chr.9: 128946055-128946055 on Build GRCh38
rs370282147 Chr.9: 128946024-128946024 on Build GRCh38
rs763260938 Chr.9: 128946095-128946095 on Build GRCh38
rs770667646 Chr.9: 128946250-128946250 on Build GRCh38
rs372057636 Chr.9: 128945989-128945989 on Build GRCh38
rs372577167 Chr.9: 128946057-128946057 on Build GRCh38
rs371882757 Chr.9: 128946239-128946239 on Build GRCh38
rs375959546 Chr.9: 128946012-128946012 on Build GRCh38
rs192515773 Chr.9: 128946134-128946134 on Build GRCh38
rs962516947 Chr.9: 128946188-128946188 on Build GRCh38
rs767133437 Chr.9: 128946107-128946107 on Build GRCh38
rs112448460 Chr.9: 128945992-128945992 on Build GRCh38
rs775869216 Chr.9: 128946166-128946166 on Build GRCh38
rs751762161 Chr.9: 128946218-128946218 on Build GRCh38
rs763859977 Chr.9: 128946207-128946207 on Build GRCh38
rs762835635 Chr.9: 128946304-128946304 on Build GRCh38
rs766759178 Chr.9: 128946390-128946390 on Build GRCh38
rs764117689 Chr.9: 128946305-128946305 on Build GRCh38
rs751318146 Chr.9: 128946309-128946309 on Build GRCh38
rs758196782 Chr.9: 128946202-128946202 on Build GRCh38
rs973710719 Chr.9: 128946083-128946083 on Build GRCh38
rs773789842 Chr.9: 128946439-128946441 on Build GRCh38
rs143854005 Chr.9: 128946382-128946382 on Build GRCh38
rs758129067 Chr.9: 128946139-128946139 on Build GRCh38
rs760249788 Chr.9: 128946109-128946109 on Build GRCh38
rs765151256 Chr.9: 128946200-128946200 on Build GRCh38
rs553112177 Chr.9: 128946165-128946165 on Build GRCh38
rs780974264 Chr.9: 128946396-128946396 on Build GRCh38
rs560708069 Chr.9: 128946158-128946158 on Build GRCh38
rs982668501 Chr.9: 128946126-128946126 on Build GRCh38
rs750702584 Chr.9: 128946337-128946337 on Build GRCh38
rs748688866 Chr.9: 128946108-128946108 on Build GRCh38
rs387907030 Chr.9: 128946082-128946082 on Build GRCh38
rs145125254 Chr.9: 128946042-128946042 on Build GRCh38
rs574562542 Chr.9: 128946018-128946018 on Build GRCh38
rs781428790 Chr.9: 128946233-128946233 on Build GRCh38
rs148118523 Chr.9: 128945990-128945990 on Build GRCh38
rs148646968 Chr.9: 128946431-128946431 on Build GRCh38
rs138453255 Chr.9: 128946225-128946225 on Build GRCh38
rs367925997 Chr.9: 128945984-128945984 on Build GRCh38
rs761583636 Chr.9: 128946430-128946430 on Build GRCh38
rs779523902 Chr.9: 128946074-128946074 on Build GRCh38
rs780038695 Chr.9: 128946339-128946339 on Build GRCh38
rs761604406 Chr.9: 128946313-128946313 on Build GRCh38
rs868232833 Chr.9: 128946175-128946175 on Build GRCh38
rs147342916 Chr.9: 128946428-128946428 on Build GRCh38
rs955235262 Chr.9: 128946099-128946099 on Build GRCh38
rs371529625 Chr.9: 128946406-128946406 on Build GRCh38
rs368251332 Chr.9: 128946238-128946238 on Build GRCh38
rs756216195 Chr.9: 128946338-128946338 on Build GRCh38
rs189623590 Chr.9: 128946021-128946021 on Build GRCh38
rs149462670 Chr.9: 128946329-128946329 on Build GRCh38
rs201143469 Chr.9: 128946414-128946414 on Build GRCh38
rs760856736 Chr.9: 128946435-128946435 on Build GRCh38
rs554098531 Chr.9: 128946410-128946410 on Build GRCh38
rs754346076 Chr.9: 128946352-128946352 on Build GRCh38
rs199535796 Chr.9: 128946237-128946237 on Build GRCh38
rs531714647 Chr.9: 128946380-128946380 on Build GRCh38
rs572213550 Chr.9: 128946094-128946094 on Build GRCh38
rs781302151 Chr.9: 128946148-128946148 on Build GRCh38
rs146395561 Chr.9: 128946190-128946190 on Build GRCh38
rs745885619 Chr.9: 128946234-128946234 on Build GRCh38
rs374317240 Chr.9: 128946174-128946174 on Build GRCh38
rs919111760 Chr.9: 128946102-128946103 on Build GRCh38
rs752585632 Chr.9: 128946201-128946201 on Build GRCh38
rs938092069 Chr.9: 128946151-128946151 on Build GRCh38
rs549060897 Chr.9: 128946191-128946191 on Build GRCh38
rs370084249 Chr.9: 128946299-128946299 on Build GRCh38
rs770030773 Chr.9: 128946022-128946022 on Build GRCh38
rs762471588 Chr.9: 128946371-128946379 on Build GRCh38
rs755389721 Chr.9: 128946367-128946367 on Build GRCh38
rs768517674 Chr.9: 128946424-128946424 on Build GRCh38
rs78758592 Chr.9: 128946220-128946220 on Build GRCh38
rs762558659 Chr.9: 128946419-128946419 on Build GRCh38
rs747023081 Chr.9: 128946054-128946054 on Build GRCh38
rs371490482 Chr.9: 128946236-128946236 on Build GRCh38
rs981981444 Chr.9: 128946333-128946333 on Build GRCh38
rs876657455 Chr.9: 128946269-128946269 on Build GRCh38
rs750733060 Chr.9: 128946161-128946161 on Build GRCh38
rs928628151 Chr.9: 128946363-128946363 on Build GRCh38

View Full Product Details