Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129620079-129620577 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf50
Entrez Gene ID
375759
Gene Name
chromosome 9 open reading frame 50
Gene Aliases
-
UniGene
Hs.124223
Gene Chromosome Location
Chr.9: 129612225-129622275 on Build GRCh38
Transcripts
NM_199350.3 XM_011518655.1 XM_011518656.1 XM_011518657.2 XM_011518658.2 XM_011518659.1 XM_011518660.1 XM_011518661.2 XM_011518662.2 XM_011518663.1 XM_017014708.1 NM_001286796.1 NM_001286797.1

Gene Symbol
NTMT1
Entrez Gene ID
28989
Gene Name
N-terminal Xaa-Pro-Lys N-methyltransferase 1
Gene Aliases
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A
UniGene
Hs.744027
Gene Chromosome Location
Chr.9: 129608884-129636742 on Build GRCh38
Transcripts
NM_199350.3 XM_011518655.1 XM_011518656.1 XM_011518657.2 XM_011518658.2 XM_011518659.1 XM_011518660.1 XM_011518661.2 XM_011518662.2 XM_011518663.1 XM_017014708.1 NM_001286796.1 NM_001286797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6240491 c.274C>A p.R92R Chr.9: 129620301-129620301 on Build GRCh38
3904708 c.57T>C p.P19P Chr.9: 129620518-129620518 on Build GRCh38
6499047 c.455_475del21 p.R152_L158delREFLHQL Chr.9: 129620100-129620120 on Build GRCh38
6616972 c.16C>T p.L6F Chr.9: 129620559-129620559 on Build GRCh38
6408674 c.401A>G p.E134G Chr.9: 129620174-129620174 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571561788 Chr.9: 129620350-129620350 on Build GRCh38
rs548738466 Chr.9: 129620155-129620155 on Build GRCh38
rs374267801 Chr.9: 129620525-129620525 on Build GRCh38
rs547222193 Chr.9: 129620432-129620432 on Build GRCh38
rs896460877 Chr.9: 129620417-129620417 on Build GRCh38
rs962226952 Chr.9: 129620290-129620290 on Build GRCh38
rs957158780 Chr.9: 129620309-129620309 on Build GRCh38
rs764915212 Chr.9: 129620538-129620538 on Build GRCh38
rs764531058 Chr.9: 129620459-129620459 on Build GRCh38
rs1001322600 Chr.9: 129620438-129620438 on Build GRCh38
rs774860449 Chr.9: 129620494-129620494 on Build GRCh38
rs551374227 Chr.9: 129620322-129620322 on Build GRCh38
rs995451980 Chr.9: 129620299-129620299 on Build GRCh38
rs752184672 Chr.9: 129620541-129620541 on Build GRCh38
rs922190541 Chr.9: 129620334-129620334 on Build GRCh38
rs754729275 Chr.9: 129620514-129620514 on Build GRCh38
rs976338600 Chr.9: 129620326-129620326 on Build GRCh38
rs1049161142 Chr.9: 129620133-129620133 on Build GRCh38
rs757183721 Chr.9: 129620549-129620549 on Build GRCh38
rs890125721 Chr.9: 129620450-129620450 on Build GRCh38
rs763807224 Chr.9: 129620515-129620515 on Build GRCh38
rs980741936 Chr.9: 129620339-129620339 on Build GRCh38
rs887884788 Chr.9: 129620159-129620159 on Build GRCh38
rs927971137 Chr.9: 129620345-129620345 on Build GRCh38
rs1015469942 Chr.9: 129620280-129620280 on Build GRCh38
rs944401966 Chr.9: 129620454-129620454 on Build GRCh38
rs988430582 Chr.9: 129620313-129620313 on Build GRCh38
rs13302395 Chr.9: 129620396-129620396 on Build GRCh38
rs1024658420 Chr.9: 129620269-129620269 on Build GRCh38
rs751493011 Chr.9: 129620544-129620544 on Build GRCh38
rs969885586 Chr.9: 129620277-129620277 on Build GRCh38
rs1036450074 Chr.9: 129620404-129620404 on Build GRCh38
rs746882730 Chr.9: 129620548-129620548 on Build GRCh38
rs768057809 Chr.9: 129620501-129620501 on Build GRCh38
rs1009152098 Chr.9: 129620264-129620264 on Build GRCh38
rs146618124 Chr.9: 129620420-129620420 on Build GRCh38
rs1034984810 Chr.9: 129620534-129620534 on Build GRCh38
rs932159897 Chr.9: 129620118-129620118 on Build GRCh38
rs1002292850 Chr.9: 129620512-129620512 on Build GRCh38
rs1036397309 Chr.9: 129620196-129620197 on Build GRCh38
rs960189428 Chr.9: 129620349-129620349 on Build GRCh38
rs547381419 Chr.9: 129620388-129620388 on Build GRCh38
rs377747523 Chr.9: 129620359-129620359 on Build GRCh38
rs1020472216 Chr.9: 129620509-129620509 on Build GRCh38
rs56148509 Chr.9: 129620511-129620511 on Build GRCh38
rs969491538 Chr.9: 129620321-129620321 on Build GRCh38
rs1042805379 Chr.9: 129620134-129620134 on Build GRCh38
rs866895180 Chr.9: 129620304-129620304 on Build GRCh38
rs1040556653 Chr.9: 129620363-129620363 on Build GRCh38
rs904304018 Chr.9: 129620142-129620142 on Build GRCh38
rs761365667 Chr.9: 129620547-129620547 on Build GRCh38
rs529361671 Chr.9: 129620130-129620130 on Build GRCh38
rs966104171 Chr.9: 129620325-129620325 on Build GRCh38
rs372378735 Chr.9: 129620440-129620440 on Build GRCh38
rs868575365 Chr.9: 129620540-129620540 on Build GRCh38
rs780122018 Chr.9: 129620502-129620502 on Build GRCh38
rs762269773 Chr.9: 129620496-129620496 on Build GRCh38
rs944859655 Chr.9: 129620348-129620348 on Build GRCh38
rs775470732 Chr.9: 129620446-129620446 on Build GRCh38
rs1036611530 Chr.9: 129620461-129620461 on Build GRCh38
rs111606531 Chr.9: 129620317-129620317 on Build GRCh38
rs763049533 Chr.9: 129620453-129620453 on Build GRCh38
rs368756086 Chr.9: 129620377-129620377 on Build GRCh38
rs925675719 Chr.9: 129620421-129620421 on Build GRCh38
rs1050064180 Chr.9: 129620195-129620195 on Build GRCh38
rs890684164 Chr.9: 129620263-129620263 on Build GRCh38
rs1004883372 Chr.9: 129620185-129620185 on Build GRCh38
rs1049297317 Chr.9: 129620362-129620362 on Build GRCh38
rs369716188 Chr.9: 129620213-129620213 on Build GRCh38
rs750800261 Chr.9: 129620503-129620503 on Build GRCh38
rs749921277 Chr.9: 129620098-129620098 on Build GRCh38
rs141348351 Chr.9: 129620423-129620423 on Build GRCh38
rs1001805189 Chr.9: 129620288-129620288 on Build GRCh38
rs898096301 Chr.9: 129620465-129620465 on Build GRCh38
rs781024658 Chr.9: 129620556-129620556 on Build GRCh38
rs995004659 Chr.9: 129620482-129620482 on Build GRCh38

View Full Product Details