Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129828939-129829446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf78
Entrez Gene ID
51759
Gene Name
chromosome 9 open reading frame 78
Gene Aliases
HCA59, HSPC220, bA409K20.3
UniGene
Hs.278429
Gene Chromosome Location
Chr.9: 129827285-129835293 on Build GRCh38
Transcripts
NM_016520.2 XM_011518780.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5596412 c.724_730delGAGCCCA p.E242fs*5 Chr.9: 129829253-129829259 on Build GRCh38
6455039 c.762G>A p.E254E Chr.9: 129829221-129829221 on Build GRCh38
3654831 c.685C>T p.H229Y Chr.9: 129829298-129829298 on Build GRCh38
7242836 c.677G>C p.R226T Chr.9: 129829407-129829407 on Build GRCh38
1460438 c.723_725delAGA p.E242delE Chr.9: 129829258-129829260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773890545 Chr.9: 129829352-129829352 on Build GRCh38
rs767660346 Chr.9: 129829342-129829342 on Build GRCh38
rs139248631 Chr.9: 129829282-129829282 on Build GRCh38
rs766977709 Chr.9: 129829374-129829374 on Build GRCh38
rs373452546 Chr.9: 129829284-129829284 on Build GRCh38
rs112483441 Chr.9: 129829026-129829026 on Build GRCh38
rs765431969 Chr.9: 129829031-129829031 on Build GRCh38
rs542228560 Chr.9: 129829100-129829100 on Build GRCh38
rs374249652 Chr.9: 129829185-129829185 on Build GRCh38
rs76146811 Chr.9: 129829121-129829121 on Build GRCh38
rs745707856 Chr.9: 129829212-129829212 on Build GRCh38
rs756834298 Chr.9: 129829061-129829061 on Build GRCh38
rs756733770 Chr.9: 129829298-129829298 on Build GRCh38
rs763698426 Chr.9: 129829387-129829387 on Build GRCh38
rs758374179 Chr.9: 129829051-129829051 on Build GRCh38
rs752890879 Chr.9: 129829140-129829140 on Build GRCh38
rs772308905 Chr.9: 129829329-129829329 on Build GRCh38
rs376381202 Chr.9: 129829325-129829325 on Build GRCh38
rs768829061 Chr.9: 129829334-129829334 on Build GRCh38
rs201417433 Chr.9: 129829192-129829192 on Build GRCh38
rs967599854 Chr.9: 129829259-129829259 on Build GRCh38
rs56195701 Chr.9: 129829130-129829130 on Build GRCh38
rs778707570 Chr.9: 129829123-129829123 on Build GRCh38
rs910607987 Chr.9: 129829351-129829351 on Build GRCh38
rs926593270 Chr.9: 129829166-129829166 on Build GRCh38
rs374427651 Chr.9: 129829376-129829376 on Build GRCh38
rs937249688 Chr.9: 129828975-129828975 on Build GRCh38
rs929881971 Chr.9: 129829397-129829397 on Build GRCh38
rs1007823265 Chr.9: 129829317-129829317 on Build GRCh38
rs3818553 Chr.9: 129829230-129829230 on Build GRCh38
rs199922761 Chr.9: 129829368-129829368 on Build GRCh38
rs1041515188 Chr.9: 129829032-129829032 on Build GRCh38
rs761852463 Chr.9: 129829337-129829337 on Build GRCh38
rs771646091 Chr.9: 129829186-129829186 on Build GRCh38
rs774602738 Chr.9: 129829335-129829335 on Build GRCh38
rs764968548 Chr.9: 129829263-129829265 on Build GRCh38
rs770840294 Chr.9: 129829195-129829195 on Build GRCh38
rs55962905 Chr.9: 129828993-129828993 on Build GRCh38
rs777087140 Chr.9: 129829189-129829189 on Build GRCh38
rs752840133 Chr.9: 129829033-129829033 on Build GRCh38
rs532995748 Chr.9: 129829164-129829164 on Build GRCh38
rs761952121 Chr.9: 129829255-129829255 on Build GRCh38
rs760491729 Chr.9: 129829131-129829131 on Build GRCh38
rs776467026 Chr.9: 129829132-129829132 on Build GRCh38
rs773375419 Chr.9: 129829330-129829330 on Build GRCh38
rs952129557 Chr.9: 129829144-129829144 on Build GRCh38
rs778389912 Chr.9: 129829069-129829069 on Build GRCh38
rs768746773 Chr.9: 129828971-129828972 on Build GRCh38
rs755335649 Chr.9: 129829378-129829378 on Build GRCh38
rs756104638 Chr.9: 129829412-129829412 on Build GRCh38
rs564233944 Chr.9: 129829161-129829161 on Build GRCh38
rs201526319 Chr.9: 129829373-129829373 on Build GRCh38
rs769525413 Chr.9: 129829225-129829225 on Build GRCh38
rs537836441 Chr.9: 129829101-129829101 on Build GRCh38
rs780612020 Chr.9: 129829394-129829394 on Build GRCh38
rs1055641598 Chr.9: 129828984-129828984 on Build GRCh38
rs751175479 Chr.9: 129829390-129829390 on Build GRCh38
rs765483588 Chr.9: 129829137-129829137 on Build GRCh38
rs758042858 Chr.9: 129829283-129829283 on Build GRCh38
rs199630059 Chr.9: 129829273-129829273 on Build GRCh38
rs745816483 Chr.9: 129829396-129829396 on Build GRCh38
rs140049867 Chr.9: 129829205-129829205 on Build GRCh38
rs750944606 Chr.9: 129829274-129829274 on Build GRCh38
rs759321072 Chr.9: 129829136-129829136 on Build GRCh38
rs7047271 Chr.9: 129829146-129829146 on Build GRCh38
rs994114431 Chr.9: 129829366-129829366 on Build GRCh38
rs199549692 Chr.9: 129829174-129829174 on Build GRCh38
rs748285215 Chr.9: 129829307-129829307 on Build GRCh38
rs754061519 Chr.9: 129829004-129829004 on Build GRCh38
rs763016070 Chr.9: 129829245-129829245 on Build GRCh38
rs764037911 Chr.9: 129829150-129829150 on Build GRCh38
rs977601405 Chr.9: 129829260-129829260 on Build GRCh38
rs200595056 Chr.9: 129829251-129829251 on Build GRCh38
rs752113900 Chr.9: 129829160-129829160 on Build GRCh38
rs756571102 Chr.9: 129829275-129829275 on Build GRCh38
rs192531241 Chr.9: 129829125-129829125 on Build GRCh38
rs746537716 Chr.9: 129829286-129829286 on Build GRCh38
rs7032945 Chr.9: 129829093-129829093 on Build GRCh38
rs549811347 Chr.9: 129828995-129828995 on Build GRCh38
rs200922853 Chr.9: 129829289-129829289 on Build GRCh38
rs769579469 Chr.9: 129829312-129829312 on Build GRCh38
rs764250406 Chr.9: 129829246-129829246 on Build GRCh38
rs902913830 Chr.9: 129829079-129829079 on Build GRCh38
rs751958133 Chr.9: 129829090-129829090 on Build GRCh38
rs756791022 Chr.9: 129829393-129829393 on Build GRCh38
rs766811260 Chr.9: 129829281-129829281 on Build GRCh38
rs372860206 Chr.9: 129829182-129829182 on Build GRCh38
rs376310519 Chr.9: 129829184-129829184 on Build GRCh38
rs750888164 Chr.9: 129829173-129829173 on Build GRCh38
rs553733967 Chr.9: 129829027-129829027 on Build GRCh38

View Full Product Details