Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 129834988-129835479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf78
Entrez Gene ID
51759
Gene Name
chromosome 9 open reading frame 78
Gene Aliases
HCA59, HSPC220, bA409K20.3
UniGene
Hs.278429
Gene Chromosome Location
Chr.9: 129827285-129835293 on Build GRCh38
Transcripts
NM_016520.2 XM_011518780.2 NM_001008563.4 NM_001110303.3 NM_006676.7 XM_005251665.3 XM_011518161.2

Gene Symbol
USP20
Entrez Gene ID
10868
Gene Name
ubiquitin specific peptidase 20
Gene Aliases
LSFR3A, VDU2, hVDU2
UniGene
Hs.5452
Gene Chromosome Location
Chr.9: 129835417-129881838 on Build GRCh38
Transcripts
NM_016520.2 XM_011518780.2 NM_001008563.4 NM_001110303.3 NM_006676.7 XM_005251665.3 XM_011518161.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
346097 c.78G>T p.E26D Chr.9: 129835144-129835144 on Build GRCh38
5960652 c.44C>T p.S15L Chr.9: 129835178-129835178 on Build GRCh38
3952383 c.37G>A p.G13S Chr.9: 129835185-129835185 on Build GRCh38
5034074 c.80T>G p.V27G Chr.9: 129835142-129835142 on Build GRCh38
1105884 c.82C>A p.R28R Chr.9: 129835140-129835140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019020931 Chr.9: 129835297-129835297 on Build GRCh38
rs766391642 Chr.9: 129835241-129835241 on Build GRCh38
rs1051122480 Chr.9: 129835085-129835085 on Build GRCh38
rs201516060 Chr.9: 129835169-129835169 on Build GRCh38
rs1048217525 Chr.9: 129835122-129835122 on Build GRCh38
rs919985997 Chr.9: 129835359-129835360 on Build GRCh38
rs557823140 Chr.9: 129835266-129835266 on Build GRCh38
rs560383269 Chr.9: 129835055-129835055 on Build GRCh38
rs760074489 Chr.9: 129835238-129835238 on Build GRCh38
rs755867968 Chr.9: 129835093-129835093 on Build GRCh38
rs902959094 Chr.9: 129835283-129835283 on Build GRCh38
rs1055967037 Chr.9: 129835397-129835397 on Build GRCh38
rs916997277 Chr.9: 129835259-129835259 on Build GRCh38
rs923249491 Chr.9: 129835015-129835015 on Build GRCh38
rs774199710 Chr.9: 129835429-129835429 on Build GRCh38
rs907424642 Chr.9: 129835267-129835267 on Build GRCh38
rs986096175 Chr.9: 129835045-129835045 on Build GRCh38
rs201895521 Chr.9: 129835231-129835231 on Build GRCh38
rs937090158 Chr.9: 129835254-129835254 on Build GRCh38
rs748868375 Chr.9: 129835272-129835272 on Build GRCh38
rs758662635 Chr.9: 129835198-129835199 on Build GRCh38
rs577451497 Chr.9: 129835279-129835279 on Build GRCh38
rs1041345660 Chr.9: 129835263-129835263 on Build GRCh38
rs965278047 Chr.9: 129835065-129835065 on Build GRCh38
rs901931914 Chr.9: 129835298-129835298 on Build GRCh38
rs773995958 Chr.9: 129835233-129835233 on Build GRCh38
rs536693008 Chr.9: 129835237-129835237 on Build GRCh38
rs572810270 Chr.9: 129835438-129835438 on Build GRCh38
rs376128096 Chr.9: 129835367-129835367 on Build GRCh38
rs371917470 Chr.9: 129835225-129835225 on Build GRCh38
rs148302016 Chr.9: 129835186-129835186 on Build GRCh38
rs761432344 Chr.9: 129835047-129835048 on Build GRCh38
rs571223625 Chr.9: 129835060-129835060 on Build GRCh38
rs1026279543 Chr.9: 129835315-129835315 on Build GRCh38
rs1030539488 Chr.9: 129835460-129835460 on Build GRCh38
rs938895293 Chr.9: 129835392-129835392 on Build GRCh38
rs1026020421 Chr.9: 129835239-129835239 on Build GRCh38
rs781737180 Chr.9: 129835255-129835255 on Build GRCh38
rs953453087 Chr.9: 129835022-129835022 on Build GRCh38
rs775892516 Chr.9: 129835261-129835261 on Build GRCh38
rs986049933 Chr.9: 129835306-129835306 on Build GRCh38
rs993460770 Chr.9: 129835305-129835305 on Build GRCh38
rs878895139 Chr.9: 129835290-129835290 on Build GRCh38
rs748345169 Chr.9: 129835216-129835216 on Build GRCh38
rs868794117 Chr.9: 129835374-129835374 on Build GRCh38
rs761390965 Chr.9: 129835043-129835044 on Build GRCh38
rs951386530 Chr.9: 129835316-129835316 on Build GRCh38
rs761344335 Chr.9: 129835191-129835191 on Build GRCh38
rs370856154 Chr.9: 129835208-129835208 on Build GRCh38
rs917007194 Chr.9: 129835372-129835372 on Build GRCh38
rs1017573626 Chr.9: 129835328-129835328 on Build GRCh38
rs749025411 Chr.9: 129835112-129835112 on Build GRCh38
rs747158880 Chr.9: 129835252-129835252 on Build GRCh38
rs937039318 Chr.9: 129835126-129835126 on Build GRCh38
rs911868183 Chr.9: 129835064-129835064 on Build GRCh38
rs765061525 Chr.9: 129835246-129835246 on Build GRCh38
rs747052904 Chr.9: 129835229-129835229 on Build GRCh38
rs977505329 Chr.9: 129835393-129835393 on Build GRCh38
rs779249106 Chr.9: 129835121-129835121 on Build GRCh38
rs991321382 Chr.9: 129835363-129835363 on Build GRCh38
rs1047753711 Chr.9: 129835154-129835154 on Build GRCh38
rs767064494 Chr.9: 129835235-129835235 on Build GRCh38
rs890291782 Chr.9: 129835461-129835461 on Build GRCh38
rs757234139 Chr.9: 129835253-129835253 on Build GRCh38
rs769068705 Chr.9: 129835115-129835115 on Build GRCh38
rs529150297 Chr.9: 129835343-129835343 on Build GRCh38
rs930094273 Chr.9: 129835366-129835366 on Build GRCh38
rs1021780803 Chr.9: 129835280-129835280 on Build GRCh38
rs17851537 Chr.9: 129835183-129835183 on Build GRCh38
rs1042028495 Chr.9: 129835201-129835201 on Build GRCh38
rs1038514233 Chr.9: 129835098-129835098 on Build GRCh38
rs374498980 Chr.9: 129835248-129835248 on Build GRCh38
rs976666926 Chr.9: 129835087-129835087 on Build GRCh38
rs766045600 Chr.9: 129835376-129835376 on Build GRCh38
rs145780019 Chr.9: 129835245-129835245 on Build GRCh38
rs531329143 Chr.9: 129835437-129835437 on Build GRCh38
rs199979480 Chr.9: 129835222-129835222 on Build GRCh38
rs11790613 Chr.9: 129835342-129835342 on Build GRCh38
rs894146856 Chr.9: 129835273-129835275 on Build GRCh38
rs761183374 Chr.9: 129835161-129835161 on Build GRCh38
rs894761829 Chr.9: 129835423-129835423 on Build GRCh38
rs901941742 Chr.9: 129835459-129835459 on Build GRCh38
rs928812599 Chr.9: 129835381-129835381 on Build GRCh38
rs370052266 Chr.9: 129835232-129835232 on Build GRCh38
rs771324874 Chr.9: 129835166-129835166 on Build GRCh38
rs772233601 Chr.9: 129835145-129835145 on Build GRCh38
rs759785863 Chr.9: 129835177-129835177 on Build GRCh38
rs761397322 Chr.9: 129835234-129835234 on Build GRCh38
rs540403157 Chr.9: 129835339-129835339 on Build GRCh38
rs755939769 Chr.9: 129835089-129835090 on Build GRCh38
rs1055053172 Chr.9: 129835274-129835274 on Build GRCh38
rs750341360 Chr.9: 129835204-129835204 on Build GRCh38
rs770309165 Chr.9: 129835273-129835273 on Build GRCh38
rs773627349 Chr.9: 129835151-129835151 on Build GRCh38
rs895457783 Chr.9: 129835180-129835180 on Build GRCh38
rs984029629 Chr.9: 129835331-129835331 on Build GRCh38
rs200977485 Chr.9: 129835249-129835249 on Build GRCh38
rs1000775996 Chr.9: 129835221-129835221 on Build GRCh38
rs753708289 Chr.9: 129835207-129835207 on Build GRCh38
rs370589788 Chr.9: 129835262-129835262 on Build GRCh38
rs779841642 Chr.9: 129835096-129835096 on Build GRCh38
rs924278199 Chr.9: 129835409-129835409 on Build GRCh38
rs756134527 Chr.9: 129835206-129835206 on Build GRCh38
rs542264057 Chr.9: 129835353-129835353 on Build GRCh38
rs770157065 Chr.9: 129835256-129835256 on Build GRCh38
rs918492887 Chr.9: 129835116-129835116 on Build GRCh38
rs766946594 Chr.9: 129835202-129835202 on Build GRCh38
rs571348027 Chr.9: 129835443-129835443 on Build GRCh38
rs779302258 Chr.9: 129835213-129835213 on Build GRCh38
rs562418793 Chr.9: 129835424-129835424 on Build GRCh38
rs958416458 Chr.9: 129835185-129835185 on Build GRCh38
rs553422756 Chr.9: 129835230-129835230 on Build GRCh38
rs199532327 Chr.9: 129835125-129835125 on Build GRCh38
rs753663441 Chr.9: 129835147-129835148 on Build GRCh38
rs13294710 Chr.9: 129835442-129835442 on Build GRCh38
rs779654293 Chr.9: 129835102-129835102 on Build GRCh38
rs998624478 Chr.9: 129835457-129835457 on Build GRCh38
rs933968446 Chr.9: 129835025-129835025 on Build GRCh38

View Full Product Details