Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50284523-50285071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNB3
Entrez Gene ID
85417
Gene Name
cyclin B3
Gene Aliases
-
UniGene
Hs.130310
Gene Chromosome Location
Chr.X: 50202713-50351914 on Build GRCh38
Transcripts
NM_033031.2 NM_033670.2 XM_017029913.1 XM_017029914.1 XM_017029915.1 XM_017029916.1 XM_017029917.1 XM_017029918.1

SNP Detail

SNP ID Chromosome Location
rs992532464 Chr.X: 50284727-50284727 on Build GRCh38
rs935884129 Chr.X: 50284999-50284999 on Build GRCh38
rs924084808 Chr.X: 50284680-50284680 on Build GRCh38
rs747516334 Chr.X: 50284854-50284854 on Build GRCh38
rs57816053 Chr.X: 50284728-50284728 on Build GRCh38
rs185089009 Chr.X: 50284559-50284559 on Build GRCh38
rs771226264 Chr.X: 50284922-50284922 on Build GRCh38
rs915278173 Chr.X: 50284771-50284771 on Build GRCh38
rs1024290327 Chr.X: 50284558-50284558 on Build GRCh38
rs12401117 Chr.X: 50284651-50284651 on Build GRCh38
rs376642965 Chr.X: 50285020-50285021 on Build GRCh38
rs189772191 Chr.X: 50284712-50284712 on Build GRCh38
rs953051856 Chr.X: 50284650-50284650 on Build GRCh38
rs773549751 Chr.X: 50284838-50284838 on Build GRCh38
rs566231294 Chr.X: 50284746-50284746 on Build GRCh38
rs73493796 Chr.X: 50284609-50284609 on Build GRCh38
rs901166630 Chr.X: 50284607-50284607 on Build GRCh38
rs780753327 Chr.X: 50284794-50284794 on Build GRCh38
rs190166916 Chr.X: 50284596-50284596 on Build GRCh38
rs748720946 Chr.X: 50284745-50284745 on Build GRCh38
rs1050388021 Chr.X: 50284818-50284818 on Build GRCh38
rs931971840 Chr.X: 50284777-50284777 on Build GRCh38
rs182365971 Chr.X: 50284630-50284630 on Build GRCh38
rs961527313 Chr.X: 50284715-50284715 on Build GRCh38
rs73493793 Chr.X: 50284583-50284583 on Build GRCh38
rs747051446 Chr.X: 50284801-50284801 on Build GRCh38
rs1036171587 Chr.X: 50284866-50284868 on Build GRCh38
rs752714416 Chr.X: 50284578-50284578 on Build GRCh38
rs73493797 Chr.X: 50285045-50285045 on Build GRCh38
rs1009618913 Chr.X: 50284579-50284579 on Build GRCh38
rs142579882 Chr.X: 50284688-50284688 on Build GRCh38
rs748411151 Chr.X: 50285005-50285005 on Build GRCh38

View Full Product Details