Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50370200-50370712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKK
Entrez Gene ID
139189
Gene Name
diacylglycerol kinase kappa
Gene Aliases
-
UniGene
Hs.38132
Gene Chromosome Location
Chr.X: 50365407-50470850 on Build GRCh38
Transcripts
NM_001013742.3 XM_017029268.1

SNP Detail

SNP ID Chromosome Location
rs956096808 Chr.X: 50370512-50370512 on Build GRCh38
rs781827565 Chr.X: 50370384-50370384 on Build GRCh38
rs782288714 Chr.X: 50370409-50370409 on Build GRCh38
rs571444713 Chr.X: 50370506-50370506 on Build GRCh38
rs782004804 Chr.X: 50370483-50370483 on Build GRCh38
rs782647091 Chr.X: 50370413-50370413 on Build GRCh38
rs782362762 Chr.X: 50370564-50370564 on Build GRCh38
rs1056517676 Chr.X: 50370381-50370381 on Build GRCh38
rs782063467 Chr.X: 50370592-50370592 on Build GRCh38
rs987595326 Chr.X: 50370529-50370529 on Build GRCh38
rs782050850 Chr.X: 50370343-50370343 on Build GRCh38
rs1047888236 Chr.X: 50370292-50370292 on Build GRCh38
rs886165459 Chr.X: 50370298-50370298 on Build GRCh38
rs782388594 Chr.X: 50370257-50370257 on Build GRCh38
rs782073603 Chr.X: 50370561-50370561 on Build GRCh38
rs903745554 Chr.X: 50370679-50370679 on Build GRCh38
rs999338085 Chr.X: 50370492-50370492 on Build GRCh38
rs150271614 Chr.X: 50370586-50370586 on Build GRCh38
rs782673050 Chr.X: 50370651-50370651 on Build GRCh38
rs41308608 Chr.X: 50370596-50370596 on Build GRCh38
rs782782712 Chr.X: 50370266-50370266 on Build GRCh38
rs781816071 Chr.X: 50370521-50370521 on Build GRCh38
rs782525357 Chr.X: 50370526-50370526 on Build GRCh38
rs1043432719 Chr.X: 50370674-50370674 on Build GRCh38
rs930778922 Chr.X: 50370275-50370275 on Build GRCh38
rs782384452 Chr.X: 50370400-50370400 on Build GRCh38
rs939096759 Chr.X: 50370634-50370634 on Build GRCh38
rs894850842 Chr.X: 50370446-50370446 on Build GRCh38
rs781955362 Chr.X: 50370566-50370566 on Build GRCh38
rs782552872 Chr.X: 50370539-50370539 on Build GRCh38
rs200519087 Chr.X: 50370469-50370469 on Build GRCh38
rs376700599 Chr.X: 50370573-50370573 on Build GRCh38
rs781800974 Chr.X: 50370268-50370268 on Build GRCh38
rs782158470 Chr.X: 50370488-50370488 on Build GRCh38
rs782012493 Chr.X: 50370587-50370587 on Build GRCh38
rs782252945 Chr.X: 50370385-50370385 on Build GRCh38
rs782558579 Chr.X: 50370619-50370619 on Build GRCh38
rs782157435 Chr.X: 50370265-50370265 on Build GRCh38
rs781809441 Chr.X: 50370502-50370502 on Build GRCh38
rs781979060 Chr.X: 50370261-50370261 on Build GRCh38
rs782189565 Chr.X: 50370552-50370552 on Build GRCh38
rs947707743 Chr.X: 50370662-50370662 on Build GRCh38
rs141122494 Chr.X: 50370538-50370538 on Build GRCh38
rs782522482 Chr.X: 50370402-50370402 on Build GRCh38
rs782749655 Chr.X: 50370312-50370312 on Build GRCh38
rs782594888 Chr.X: 50370542-50370542 on Build GRCh38
rs782099622 Chr.X: 50370464-50370464 on Build GRCh38
rs782738180 Chr.X: 50370514-50370514 on Build GRCh38
rs782620144 Chr.X: 50370534-50370534 on Build GRCh38
rs782791352 Chr.X: 50370497-50370497 on Build GRCh38
rs781924107 Chr.X: 50370433-50370433 on Build GRCh38
rs782747182 Chr.X: 50370346-50370346 on Build GRCh38
rs894897195 Chr.X: 50370661-50370661 on Build GRCh38

View Full Product Details