Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 50375918-50376424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DGKK
Entrez Gene ID
139189
Gene Name
diacylglycerol kinase kappa
Gene Aliases
-
UniGene
Hs.38132
Gene Chromosome Location
Chr.X: 50365407-50470850 on Build GRCh38
Transcripts
NM_001013742.3 XM_017029268.1

SNP Detail

SNP ID Chromosome Location
rs942780893 Chr.X: 50376176-50376176 on Build GRCh38
rs782258880 Chr.X: 50376079-50376079 on Build GRCh38
rs929944960 Chr.X: 50376271-50376271 on Build GRCh38
rs781822025 Chr.X: 50375987-50375987 on Build GRCh38
rs369521039 Chr.X: 50376102-50376102 on Build GRCh38
rs4074319 Chr.X: 50376189-50376189 on Build GRCh38
rs782050571 Chr.X: 50375978-50375978 on Build GRCh38
rs782533063 Chr.X: 50376032-50376032 on Build GRCh38
rs977116150 Chr.X: 50376062-50376062 on Build GRCh38
rs782445375 Chr.X: 50376208-50376208 on Build GRCh38
rs782038459 Chr.X: 50376101-50376101 on Build GRCh38
rs782737383 Chr.X: 50375981-50375981 on Build GRCh38
rs910425461 Chr.X: 50376122-50376122 on Build GRCh38
rs376715528 Chr.X: 50376347-50376347 on Build GRCh38
rs370337711 Chr.X: 50376123-50376123 on Build GRCh38
rs185005508 Chr.X: 50376028-50376028 on Build GRCh38
rs781976960 Chr.X: 50376168-50376168 on Build GRCh38
rs782727183 Chr.X: 50376161-50376161 on Build GRCh38
rs188642156 Chr.X: 50376075-50376075 on Build GRCh38
rs782794033 Chr.X: 50376322-50376322 on Build GRCh38
rs181431039 Chr.X: 50376323-50376323 on Build GRCh38
rs782553594 Chr.X: 50376348-50376348 on Build GRCh38
rs372991252 Chr.X: 50376107-50376107 on Build GRCh38
rs782771151 Chr.X: 50375992-50375992 on Build GRCh38
rs782189272 Chr.X: 50376049-50376049 on Build GRCh38
rs1047071679 Chr.X: 50376310-50376310 on Build GRCh38
rs4074321 Chr.X: 50375958-50375958 on Build GRCh38
rs782078073 Chr.X: 50376110-50376110 on Build GRCh38
rs898584085 Chr.X: 50376237-50376237 on Build GRCh38
rs4074320 Chr.X: 50376086-50376086 on Build GRCh38
rs782588606 Chr.X: 50376044-50376044 on Build GRCh38
rs782789213 Chr.X: 50376191-50376191 on Build GRCh38
rs192664195 Chr.X: 50375947-50375947 on Build GRCh38
rs782416103 Chr.X: 50376318-50376318 on Build GRCh38
rs782505508 Chr.X: 50375988-50375988 on Build GRCh38
rs782503001 Chr.X: 50376074-50376074 on Build GRCh38
rs782113744 Chr.X: 50376198-50376198 on Build GRCh38

View Full Product Details