Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54448719-54449224 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD1
Entrez Gene ID
2245
Gene Name
FYVE, RhoGEF and PH domain containing 1
Gene Aliases
AAS, FGDY, MRXS16, ZFYVE3
UniGene
Hs.709201
Gene Chromosome Location
Chr.X: 54445454-54496166 on Build GRCh38
Transcripts
NM_004463.2

SNP Detail

SNP ID Chromosome Location
rs1057418327 Chr.X: 54449052-54449052 on Build GRCh38
rs763913024 Chr.X: 54449128-54449128 on Build GRCh38
rs781020308 Chr.X: 54448988-54448988 on Build GRCh38
rs759432830 Chr.X: 54449115-54449115 on Build GRCh38
rs947151239 Chr.X: 54449020-54449020 on Build GRCh38
rs765426125 Chr.X: 54448902-54448902 on Build GRCh38
rs751512554 Chr.X: 54448933-54448933 on Build GRCh38
rs762003845 Chr.X: 54448890-54448890 on Build GRCh38
rs398124163 Chr.X: 54449160-54449160 on Build GRCh38
rs770514575 Chr.X: 54448802-54448802 on Build GRCh38
rs934603263 Chr.X: 54449050-54449050 on Build GRCh38
rs774020303 Chr.X: 54449122-54449122 on Build GRCh38
rs774928867 Chr.X: 54449010-54449010 on Build GRCh38
rs35977564 Chr.X: 54449056-54449056 on Build GRCh38
rs368521395 Chr.X: 54449127-54449127 on Build GRCh38
rs370134555 Chr.X: 54448997-54448997 on Build GRCh38
rs201698944 Chr.X: 54449102-54449102 on Build GRCh38
rs768912998 Chr.X: 54448863-54448863 on Build GRCh38
rs376552943 Chr.X: 54448758-54448758 on Build GRCh38
rs780985219 Chr.X: 54448755-54448755 on Build GRCh38
rs185771737 Chr.X: 54448769-54448769 on Build GRCh38
rs201232039 Chr.X: 54449110-54449110 on Build GRCh38
rs769198571 Chr.X: 54449083-54449083 on Build GRCh38
rs748784763 Chr.X: 54448801-54448801 on Build GRCh38
rs761173572 Chr.X: 54448844-54448844 on Build GRCh38
rs150865566 Chr.X: 54448953-54448953 on Build GRCh38
rs372642699 Chr.X: 54449005-54449005 on Build GRCh38
rs763053573 Chr.X: 54448923-54448923 on Build GRCh38
rs374192869 Chr.X: 54449124-54449124 on Build GRCh38
rs190570287 Chr.X: 54449055-54449055 on Build GRCh38
rs376026003 Chr.X: 54448973-54448973 on Build GRCh38
rs772753650 Chr.X: 54448746-54448746 on Build GRCh38
rs747523280 Chr.X: 54449069-54449069 on Build GRCh38
rs773377324 Chr.X: 54448917-54448917 on Build GRCh38
rs777036753 Chr.X: 54448884-54448884 on Build GRCh38
rs754931200 Chr.X: 54448939-54448939 on Build GRCh38
rs769484631 Chr.X: 54448779-54448779 on Build GRCh38
rs910292238 Chr.X: 54448895-54448895 on Build GRCh38
rs778629490 Chr.X: 54449204-54449204 on Build GRCh38
rs776913174 Chr.X: 54449098-54449098 on Build GRCh38
rs776170341 Chr.X: 54448833-54448833 on Build GRCh38
rs373950992 Chr.X: 54448928-54448928 on Build GRCh38
rs756828939 Chr.X: 54449009-54449009 on Build GRCh38
rs61734178 Chr.X: 54449149-54449149 on Build GRCh38
rs773430334 Chr.X: 54449109-54449109 on Build GRCh38
rs202213679 Chr.X: 54449194-54449194 on Build GRCh38
rs777465420 Chr.X: 54448795-54448795 on Build GRCh38
rs770106297 Chr.X: 54449107-54449107 on Build GRCh38
rs375922720 Chr.X: 54449099-54449099 on Build GRCh38

View Full Product Details