Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 54449570-54450067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD1
Entrez Gene ID
2245
Gene Name
FYVE, RhoGEF and PH domain containing 1
Gene Aliases
AAS, FGDY, MRXS16, ZFYVE3
UniGene
Hs.709201
Gene Chromosome Location
Chr.X: 54445454-54496166 on Build GRCh38
Transcripts
NM_004463.2

SNP Detail

SNP ID Chromosome Location
rs191379839 Chr.X: 54449854-54449854 on Build GRCh38
rs904182044 Chr.X: 54449835-54449835 on Build GRCh38
rs12011120 Chr.X: 54449716-54449716 on Build GRCh38
rs398124162 Chr.X: 54449671-54449672 on Build GRCh38
rs941339199 Chr.X: 54449675-54449675 on Build GRCh38
rs765315414 Chr.X: 54449999-54449999 on Build GRCh38
rs746008535 Chr.X: 54449731-54449731 on Build GRCh38
rs1031406199 Chr.X: 54449932-54449932 on Build GRCh38
rs955927834 Chr.X: 54449942-54449942 on Build GRCh38
rs999986584 Chr.X: 54449872-54449872 on Build GRCh38
rs10127419 Chr.X: 54449956-54449956 on Build GRCh38
rs368646447 Chr.X: 54449765-54449765 on Build GRCh38
rs12560189 Chr.X: 54449964-54449964 on Build GRCh38
rs772204559 Chr.X: 54449749-54449749 on Build GRCh38
rs750008353 Chr.X: 54449642-54449642 on Build GRCh38
rs757279999 Chr.X: 54449624-54449624 on Build GRCh38
rs775415679 Chr.X: 54449752-54449752 on Build GRCh38
rs776613296 Chr.X: 54449800-54449800 on Build GRCh38
rs182805935 Chr.X: 54449899-54449899 on Build GRCh38
rs761542919 Chr.X: 54449647-54449647 on Build GRCh38
rs897246819 Chr.X: 54449725-54449725 on Build GRCh38
rs764968569 Chr.X: 54449634-54449634 on Build GRCh38
rs1126744 Chr.X: 54449671-54449671 on Build GRCh38
rs1047575842 Chr.X: 54449792-54449792 on Build GRCh38
rs970381483 Chr.X: 54449990-54449990 on Build GRCh38
rs1010522109 Chr.X: 54449959-54449959 on Build GRCh38
rs779226954 Chr.X: 54449726-54449726 on Build GRCh38
rs746687912 Chr.X: 54449834-54449834 on Build GRCh38
rs926658541 Chr.X: 54450012-54450012 on Build GRCh38
rs1024116526 Chr.X: 54449960-54449960 on Build GRCh38
rs398124161 Chr.X: 54449694-54449695 on Build GRCh38
rs768678687 Chr.X: 54449791-54449791 on Build GRCh38
rs757933033 Chr.X: 54449661-54449661 on Build GRCh38
rs749630642 Chr.X: 54449701-54449701 on Build GRCh38
rs753784232 Chr.X: 54449608-54449608 on Build GRCh38
rs993082966 Chr.X: 54449958-54449958 on Build GRCh38
rs375527081 Chr.X: 54449626-54449626 on Build GRCh38
rs139366395 Chr.X: 54449823-54449823 on Build GRCh38
rs762287335 Chr.X: 54449952-54449952 on Build GRCh38
rs536148280 Chr.X: 54449824-54449824 on Build GRCh38
rs778151689 Chr.X: 54449698-54449698 on Build GRCh38
rs922736654 Chr.X: 54449598-54449598 on Build GRCh38
rs756611520 Chr.X: 54449683-54449683 on Build GRCh38

View Full Product Details