Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55624835-55625379 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXR2
Entrez Gene ID
139628
Gene Name
forkhead box R2
Gene Aliases
FOXN6
UniGene
Hs.364260
Gene Chromosome Location
Chr.X: 55623400-55626188 on Build GRCh38
Transcripts
NM_198451.3

SNP Detail

SNP ID Chromosome Location
rs987463472 Chr.X: 55625207-55625207 on Build GRCh38
rs758537552 Chr.X: 55624903-55624903 on Build GRCh38
rs749019104 Chr.X: 55625253-55625253 on Build GRCh38
rs186221540 Chr.X: 55624871-55624871 on Build GRCh38
rs761473834 Chr.X: 55625238-55625238 on Build GRCh38
rs879041211 Chr.X: 55624903-55624903 on Build GRCh38
rs935829005 Chr.X: 55625356-55625356 on Build GRCh38
rs1018925688 Chr.X: 55625263-55625263 on Build GRCh38
rs753583404 Chr.X: 55625031-55625031 on Build GRCh38
rs1017112988 Chr.X: 55624992-55624992 on Build GRCh38
rs764826929 Chr.X: 55625276-55625276 on Build GRCh38
rs961645678 Chr.X: 55625127-55625127 on Build GRCh38
rs748098451 Chr.X: 55625010-55625010 on Build GRCh38
rs780223322 Chr.X: 55624988-55624988 on Build GRCh38
rs905544702 Chr.X: 55624982-55624982 on Build GRCh38
rs780521741 Chr.X: 55625312-55625312 on Build GRCh38
rs950984002 Chr.X: 55625145-55625145 on Build GRCh38
rs777729231 Chr.X: 55625201-55625201 on Build GRCh38
rs973383038 Chr.X: 55625217-55625217 on Build GRCh38
rs769810519 Chr.X: 55625030-55625030 on Build GRCh38
rs749901548 Chr.X: 55625301-55625301 on Build GRCh38
rs949772264 Chr.X: 55625246-55625246 on Build GRCh38
rs995443933 Chr.X: 55625108-55625108 on Build GRCh38
rs998302466 Chr.X: 55624999-55624999 on Build GRCh38
rs984260744 Chr.X: 55625048-55625048 on Build GRCh38
rs926056759 Chr.X: 55625315-55625315 on Build GRCh38
rs867959911 Chr.X: 55625054-55625054 on Build GRCh38
rs73489626 Chr.X: 55625311-55625311 on Build GRCh38
rs764020624 Chr.X: 55624935-55624935 on Build GRCh38
rs1017018882 Chr.X: 55625117-55625117 on Build GRCh38

View Full Product Details