Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 57137175-57137675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAAH2
Entrez Gene ID
158584
Gene Name
fatty acid amide hydrolase 2
Gene Aliases
AMDD
UniGene
Hs.496205
Gene Chromosome Location
Chr.X: 57121861-57489196 on Build GRCh38
Transcripts
XM_011530767.2 NM_019003.3 XM_005262019.3 XM_017029598.1 XM_017029599.1

Gene Symbol
SPIN2A
Entrez Gene ID
54466
Gene Name
spindlin family member 2A
Gene Aliases
DXF34, SPIN2, TDRD25, dJ323P24.1
UniGene
Hs.460725
Gene Chromosome Location
Chr.X: 57134259-57138688 on Build GRCh38
Transcripts
XM_011530767.2 NM_019003.3 XM_005262019.3 XM_017029598.1 XM_017029599.1

SNP Detail

SNP ID Chromosome Location
rs150780723 Chr.X: 57137382-57137382 on Build GRCh38
rs1045814142 Chr.X: 57137497-57137497 on Build GRCh38
rs940598592 Chr.X: 57137279-57137279 on Build GRCh38
rs951783484 Chr.X: 57137439-57137439 on Build GRCh38
rs907573195 Chr.X: 57137278-57137278 on Build GRCh38
rs766695625 Chr.X: 57137313-57137313 on Build GRCh38
rs1050264840 Chr.X: 57137593-57137593 on Build GRCh38
rs934824381 Chr.X: 57137542-57137542 on Build GRCh38
rs923183002 Chr.X: 57137566-57137566 on Build GRCh38
rs1020285120 Chr.X: 57137406-57137406 on Build GRCh38
rs907544718 Chr.X: 57137470-57137470 on Build GRCh38
rs914436971 Chr.X: 57137595-57137595 on Build GRCh38
rs773469718 Chr.X: 57137257-57137257 on Build GRCh38
rs893243742 Chr.X: 57137380-57137380 on Build GRCh38
rs1041864922 Chr.X: 57137642-57137642 on Build GRCh38
rs1055011751 Chr.X: 57137621-57137621 on Build GRCh38
rs766813396 Chr.X: 57137247-57137247 on Build GRCh38
rs748443141 Chr.X: 57137228-57137228 on Build GRCh38
rs760359477 Chr.X: 57137618-57137618 on Build GRCh38
rs1035845643 Chr.X: 57137242-57137242 on Build GRCh38
rs762975059 Chr.X: 57137264-57137264 on Build GRCh38
rs903207067 Chr.X: 57137427-57137427 on Build GRCh38
rs901667640 Chr.X: 57137360-57137360 on Build GRCh38
rs930275057 Chr.X: 57137262-57137262 on Build GRCh38
rs767926064 Chr.X: 57137404-57137404 on Build GRCh38
rs1003125649 Chr.X: 57137431-57137431 on Build GRCh38
rs975989888 Chr.X: 57137517-57137517 on Build GRCh38
rs781562866 Chr.X: 57137225-57137225 on Build GRCh38
rs371005349 Chr.X: 57137212-57137212 on Build GRCh38
rs11091631 Chr.X: 57137492-57137492 on Build GRCh38
rs893330940 Chr.X: 57137639-57137639 on Build GRCh38
rs1023320105 Chr.X: 57137439-57137439 on Build GRCh38
rs903178199 Chr.X: 57137649-57137649 on Build GRCh38
rs1040730949 Chr.X: 57137315-57137315 on Build GRCh38
rs183579812 Chr.X: 57137365-57137365 on Build GRCh38
rs991621969 Chr.X: 57137438-57137438 on Build GRCh38
rs192199444 Chr.X: 57137248-57137248 on Build GRCh38
rs751930639 Chr.X: 57137456-57137456 on Build GRCh38
rs1050461548 Chr.X: 57137374-57137374 on Build GRCh38
rs111515245 Chr.X: 57137372-57137372 on Build GRCh38
rs903282198 Chr.X: 57137513-57137513 on Build GRCh38
rs984527142 Chr.X: 57137458-57137458 on Build GRCh38
rs931809781 Chr.X: 57137586-57137586 on Build GRCh38
rs755496102 Chr.X: 57137488-57137488 on Build GRCh38
rs958870485 Chr.X: 57137435-57137435 on Build GRCh38
rs1036390025 Chr.X: 57137434-57137434 on Build GRCh38

View Full Product Details