Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 66597483-66597977 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDA2R
Entrez Gene ID
60401
Gene Name
ectodysplasin A2 receptor
Gene Aliases
EDA-A2R, EDAA2R, TNFRSF27, XEDAR
UniGene
Hs.302017
Gene Chromosome Location
Chr.X: 66594384-66639303 on Build GRCh38
Transcripts
NM_001199687.2 NM_001242310.1 NM_001324199.1 NM_001324201.1 NM_001324202.1 NM_001324204.1 NM_001324205.1 NM_001324206.1 NM_021783.4 NR_136726.1 NR_136727.1 XM_011531000.1 XM_011531001.1 XM_011531002.1 XM_011531003.2 XM_011531005.2 XM_011531006.2 XM_017029703.1 XM_017029704.1 XM_017029705.1 XM_017029706.1 XM_017029707.1 XM_017029708.1

SNP Detail

SNP ID Chromosome Location
rs962722253 Chr.X: 66597528-66597528 on Build GRCh38
rs1054677154 Chr.X: 66597857-66597857 on Build GRCh38
rs768305887 Chr.X: 66597653-66597654 on Build GRCh38
rs775539008 Chr.X: 66597851-66597851 on Build GRCh38
rs189490989 Chr.X: 66597709-66597709 on Build GRCh38
rs1048049111 Chr.X: 66597879-66597879 on Build GRCh38
rs775774109 Chr.X: 66597609-66597610 on Build GRCh38
rs193006688 Chr.X: 66597909-66597909 on Build GRCh38
rs945644771 Chr.X: 66597710-66597710 on Build GRCh38
rs113456229 Chr.X: 66597871-66597871 on Build GRCh38
rs975382427 Chr.X: 66597530-66597530 on Build GRCh38
rs1023690067 Chr.X: 66597586-66597586 on Build GRCh38
rs766934065 Chr.X: 66597892-66597892 on Build GRCh38
rs1011948860 Chr.X: 66597515-66597515 on Build GRCh38
rs4827380 Chr.X: 66597898-66597898 on Build GRCh38
rs763482978 Chr.X: 66597651-66597651 on Build GRCh38
rs773251908 Chr.X: 66597617-66597617 on Build GRCh38
rs975440752 Chr.X: 66597872-66597872 on Build GRCh38
rs766286706 Chr.X: 66597920-66597920 on Build GRCh38
rs1047372270 Chr.X: 66597923-66597923 on Build GRCh38
rs970673776 Chr.X: 66597587-66597587 on Build GRCh38
rs766544215 Chr.X: 66597794-66597794 on Build GRCh38
rs964585046 Chr.X: 66597921-66597921 on Build GRCh38
rs1038540204 Chr.X: 66597954-66597954 on Build GRCh38
rs934089182 Chr.X: 66597610-66597611 on Build GRCh38
rs12855916 Chr.X: 66597680-66597680 on Build GRCh38
rs767481975 Chr.X: 66597589-66597589 on Build GRCh38
rs989257059 Chr.X: 66597788-66597788 on Build GRCh38
rs772536506 Chr.X: 66597652-66597653 on Build GRCh38
rs896262232 Chr.X: 66597862-66597862 on Build GRCh38
rs765511051 Chr.X: 66597949-66597949 on Build GRCh38
rs1006034522 Chr.X: 66597918-66597918 on Build GRCh38
rs914062164 Chr.X: 66597637-66597637 on Build GRCh38
rs908775888 Chr.X: 66597931-66597931 on Build GRCh38
rs1043976119 Chr.X: 66597787-66597787 on Build GRCh38
rs796146247 Chr.X: 66597583-66597583 on Build GRCh38

View Full Product Details