Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 67545979-67546500 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AR
Entrez Gene ID
367
Gene Name
androgen receptor
Gene Aliases
AIS, AR8, DHTR, HUMARA, HYSP1, KD, NR3C4, SBMA, SMAX1, TFM
UniGene
Hs.76704
Gene Chromosome Location
Chr.X: 67544032-67730619 on Build GRCh38
Transcripts
NM_000044.3

SNP Detail

SNP ID Chromosome Location
rs774171864 Chr.X: 67546042-67546042 on Build GRCh38
rs761215870 Chr.X: 67546365-67546365 on Build GRCh38
rs745333161 Chr.X: 67546030-67546030 on Build GRCh38
rs748457992 Chr.X: 67546447-67546447 on Build GRCh38
rs764690821 Chr.X: 67546373-67546373 on Build GRCh38
rs747067108 Chr.X: 67546219-67546219 on Build GRCh38
rs759045198 Chr.X: 67546288-67546288 on Build GRCh38
rs758022551 Chr.X: 67546295-67546295 on Build GRCh38
rs746981310 Chr.X: 67546186-67546186 on Build GRCh38
rs770988211 Chr.X: 67546231-67546231 on Build GRCh38
rs145233858 Chr.X: 67546187-67546187 on Build GRCh38
rs772490323 Chr.X: 67546354-67546354 on Build GRCh38
rs372682890 Chr.X: 67546126-67546126 on Build GRCh38
rs201934623 Chr.X: 67546320-67546320 on Build GRCh38
rs763834817 Chr.X: 67546297-67546297 on Build GRCh38
rs755088348 Chr.X: 67546434-67546434 on Build GRCh38
rs759882189 Chr.X: 67546120-67546120 on Build GRCh38
rs771949945 Chr.X: 67546060-67546060 on Build GRCh38
rs764432915 Chr.X: 67546215-67546215 on Build GRCh38
rs748391948 Chr.X: 67546352-67546352 on Build GRCh38
rs1035114992 Chr.X: 67546357-67546357 on Build GRCh38
rs867052370 Chr.X: 67546337-67546337 on Build GRCh38
rs760745000 Chr.X: 67546092-67546092 on Build GRCh38
rs776887144 Chr.X: 67546196-67546196 on Build GRCh38
rs781297287 Chr.X: 67546263-67546263 on Build GRCh38
rs113983339 Chr.X: 67546175-67546175 on Build GRCh38
rs374264537 Chr.X: 67546005-67546005 on Build GRCh38
rs773996740 Chr.X: 67546321-67546321 on Build GRCh38
rs778172639 Chr.X: 67546016-67546016 on Build GRCh38
rs571209658 Chr.X: 67546273-67546273 on Build GRCh38
rs767478793 Chr.X: 67546408-67546408 on Build GRCh38
rs778472979 Chr.X: 67546236-67546236 on Build GRCh38
rs988768657 Chr.X: 67546100-67546100 on Build GRCh38
rs377260719 Chr.X: 67546189-67546189 on Build GRCh38
rs138454018 Chr.X: 67546171-67546171 on Build GRCh38
rs773692090 Chr.X: 67546358-67546358 on Build GRCh38
rs779789625 Chr.X: 67546141-67546141 on Build GRCh38
rs533784386 Chr.X: 67546172-67546172 on Build GRCh38
rs749268474 Chr.X: 67546426-67546426 on Build GRCh38
rs968098233 Chr.X: 67546162-67546162 on Build GRCh38
rs777765515 Chr.X: 67546182-67546182 on Build GRCh38
rs774540165 Chr.X: 67546280-67546280 on Build GRCh38
rs767010987 Chr.X: 67546389-67546389 on Build GRCh38
rs960882208 Chr.X: 67546084-67546084 on Build GRCh38
rs746929441 Chr.X: 67546261-67546261 on Build GRCh38
rs769033307 Chr.X: 67546275-67546275 on Build GRCh38
rs780162524 Chr.X: 67546417-67546417 on Build GRCh38
rs145820446 Chr.X: 67546160-67546160 on Build GRCh38
rs776903759 Chr.X: 67546101-67546101 on Build GRCh38
rs765750147 Chr.X: 67546001-67546001 on Build GRCh38
rs956270286 Chr.X: 67546012-67546012 on Build GRCh38
rs755644981 Chr.X: 67546161-67546161 on Build GRCh38
rs753297907 Chr.X: 67546222-67546222 on Build GRCh38
rs751260108 Chr.X: 67546298-67546298 on Build GRCh38
rs866450936 Chr.X: 67546351-67546351 on Build GRCh38
rs548206951 Chr.X: 67546132-67546132 on Build GRCh38
rs781574525 Chr.X: 67546240-67546240 on Build GRCh38
rs773716161 Chr.X: 67546287-67546287 on Build GRCh38
rs763430523 Chr.X: 67546133-67546133 on Build GRCh38
rs779690243 Chr.X: 67546034-67546034 on Build GRCh38
rs555361360 Chr.X: 67546303-67546303 on Build GRCh38
rs1037339401 Chr.X: 67546289-67546289 on Build GRCh38
rs777482882 Chr.X: 67546252-67546252 on Build GRCh38
rs199522810 Chr.X: 67546167-67546167 on Build GRCh38
rs367604031 Chr.X: 67546209-67546209 on Build GRCh38
rs762267471 Chr.X: 67546220-67546220 on Build GRCh38
rs773040093 Chr.X: 67546065-67546065 on Build GRCh38
rs202150225 Chr.X: 67546332-67546332 on Build GRCh38
rs368729543 Chr.X: 67546469-67546469 on Build GRCh38
rs939465023 Chr.X: 67546371-67546371 on Build GRCh38
rs752328862 Chr.X: 67546292-67546292 on Build GRCh38
rs750364682 Chr.X: 67546410-67546410 on Build GRCh38
rs745883223 Chr.X: 67546271-67546271 on Build GRCh38
rs772400835 Chr.X: 67546471-67546471 on Build GRCh38
rs746338191 Chr.X: 67546248-67546248 on Build GRCh38
rs765067739 Chr.X: 67546269-67546277 on Build GRCh38
rs753457990 Chr.X: 67546170-67546170 on Build GRCh38
rs763341870 Chr.X: 67546212-67546212 on Build GRCh38
rs773170325 Chr.X: 67546412-67546412 on Build GRCh38
rs759716252 Chr.X: 67546208-67546208 on Build GRCh38
rs750324117 Chr.X: 67546011-67546011 on Build GRCh38
rs747970053 Chr.X: 67546046-67546046 on Build GRCh38
rs756250421 Chr.X: 67546309-67546309 on Build GRCh38
rs752012251 Chr.X: 67546158-67546158 on Build GRCh38
rs368345903 Chr.X: 67546103-67546103 on Build GRCh38
rs200510049 Chr.X: 67546285-67546285 on Build GRCh38
rs778996505 Chr.X: 67546439-67546439 on Build GRCh38
rs755590843 Chr.X: 67546031-67546031 on Build GRCh38
rs979157286 Chr.X: 67546163-67546163 on Build GRCh38
rs147994089 Chr.X: 67546250-67546250 on Build GRCh38
rs764789232 Chr.X: 67546290-67546290 on Build GRCh38
rs867505965 Chr.X: 67546342-67546342 on Build GRCh38
rs770987448 Chr.X: 67546098-67546098 on Build GRCh38
rs756999259 Chr.X: 67546405-67546405 on Build GRCh38
rs762262181 Chr.X: 67546283-67546283 on Build GRCh38
rs1021778539 Chr.X: 67546096-67546096 on Build GRCh38
rs774826391 Chr.X: 67546380-67546380 on Build GRCh38
rs200067740 Chr.X: 67546299-67546299 on Build GRCh38
rs774645775 Chr.X: 67546341-67546341 on Build GRCh38
rs201038724 Chr.X: 67546340-67546340 on Build GRCh38
rs749006575 Chr.X: 67546035-67546035 on Build GRCh38
rs1021234843 Chr.X: 67546113-67546113 on Build GRCh38
rs771770717 Chr.X: 67546177-67546177 on Build GRCh38
rs1001530027 Chr.X: 67546456-67546456 on Build GRCh38

View Full Product Details