Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 68829245-68829735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFNB1
Entrez Gene ID
1947
Gene Name
ephrin B1
Gene Aliases
CFND, CFNS, EFB1, EFL3, EPLG2, Elk-L, LERK2
UniGene
Hs.144700
Gene Chromosome Location
Chr.X: 68828997-68842164 on Build GRCh38
Transcripts
NM_004429.4

SNP Detail

SNP ID Chromosome Location
rs939985992 Chr.X: 68829716-68829716 on Build GRCh38
rs1035372569 Chr.X: 68829471-68829471 on Build GRCh38
rs969981767 Chr.X: 68829439-68829439 on Build GRCh38
rs1004268110 Chr.X: 68829318-68829318 on Build GRCh38
rs139698751 Chr.X: 68829548-68829548 on Build GRCh38
rs898377370 Chr.X: 68829634-68829634 on Build GRCh38
rs1045624352 Chr.X: 68829580-68829580 on Build GRCh38
rs1012656071 Chr.X: 68829579-68829579 on Build GRCh38
rs781366089 Chr.X: 68829338-68829338 on Build GRCh38
rs774575536 Chr.X: 68829563-68829563 on Build GRCh38
rs935700347 Chr.X: 68829472-68829472 on Build GRCh38
rs748040560 Chr.X: 68829585-68829585 on Build GRCh38
rs184030244 Chr.X: 68829269-68829269 on Build GRCh38
rs932937496 Chr.X: 68829609-68829609 on Build GRCh38
rs1023332104 Chr.X: 68829402-68829402 on Build GRCh38
rs912572510 Chr.X: 68829666-68829666 on Build GRCh38
rs1015528229 Chr.X: 68829608-68829608 on Build GRCh38
rs1028261378 Chr.X: 68829652-68829652 on Build GRCh38
rs968242566 Chr.X: 68829538-68829538 on Build GRCh38
rs780296718 Chr.X: 68829508-68829508 on Build GRCh38
rs962743262 Chr.X: 68829297-68829297 on Build GRCh38
rs1015891281 Chr.X: 68829294-68829294 on Build GRCh38
rs756470369 Chr.X: 68829498-68829498 on Build GRCh38
rs945653200 Chr.X: 68829411-68829411 on Build GRCh38
rs1004412683 Chr.X: 68829586-68829586 on Build GRCh38
rs1051085043 Chr.X: 68829301-68829301 on Build GRCh38
rs1054501922 Chr.X: 68829489-68829489 on Build GRCh38
rs1037044482 Chr.X: 68829391-68829391 on Build GRCh38
rs986839033 Chr.X: 68829677-68829677 on Build GRCh38
rs914324008 Chr.X: 68829536-68829536 on Build GRCh38
rs891103381 Chr.X: 68829315-68829315 on Build GRCh38
rs187341605 Chr.X: 68829392-68829392 on Build GRCh38
rs773063048 Chr.X: 68829643-68829643 on Build GRCh38
rs772069321 Chr.X: 68829597-68829597 on Build GRCh38
rs760496981 Chr.X: 68829654-68829654 on Build GRCh38
rs1003177738 Chr.X: 68829443-68829443 on Build GRCh38
rs912728013 Chr.X: 68829408-68829408 on Build GRCh38
rs921538487 Chr.X: 68829598-68829598 on Build GRCh38
rs995291342 Chr.X: 68829393-68829393 on Build GRCh38
rs768850247 Chr.X: 68829562-68829562 on Build GRCh38

View Full Product Details