Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70235878-70236378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AWAT1
Entrez Gene ID
158833
Gene Name
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases
DGA2, DGAT2L3
UniGene
Hs.407641
Gene Chromosome Location
Chr.X: 70233489-70240661 on Build GRCh38
Transcripts
NM_001013579.2 XM_017029295.1

SNP Detail

SNP ID Chromosome Location
rs1009328138 Chr.X: 70236299-70236299 on Build GRCh38
rs200977849 Chr.X: 70236081-70236081 on Build GRCh38
rs773913085 Chr.X: 70236078-70236078 on Build GRCh38
rs1030335688 Chr.X: 70236214-70236214 on Build GRCh38
rs753977336 Chr.X: 70236159-70236159 on Build GRCh38
rs968352178 Chr.X: 70236336-70236336 on Build GRCh38
rs745516238 Chr.X: 70236235-70236235 on Build GRCh38
rs761665299 Chr.X: 70236130-70236130 on Build GRCh38
rs774104029 Chr.X: 70236126-70236126 on Build GRCh38
rs756086018 Chr.X: 70236147-70236147 on Build GRCh38
rs967754322 Chr.X: 70235975-70235975 on Build GRCh38
rs979186657 Chr.X: 70236163-70236163 on Build GRCh38
rs988023072 Chr.X: 70235970-70235970 on Build GRCh38
rs965639074 Chr.X: 70236110-70236110 on Build GRCh38
rs139444456 Chr.X: 70235971-70235971 on Build GRCh38
rs866032694 Chr.X: 70236131-70236131 on Build GRCh38
rs955323335 Chr.X: 70235947-70235947 on Build GRCh38
rs774998010 Chr.X: 70236347-70236347 on Build GRCh38
rs767429250 Chr.X: 70236143-70236143 on Build GRCh38
rs748561802 Chr.X: 70236073-70236073 on Build GRCh38
rs951363799 Chr.X: 70236017-70236017 on Build GRCh38
rs758588341 Chr.X: 70236010-70236010 on Build GRCh38
rs182294660 Chr.X: 70235969-70235969 on Build GRCh38
rs200739971 Chr.X: 70236108-70236108 on Build GRCh38
rs377034950 Chr.X: 70236011-70236011 on Build GRCh38
rs779342837 Chr.X: 70236164-70236164 on Build GRCh38
rs375457191 Chr.X: 70236144-70236144 on Build GRCh38
rs748506798 Chr.X: 70236167-70236167 on Build GRCh38
rs375778100 Chr.X: 70236189-70236189 on Build GRCh38
rs1045269917 Chr.X: 70236335-70236335 on Build GRCh38
rs375099042 Chr.X: 70236020-70236020 on Build GRCh38
rs182767473 Chr.X: 70236324-70236324 on Build GRCh38
rs768955308 Chr.X: 70236054-70236054 on Build GRCh38
rs185484224 Chr.X: 70236082-70236082 on Build GRCh38
rs200111885 Chr.X: 70236077-70236077 on Build GRCh38
rs750500757 Chr.X: 70236021-70236021 on Build GRCh38
rs763983436 Chr.X: 70236157-70236157 on Build GRCh38
rs750082950 Chr.X: 70236051-70236053 on Build GRCh38
rs751551827 Chr.X: 70236165-70236166 on Build GRCh38
rs763854973 Chr.X: 70236123-70236123 on Build GRCh38
rs4314814 Chr.X: 70236068-70236068 on Build GRCh38
rs756140842 Chr.X: 70236181-70236181 on Build GRCh38
rs376144288 Chr.X: 70236216-70236216 on Build GRCh38
rs763915506 Chr.X: 70236140-70236143 on Build GRCh38
rs779373625 Chr.X: 70236061-70236061 on Build GRCh38
rs190171806 Chr.X: 70236101-70236101 on Build GRCh38
rs372091173 Chr.X: 70236139-70236139 on Build GRCh38
rs934977511 Chr.X: 70236206-70236206 on Build GRCh38
rs111373170 Chr.X: 70236128-70236128 on Build GRCh38
rs748626244 Chr.X: 70236155-70236155 on Build GRCh38
rs953224620 Chr.X: 70236162-70236162 on Build GRCh38
rs756210452 Chr.X: 70236033-70236033 on Build GRCh38
rs749404580 Chr.X: 70236052-70236052 on Build GRCh38
rs780196274 Chr.X: 70236035-70236035 on Build GRCh38
rs762031058 Chr.X: 70235931-70235931 on Build GRCh38
rs73634825 Chr.X: 70235929-70235929 on Build GRCh38

View Full Product Details