Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70426283-70426788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDPD2
Entrez Gene ID
54857
Gene Name
glycerophosphodiester phosphodiesterase domain containing 2
Gene Aliases
GDE3, OBDPF
UniGene
Hs.438712
Gene Chromosome Location
Chr.X: 70423031-70433391 on Build GRCh38
Transcripts
NM_001171191.1 NM_001171192.1 NM_001171193.1 NM_017711.3 XM_011530977.1 XM_017029614.1

SNP Detail

SNP ID Chromosome Location
rs112411662 Chr.X: 70426727-70426727 on Build GRCh38
rs780523750 Chr.X: 70426320-70426320 on Build GRCh38
rs113047584 Chr.X: 70426360-70426360 on Build GRCh38
rs898507845 Chr.X: 70426749-70426749 on Build GRCh38
rs745602670 Chr.X: 70426625-70426625 on Build GRCh38
rs1034019390 Chr.X: 70426621-70426621 on Build GRCh38
rs199835031 Chr.X: 70426718-70426718 on Build GRCh38
rs778456600 Chr.X: 70426362-70426363 on Build GRCh38
rs1050395619 Chr.X: 70426433-70426433 on Build GRCh38
rs769578225 Chr.X: 70426656-70426656 on Build GRCh38
rs1046578900 Chr.X: 70426560-70426560 on Build GRCh38
rs770450000 Chr.X: 70426347-70426347 on Build GRCh38
rs1018632698 Chr.X: 70426442-70426442 on Build GRCh38
rs369984051 Chr.X: 70426513-70426513 on Build GRCh38
rs777077368 Chr.X: 70426451-70426451 on Build GRCh38
rs775565226 Chr.X: 70426402-70426402 on Build GRCh38
rs775370757 Chr.X: 70426671-70426671 on Build GRCh38
rs769771497 Chr.X: 70426401-70426401 on Build GRCh38
rs200660900 Chr.X: 70426600-70426600 on Build GRCh38
rs762919987 Chr.X: 70426403-70426403 on Build GRCh38
rs762866982 Chr.X: 70426675-70426675 on Build GRCh38
rs779427335 Chr.X: 70426599-70426599 on Build GRCh38
rs1009762845 Chr.X: 70426558-70426558 on Build GRCh38
rs751645183 Chr.X: 70426448-70426448 on Build GRCh38
rs769155050 Chr.X: 70426335-70426335 on Build GRCh38
rs774392762 Chr.X: 70426681-70426681 on Build GRCh38
rs1020891780 Chr.X: 70426418-70426418 on Build GRCh38
rs906965267 Chr.X: 70426562-70426562 on Build GRCh38
rs375646779 Chr.X: 70426490-70426490 on Build GRCh38
rs754161536 Chr.X: 70426724-70426724 on Build GRCh38
rs749856799 Chr.X: 70426328-70426328 on Build GRCh38
rs372366721 Chr.X: 70426395-70426395 on Build GRCh38
rs374641666 Chr.X: 70426762-70426762 on Build GRCh38
rs755375130 Chr.X: 70426726-70426726 on Build GRCh38
rs750471316 Chr.X: 70426702-70426702 on Build GRCh38
rs201731012 Chr.X: 70426736-70426736 on Build GRCh38
rs764143616 Chr.X: 70426417-70426417 on Build GRCh38
rs764611851 Chr.X: 70426640-70426640 on Build GRCh38
rs776387322 Chr.X: 70426354-70426354 on Build GRCh38
rs770524254 Chr.X: 70426622-70426622 on Build GRCh38
rs923380080 Chr.X: 70426406-70426406 on Build GRCh38
rs770846352 Chr.X: 70426311-70426311 on Build GRCh38
rs367831079 Chr.X: 70426339-70426339 on Build GRCh38
rs909118378 Chr.X: 70426322-70426322 on Build GRCh38
rs200921949 Chr.X: 70426750-70426750 on Build GRCh38
rs147569276 Chr.X: 70426456-70426456 on Build GRCh38
rs990797016 Chr.X: 70426542-70426542 on Build GRCh38
rs775617807 Chr.X: 70426497-70426497 on Build GRCh38
rs892699218 Chr.X: 70426523-70426523 on Build GRCh38
rs768678443 Chr.X: 70426680-70426680 on Build GRCh38
rs766609273 Chr.X: 70426720-70426720 on Build GRCh38
rs745890557 Chr.X: 70426342-70426342 on Build GRCh38
rs778249845 Chr.X: 70426700-70426700 on Build GRCh38
rs756427350 Chr.X: 70426487-70426487 on Build GRCh38
rs766853311 Chr.X: 70426363-70426363 on Build GRCh38
rs761987096 Chr.X: 70426455-70426455 on Build GRCh38
rs767700057 Chr.X: 70426693-70426693 on Build GRCh38

View Full Product Details