Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 148003195-148003705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMR1NB
Entrez Gene ID
158521
Gene Name
fragile X mental retardation 1 neighbor
Gene Aliases
CT37, NY-SAR-35, NYSAR35
UniGene
Hs.128580
Gene Chromosome Location
Chr.X: 147981329-148026667 on Build GRCh38
Transcripts
NM_152578.2

SNP Detail

SNP ID Chromosome Location
rs781835958 Chr.X: 148003238-148003238 on Build GRCh38
rs782113740 Chr.X: 148003573-148003573 on Build GRCh38
rs782753814 Chr.X: 148003231-148003231 on Build GRCh38
rs782029560 Chr.X: 148003343-148003343 on Build GRCh38
rs1031041013 Chr.X: 148003429-148003429 on Build GRCh38
rs912449926 Chr.X: 148003622-148003622 on Build GRCh38
rs781888123 Chr.X: 148003252-148003252 on Build GRCh38
rs782377539 Chr.X: 148003352-148003352 on Build GRCh38
rs782370235 Chr.X: 148003308-148003308 on Build GRCh38
rs782095078 Chr.X: 148003372-148003372 on Build GRCh38
rs782514241 Chr.X: 148003585-148003585 on Build GRCh38
rs781815024 Chr.X: 148003221-148003221 on Build GRCh38
rs1000166641 Chr.X: 148003420-148003420 on Build GRCh38
rs191709453 Chr.X: 148003599-148003599 on Build GRCh38
rs919744367 Chr.X: 148003570-148003570 on Build GRCh38
rs782172100 Chr.X: 148003286-148003286 on Build GRCh38
rs1039363236 Chr.X: 148003629-148003629 on Build GRCh38
rs782571623 Chr.X: 148003242-148003242 on Build GRCh38
rs782322650 Chr.X: 148003669-148003669 on Build GRCh38
rs183434361 Chr.X: 148003630-148003630 on Build GRCh38
rs921029849 Chr.X: 148003637-148003637 on Build GRCh38
rs782306158 Chr.X: 148003338-148003338 on Build GRCh38
rs782204003 Chr.X: 148003333-148003333 on Build GRCh38
rs969420671 Chr.X: 148003522-148003522 on Build GRCh38
rs782246735 Chr.X: 148003258-148003258 on Build GRCh38
rs188827810 Chr.X: 148003239-148003239 on Build GRCh38
rs1022444331 Chr.X: 148003339-148003339 on Build GRCh38
rs781819089 Chr.X: 148003611-148003611 on Build GRCh38
rs782272481 Chr.X: 148003344-148003344 on Build GRCh38
rs782004549 Chr.X: 148003491-148003491 on Build GRCh38
rs904134690 Chr.X: 148003402-148003402 on Build GRCh38
rs1023675222 Chr.X: 148003518-148003518 on Build GRCh38
rs5904833 Chr.X: 148003628-148003628 on Build GRCh38
rs782598084 Chr.X: 148003314-148003314 on Build GRCh38
rs996364898 Chr.X: 148003235-148003235 on Build GRCh38
rs781984378 Chr.X: 148003362-148003362 on Build GRCh38
rs781907275 Chr.X: 148003248-148003248 on Build GRCh38
rs782441442 Chr.X: 148003229-148003229 on Build GRCh38
rs782803815 Chr.X: 148003454-148003454 on Build GRCh38
rs782724893 Chr.X: 148003337-148003337 on Build GRCh38
rs973888137 Chr.X: 148003563-148003563 on Build GRCh38

View Full Product Details